Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37699312-37699818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF607
Entrez Gene ID
84775
Gene Name
zinc finger protein 607
Gene Aliases
-
UniGene
Hs.116622
Gene Chromosome Location
Chr.19: 37696363-37719790 on Build GRCh38
Transcripts
NM_001172677.1 NM_032689.4 XM_006723435.3 XM_006723436.2 XM_006723437.2 XM_006723440.1 XM_011527414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4077615 c.811C>T p.H271Y Chr.19: 37699320-37699320 on Build GRCh38
5748584 c.329delG p.G110fs*415 Chr.19: 37699802-37699802 on Build GRCh38
566014 c.317G>A p.R106K Chr.19: 37699814-37699814 on Build GRCh38
995816 c.755C>T p.P252L Chr.19: 37699376-37699376 on Build GRCh38
4077619 c.420A>G p.E140E Chr.19: 37699711-37699711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748055161 Chr.19: 37699762-37699762 on Build GRCh38
rs757826211 Chr.19: 37699369-37699369 on Build GRCh38
rs747088664 Chr.19: 37699553-37699553 on Build GRCh38
rs778557842 Chr.19: 37699721-37699721 on Build GRCh38
rs750763507 Chr.19: 37699603-37699603 on Build GRCh38
rs139137709 Chr.19: 37699759-37699759 on Build GRCh38
rs921645200 Chr.19: 37699751-37699751 on Build GRCh38
rs777718238 Chr.19: 37699542-37699542 on Build GRCh38
rs746114049 Chr.19: 37699717-37699717 on Build GRCh38
rs779721597 Chr.19: 37699745-37699745 on Build GRCh38
rs35735839 Chr.19: 37699764-37699764 on Build GRCh38
rs977519497 Chr.19: 37699423-37699423 on Build GRCh38
rs761621901 Chr.19: 37699497-37699497 on Build GRCh38
rs199598374 Chr.19: 37699541-37699541 on Build GRCh38
rs757910712 Chr.19: 37699789-37699790 on Build GRCh38
rs762452134 Chr.19: 37699526-37699527 on Build GRCh38
rs762817055 Chr.19: 37699599-37699599 on Build GRCh38
rs771201625 Chr.19: 37699556-37699556 on Build GRCh38
rs146641168 Chr.19: 37699440-37699440 on Build GRCh38
rs1036174517 Chr.19: 37699439-37699439 on Build GRCh38
rs978097059 Chr.19: 37699652-37699652 on Build GRCh38
rs1011546607 Chr.19: 37699575-37699575 on Build GRCh38
rs781211384 Chr.19: 37699702-37699702 on Build GRCh38
rs990304726 Chr.19: 37699775-37699775 on Build GRCh38
rs769820971 Chr.19: 37699475-37699475 on Build GRCh38
rs758896182 Chr.19: 37699675-37699675 on Build GRCh38
rs745704035 Chr.19: 37699570-37699570 on Build GRCh38
rs866245354 Chr.19: 37699614-37699614 on Build GRCh38
rs752303193 Chr.19: 37699739-37699739 on Build GRCh38
rs767315321 Chr.19: 37699626-37699626 on Build GRCh38
rs753039543 Chr.19: 37699655-37699655 on Build GRCh38
rs753792754 Chr.19: 37699792-37699792 on Build GRCh38
rs112274112 Chr.19: 37699684-37699684 on Build GRCh38
rs991923165 Chr.19: 37699367-37699367 on Build GRCh38
rs779294627 Chr.19: 37699517-37699517 on Build GRCh38
rs755596287 Chr.19: 37699432-37699432 on Build GRCh38
rs967628291 Chr.19: 37699610-37699610 on Build GRCh38
rs775262661 Chr.19: 37699479-37699479 on Build GRCh38
rs539672039 Chr.19: 37699451-37699451 on Build GRCh38
rs759314986 Chr.19: 37699473-37699473 on Build GRCh38
rs751060098 Chr.19: 37699489-37699489 on Build GRCh38
rs756306028 Chr.19: 37699723-37699723 on Build GRCh38
rs774059598 Chr.19: 37699604-37699604 on Build GRCh38
rs763539688 Chr.19: 37699487-37699487 on Build GRCh38
rs776477357 Chr.19: 37699471-37699471 on Build GRCh38
rs757297863 Chr.19: 37699700-37699700 on Build GRCh38
rs769511723 Chr.19: 37699578-37699578 on Build GRCh38
rs749661817 Chr.19: 37699416-37699416 on Build GRCh38
rs756573598 Chr.19: 37699668-37699669 on Build GRCh38
rs373378298 Chr.19: 37699502-37699502 on Build GRCh38
rs750129424 Chr.19: 37699634-37699634 on Build GRCh38
rs370452757 Chr.19: 37699648-37699648 on Build GRCh38
rs1002946230 Chr.19: 37699446-37699446 on Build GRCh38
rs765531720 Chr.19: 37699635-37699635 on Build GRCh38
rs768575302 Chr.19: 37699600-37699600 on Build GRCh38
rs747244244 Chr.19: 37699456-37699456 on Build GRCh38
rs924789501 Chr.19: 37699426-37699426 on Build GRCh38
rs367890090 Chr.19: 37699585-37699585 on Build GRCh38
rs530017688 Chr.19: 37699791-37699791 on Build GRCh38
rs917328415 Chr.19: 37699415-37699415 on Build GRCh38
rs1024729702 Chr.19: 37699673-37699673 on Build GRCh38
rs759474501 Chr.19: 37699349-37699349 on Build GRCh38
rs762910706 Chr.19: 37699481-37699481 on Build GRCh38
rs566785304 Chr.19: 37699689-37699689 on Build GRCh38
rs775308801 Chr.19: 37699593-37699593 on Build GRCh38
rs781371851 Chr.19: 37699557-37699557 on Build GRCh38
rs924736994 Chr.19: 37699703-37699703 on Build GRCh38
rs767975665 Chr.19: 37699564-37699564 on Build GRCh38
rs199835706 Chr.19: 37699676-37699676 on Build GRCh38
rs35714505 Chr.19: 37699785-37699785 on Build GRCh38
rs959581159 Chr.19: 37699639-37699639 on Build GRCh38
rs777857755 Chr.19: 37699455-37699455 on Build GRCh38
rs894478479 Chr.19: 37699523-37699523 on Build GRCh38
rs751793603 Chr.19: 37699407-37699407 on Build GRCh38
rs749092790 Chr.19: 37699747-37699747 on Build GRCh38
rs768520576 Chr.19: 37699748-37699748 on Build GRCh38
rs1019765875 Chr.19: 37699658-37699658 on Build GRCh38
rs755263277 Chr.19: 37699513-37699513 on Build GRCh38
rs188971570 Chr.19: 37699397-37699397 on Build GRCh38
rs761380832 Chr.19: 37699611-37699611 on Build GRCh38
rs141287760 Chr.19: 37699406-37699406 on Build GRCh38
rs752312002 Chr.19: 37699357-37699357 on Build GRCh38
rs971470834 Chr.19: 37699683-37699683 on Build GRCh38
rs772788044 Chr.19: 37699777-37699777 on Build GRCh38
rs771075154 Chr.19: 37699467-37699467 on Build GRCh38
rs376028633 Chr.19: 37699503-37699503 on Build GRCh38
rs760426350 Chr.19: 37699491-37699491 on Build GRCh38
rs753164111 Chr.19: 37699530-37699530 on Build GRCh38
rs752105723 Chr.19: 37699492-37699495 on Build GRCh38
rs371183320 Chr.19: 37699758-37699758 on Build GRCh38
rs763850910 Chr.19: 37699373-37699373 on Build GRCh38
rs764140510 Chr.19: 37699357-37699365 on Build GRCh38

View Full Product Details