Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38801544-38802040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGALS4
Entrez Gene ID
3960
Gene Name
galectin 4
Gene Aliases
GAL4, L36LBP
UniGene
Hs.5302
Gene Chromosome Location
Chr.19: 38801671-38813544 on Build GRCh38
Transcripts
NM_006149.3 XM_011526973.1 XM_011526974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6301267 c.961G>A p.V321I Chr.19: 38801775-38801775 on Build GRCh38
3222975 c.892G>T p.A298S Chr.19: 38801844-38801844 on Build GRCh38
7298887 c.826-6C>A p.? Chr.19: 38801916-38801916 on Build GRCh38
109322 c.920T>G p.V307G Chr.19: 38801816-38801816 on Build GRCh38
3222980 c.811G>A p.G271R Chr.19: 38802006-38802006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140940108 Chr.19: 38801885-38801885 on Build GRCh38
rs988874147 Chr.19: 38801660-38801660 on Build GRCh38
rs773550046 Chr.19: 38801737-38801737 on Build GRCh38
rs753516631 Chr.19: 38801911-38801911 on Build GRCh38
rs770171438 Chr.19: 38801867-38801867 on Build GRCh38
rs768302906 Chr.19: 38801732-38801732 on Build GRCh38
rs180707929 Chr.19: 38801648-38801648 on Build GRCh38
rs138386804 Chr.19: 38801871-38801871 on Build GRCh38
rs771531951 Chr.19: 38801856-38801856 on Build GRCh38
rs199674948 Chr.19: 38801929-38801929 on Build GRCh38
rs555313734 Chr.19: 38801745-38801745 on Build GRCh38
rs765202455 Chr.19: 38801585-38801585 on Build GRCh38
rs780764230 Chr.19: 38802006-38802006 on Build GRCh38
rs759669904 Chr.19: 38801768-38801768 on Build GRCh38
rs369813600 Chr.19: 38801947-38801947 on Build GRCh38
rs764882551 Chr.19: 38802018-38802018 on Build GRCh38
rs911250049 Chr.19: 38801698-38801698 on Build GRCh38
rs148766456 Chr.19: 38801816-38801816 on Build GRCh38
rs922655324 Chr.19: 38801753-38801753 on Build GRCh38
rs764192527 Chr.19: 38801791-38801791 on Build GRCh38
rs941520397 Chr.19: 38801837-38801837 on Build GRCh38
rs755977666 Chr.19: 38801830-38801830 on Build GRCh38
rs199814280 Chr.19: 38801831-38801831 on Build GRCh38
rs879253533 Chr.19: 38801991-38801991 on Build GRCh38
rs865975578 Chr.19: 38801702-38801702 on Build GRCh38
rs774441106 Chr.19: 38801988-38801988 on Build GRCh38
rs144672000 Chr.19: 38802007-38802007 on Build GRCh38
rs1011981059 Chr.19: 38801641-38801641 on Build GRCh38
rs572457509 Chr.19: 38802003-38802003 on Build GRCh38
rs370544665 Chr.19: 38801967-38801967 on Build GRCh38
rs113477563 Chr.19: 38801678-38801678 on Build GRCh38
rs756910152 Chr.19: 38801872-38801872 on Build GRCh38
rs748749809 Chr.19: 38801730-38801730 on Build GRCh38
rs914639169 Chr.19: 38801664-38801664 on Build GRCh38
rs768992038 Chr.19: 38801976-38801976 on Build GRCh38
rs775884503 Chr.19: 38801957-38801957 on Build GRCh38
rs896786667 Chr.19: 38801595-38801595 on Build GRCh38
rs759407296 Chr.19: 38802012-38802012 on Build GRCh38
rs761895109 Chr.19: 38801992-38801992 on Build GRCh38
rs767323404 Chr.19: 38801760-38801764 on Build GRCh38
rs551176335 Chr.19: 38801606-38801606 on Build GRCh38
rs757855126 Chr.19: 38801933-38801933 on Build GRCh38
rs761654023 Chr.19: 38801886-38801886 on Build GRCh38
rs970269044 Chr.19: 38801630-38801630 on Build GRCh38
rs1049745796 Chr.19: 38801689-38801689 on Build GRCh38
rs374683316 Chr.19: 38801712-38801712 on Build GRCh38
rs910014012 Chr.19: 38801836-38801836 on Build GRCh38
rs757270236 Chr.19: 38801800-38801800 on Build GRCh38
rs201661817 Chr.19: 38801835-38801835 on Build GRCh38
rs745488080 Chr.19: 38801723-38801723 on Build GRCh38
rs767421979 Chr.19: 38801900-38801900 on Build GRCh38
rs878876745 Chr.19: 38801914-38801914 on Build GRCh38
rs767203886 Chr.19: 38801809-38801809 on Build GRCh38
rs1048261447 Chr.19: 38801851-38801852 on Build GRCh38
rs772075044 Chr.19: 38802000-38802000 on Build GRCh38
rs879001172 Chr.19: 38801913-38801913 on Build GRCh38
rs775706738 Chr.19: 38801772-38801772 on Build GRCh38
rs777741497 Chr.19: 38801838-38801838 on Build GRCh38
rs755932424 Chr.19: 38801904-38801904 on Build GRCh38
rs766315604 Chr.19: 38801905-38801905 on Build GRCh38
rs989378299 Chr.19: 38801975-38801976 on Build GRCh38
rs116200669 Chr.19: 38801726-38801726 on Build GRCh38
rs748978981 Chr.19: 38801833-38801833 on Build GRCh38
rs751423521 Chr.19: 38801796-38801796 on Build GRCh38
rs981680196 Chr.19: 38801639-38801639 on Build GRCh38
rs755749994 Chr.19: 38801972-38801972 on Build GRCh38
rs956040331 Chr.19: 38801739-38801739 on Build GRCh38
rs1003634716 Chr.19: 38801603-38801603 on Build GRCh38
rs752695459 Chr.19: 38801750-38801750 on Build GRCh38
rs747456110 Chr.19: 38801844-38801844 on Build GRCh38
rs372425632 Chr.19: 38801604-38801604 on Build GRCh38
rs577813567 Chr.19: 38801901-38801901 on Build GRCh38
rs374092153 Chr.19: 38801863-38801863 on Build GRCh38
rs34820201 Chr.19: 38801645-38801646 on Build GRCh38
rs751351750 Chr.19: 38801870-38801870 on Build GRCh38
rs867854421 Chr.19: 38801721-38801721 on Build GRCh38
rs769629325 Chr.19: 38801725-38801725 on Build GRCh38
rs1030855537 Chr.19: 38802009-38802009 on Build GRCh38
rs901743126 Chr.19: 38802011-38802011 on Build GRCh38
rs1022080504 Chr.19: 38801647-38801647 on Build GRCh38
rs770116328 Chr.19: 38801955-38801955 on Build GRCh38
rs762975509 Chr.19: 38801785-38801785 on Build GRCh38
rs777005220 Chr.19: 38801859-38801859 on Build GRCh38
rs771175613 Chr.19: 38801749-38801749 on Build GRCh38
rs369408666 Chr.19: 38801769-38801769 on Build GRCh38
rs947529953 Chr.19: 38801669-38801669 on Build GRCh38
rs746408671 Chr.19: 38801949-38801949 on Build GRCh38
rs985583413 Chr.19: 38801828-38801828 on Build GRCh38
rs749918987 Chr.19: 38801940-38801940 on Build GRCh38
rs200693237 Chr.19: 38801915-38801915 on Build GRCh38
rs778279131 Chr.19: 38801919-38801919 on Build GRCh38
rs1043462644 Chr.19: 38801998-38801998 on Build GRCh38

View Full Product Details