Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39507740-39508252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL3
Entrez Gene ID
10683
Gene Name
delta like canonical Notch ligand 3
Gene Aliases
SCDO1
UniGene
Hs.127792
Gene Chromosome Location
Chr.19: 39498917-39508481 on Build GRCh38
Transcripts
NM_016941.3 NM_203486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7160521 c.1708G>A p.D570N Chr.19: 39507864-39507864 on Build GRCh38
1712271 c.1676C>T p.S559L Chr.19: 39507832-39507832 on Build GRCh38
3892539 c.1736C>T p.A579V Chr.19: 39507892-39507892 on Build GRCh38
4753375 c.1753C>T p.R585W Chr.19: 39507909-39507909 on Build GRCh38
5855741 c.1758+5C>T p.? Chr.19: 39507919-39507919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982368816 Chr.19: 39508129-39508129 on Build GRCh38
rs200183802 Chr.19: 39507962-39507962 on Build GRCh38
rs1001161913 Chr.19: 39508115-39508115 on Build GRCh38
rs778974437 Chr.19: 39507919-39507919 on Build GRCh38
rs746856386 Chr.19: 39508001-39508001 on Build GRCh38
rs765459212 Chr.19: 39508222-39508222 on Build GRCh38
rs772533182 Chr.19: 39508131-39508131 on Build GRCh38
rs749430803 Chr.19: 39507910-39507910 on Build GRCh38
rs769073913 Chr.19: 39507822-39507823 on Build GRCh38
rs766655092 Chr.19: 39508210-39508210 on Build GRCh38
rs908776991 Chr.19: 39507803-39507803 on Build GRCh38
rs145264173 Chr.19: 39507894-39507894 on Build GRCh38
rs543757649 Chr.19: 39507974-39507974 on Build GRCh38
rs940278862 Chr.19: 39507818-39507818 on Build GRCh38
rs367794721 Chr.19: 39508190-39508190 on Build GRCh38
rs371433358 Chr.19: 39507909-39507909 on Build GRCh38
rs1055205721 Chr.19: 39507856-39507856 on Build GRCh38
rs764651686 Chr.19: 39507969-39507969 on Build GRCh38
rs774889509 Chr.19: 39508010-39508010 on Build GRCh38
rs754172178 Chr.19: 39508214-39508214 on Build GRCh38
rs758226555 Chr.19: 39508054-39508056 on Build GRCh38
rs550561051 Chr.19: 39507936-39507936 on Build GRCh38
rs776323833 Chr.19: 39507932-39507932 on Build GRCh38
rs770611082 Chr.19: 39507935-39507935 on Build GRCh38
rs758363118 Chr.19: 39507994-39507994 on Build GRCh38
rs1042641489 Chr.19: 39507960-39507960 on Build GRCh38
rs1016194728 Chr.19: 39508091-39508091 on Build GRCh38
rs995833469 Chr.19: 39508014-39508014 on Build GRCh38
rs768002373 Chr.19: 39507907-39507907 on Build GRCh38
rs759531850 Chr.19: 39507961-39507962 on Build GRCh38
rs763249149 Chr.19: 39507799-39507800 on Build GRCh38
rs1005689624 Chr.19: 39508090-39508090 on Build GRCh38
rs761723380 Chr.19: 39507823-39507823 on Build GRCh38
rs750763232 Chr.19: 39507908-39507908 on Build GRCh38
rs140715276 Chr.19: 39508004-39508004 on Build GRCh38
rs759145657 Chr.19: 39507959-39507959 on Build GRCh38
rs868640234 Chr.19: 39507868-39507868 on Build GRCh38
rs569619393 Chr.19: 39508155-39508155 on Build GRCh38
rs539055025 Chr.19: 39507814-39507814 on Build GRCh38
rs942672323 Chr.19: 39507955-39507955 on Build GRCh38
rs959355073 Chr.19: 39508191-39508191 on Build GRCh38
rs749284798 Chr.19: 39507804-39507804 on Build GRCh38
rs147582946 Chr.19: 39507971-39507971 on Build GRCh38
rs763967582 Chr.19: 39508203-39508204 on Build GRCh38
rs1025925604 Chr.19: 39508170-39508170 on Build GRCh38
rs765968413 Chr.19: 39507850-39507850 on Build GRCh38
rs775821436 Chr.19: 39507792-39507793 on Build GRCh38
rs558083766 Chr.19: 39508226-39508227 on Build GRCh38
rs201994238 Chr.19: 39508028-39508028 on Build GRCh38
rs751999027 Chr.19: 39507900-39507900 on Build GRCh38
rs374551752 Chr.19: 39508056-39508056 on Build GRCh38
rs760398713 Chr.19: 39507927-39507928 on Build GRCh38
rs951910938 Chr.19: 39508050-39508050 on Build GRCh38
rs748176684 Chr.19: 39507989-39507989 on Build GRCh38
rs748080617 Chr.19: 39507921-39507921 on Build GRCh38
rs993856532 Chr.19: 39508141-39508141 on Build GRCh38
rs200669115 Chr.19: 39508221-39508221 on Build GRCh38
rs886054434 Chr.19: 39508090-39508090 on Build GRCh38
rs577591982 Chr.19: 39507837-39507837 on Build GRCh38
rs559109825 Chr.19: 39508168-39508168 on Build GRCh38
rs762786941 Chr.19: 39508199-39508199 on Build GRCh38
rs777658037 Chr.19: 39507995-39507995 on Build GRCh38
rs762517319 Chr.19: 39508012-39508012 on Build GRCh38
rs756522236 Chr.19: 39507982-39507982 on Build GRCh38
rs772701329 Chr.19: 39507904-39507905 on Build GRCh38
rs776415255 Chr.19: 39508003-39508003 on Build GRCh38
rs866809563 Chr.19: 39508193-39508193 on Build GRCh38
rs151052276 Chr.19: 39507975-39507975 on Build GRCh38
rs765903171 Chr.19: 39507986-39507986 on Build GRCh38
rs961581325 Chr.19: 39508127-39508127 on Build GRCh38
rs752685181 Chr.19: 39508017-39508017 on Build GRCh38
rs962449868 Chr.19: 39508130-39508130 on Build GRCh38
rs144011919 Chr.19: 39507885-39507885 on Build GRCh38
rs757766484 Chr.19: 39507905-39507905 on Build GRCh38
rs773012836 Chr.19: 39507932-39507932 on Build GRCh38
rs771849483 Chr.19: 39507832-39507832 on Build GRCh38
rs778871834 Chr.19: 39507988-39507988 on Build GRCh38
rs780299632 Chr.19: 39507782-39507782 on Build GRCh38
rs766679747 Chr.19: 39507986-39507986 on Build GRCh38
rs267605476 Chr.19: 39507926-39507926 on Build GRCh38
rs758939727 Chr.19: 39507891-39507891 on Build GRCh38
rs116099821 Chr.19: 39507820-39507820 on Build GRCh38
rs368772580 Chr.19: 39508005-39508005 on Build GRCh38
rs754000867 Chr.19: 39507987-39507987 on Build GRCh38
rs751705871 Chr.19: 39508208-39508214 on Build GRCh38
rs750855068 Chr.19: 39507977-39507977 on Build GRCh38
rs576748683 Chr.19: 39508101-39508101 on Build GRCh38
rs868518306 Chr.19: 39507874-39507874 on Build GRCh38
rs754997290 Chr.19: 39507916-39507916 on Build GRCh38
rs199745659 Chr.19: 39507941-39507941 on Build GRCh38
rs772924663 Chr.19: 39507833-39507833 on Build GRCh38
rs754576543 Chr.19: 39507839-39507839 on Build GRCh38
rs935373777 Chr.19: 39507981-39507981 on Build GRCh38
rs771937712 Chr.19: 39507923-39507923 on Build GRCh38
rs765785746 Chr.19: 39507931-39507932 on Build GRCh38
rs778339882 Chr.19: 39507934-39507934 on Build GRCh38
rs911245932 Chr.19: 39507938-39507938 on Build GRCh38

View Full Product Details