Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47438045-47438538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD2
Entrez Gene ID
2306
Gene Name
forkhead box D2
Gene Aliases
FKHL17, FREAC-9, FREAC9
UniGene
Hs.166188
Gene Chromosome Location
Chr.1: 47436017-47440691 on Build GRCh38
Transcripts
NM_004474.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392300 c.140C>A p.A47D Chr.1: 47438275-47438275 on Build GRCh38
6391059 c.323C>T p.A108V Chr.1: 47438458-47438458 on Build GRCh38
4144062 c.331G>A p.G111R Chr.1: 47438466-47438466 on Build GRCh38
4144061 c.193G>A p.A65T Chr.1: 47438328-47438328 on Build GRCh38
7415953 c.244C>A p.P82T Chr.1: 47438379-47438379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538848817 Chr.1: 47438246-47438246 on Build GRCh38
rs1022868233 Chr.1: 47438094-47438094 on Build GRCh38
rs534680288 Chr.1: 47438141-47438141 on Build GRCh38
rs78469326 Chr.1: 47438328-47438328 on Build GRCh38
rs766707957 Chr.1: 47438427-47438465 on Build GRCh38
rs751327040 Chr.1: 47438262-47438262 on Build GRCh38
rs763655499 Chr.1: 47438257-47438257 on Build GRCh38
rs974860463 Chr.1: 47438434-47438434 on Build GRCh38
rs112187678 Chr.1: 47438191-47438191 on Build GRCh38
rs749571018 Chr.1: 47438125-47438125 on Build GRCh38
rs1042256690 Chr.1: 47438260-47438260 on Build GRCh38
rs1025385729 Chr.1: 47438086-47438086 on Build GRCh38
rs964252467 Chr.1: 47438424-47438424 on Build GRCh38
rs1046887529 Chr.1: 47438507-47438507 on Build GRCh38
rs759089664 Chr.1: 47438241-47438241 on Build GRCh38
rs1018851303 Chr.1: 47438378-47438378 on Build GRCh38
rs866732679 Chr.1: 47438442-47438442 on Build GRCh38
rs779272089 Chr.1: 47438162-47438162 on Build GRCh38
rs778425099 Chr.1: 47438489-47438489 on Build GRCh38
rs758877986 Chr.1: 47438484-47438484 on Build GRCh38
rs1030101333 Chr.1: 47438435-47438435 on Build GRCh38
rs756174143 Chr.1: 47438453-47438453 on Build GRCh38
rs755190355 Chr.1: 47438471-47438471 on Build GRCh38
rs1025918121 Chr.1: 47438111-47438112 on Build GRCh38
rs936584544 Chr.1: 47438450-47438450 on Build GRCh38
rs866220574 Chr.1: 47438397-47438397 on Build GRCh38
rs953211571 Chr.1: 47438248-47438248 on Build GRCh38
rs773498235 Chr.1: 47438411-47438452 on Build GRCh38
rs969943066 Chr.1: 47438098-47438098 on Build GRCh38
rs994513706 Chr.1: 47438103-47438103 on Build GRCh38
rs756235492 Chr.1: 47438089-47438089 on Build GRCh38
rs561234231 Chr.1: 47438466-47438466 on Build GRCh38
rs145909683 Chr.1: 47438495-47438495 on Build GRCh38
rs1025680937 Chr.1: 47438359-47438359 on Build GRCh38
rs540678652 Chr.1: 47438497-47438497 on Build GRCh38
rs1005970122 Chr.1: 47438290-47438290 on Build GRCh38
rs1044114874 Chr.1: 47438073-47438073 on Build GRCh38
rs748495731 Chr.1: 47438182-47438182 on Build GRCh38
rs1054025619 Chr.1: 47438289-47438289 on Build GRCh38
rs1052972180 Chr.1: 47438081-47438081 on Build GRCh38
rs761533875 Chr.1: 47438271-47438271 on Build GRCh38
rs952924879 Chr.1: 47438362-47438362 on Build GRCh38
rs932991925 Chr.1: 47438249-47438249 on Build GRCh38
rs750452322 Chr.1: 47438087-47438087 on Build GRCh38
rs773004685 Chr.1: 47438510-47438510 on Build GRCh38
rs768774247 Chr.1: 47438142-47438142 on Build GRCh38
rs923846438 Chr.1: 47438266-47438266 on Build GRCh38
rs902599712 Chr.1: 47438076-47438076 on Build GRCh38
rs765573047 Chr.1: 47438112-47438114 on Build GRCh38
rs763229709 Chr.1: 47438422-47438461 on Build GRCh38
rs1014025667 Chr.1: 47438083-47438083 on Build GRCh38
rs999985459 Chr.1: 47438080-47438080 on Build GRCh38
rs946733181 Chr.1: 47438071-47438071 on Build GRCh38
rs895011041 Chr.1: 47438323-47438323 on Build GRCh38
rs780051451 Chr.1: 47438465-47438465 on Build GRCh38
rs767419027 Chr.1: 47438296-47438296 on Build GRCh38
rs922721593 Chr.1: 47438437-47438446 on Build GRCh38
rs543379385 Chr.1: 47438403-47438403 on Build GRCh38
rs761637448 Chr.1: 47438505-47438505 on Build GRCh38
rs779029024 Chr.1: 47438477-47438477 on Build GRCh38
rs964816097 Chr.1: 47438232-47438232 on Build GRCh38
rs895631777 Chr.1: 47438082-47438082 on Build GRCh38
rs578093538 Chr.1: 47438243-47438243 on Build GRCh38
rs748527474 Chr.1: 47438478-47438478 on Build GRCh38
rs747572734 Chr.1: 47438494-47438494 on Build GRCh38
rs1005849071 Chr.1: 47438375-47438375 on Build GRCh38
rs1012710003 Chr.1: 47438327-47438327 on Build GRCh38
rs944958034 Chr.1: 47438064-47438064 on Build GRCh38
rs780250177 Chr.1: 47438123-47438123 on Build GRCh38
rs554919952 Chr.1: 47438065-47438065 on Build GRCh38
rs552814879 Chr.1: 47438231-47438231 on Build GRCh38
rs575986501 Chr.1: 47438345-47438345 on Build GRCh38
rs994185602 Chr.1: 47438340-47438340 on Build GRCh38
rs772585833 Chr.1: 47438215-47438215 on Build GRCh38
rs900741724 Chr.1: 47438070-47438070 on Build GRCh38
rs977709123 Chr.1: 47438438-47438438 on Build GRCh38
rs773923786 Chr.1: 47438504-47438504 on Build GRCh38
rs911627019 Chr.1: 47438156-47438156 on Build GRCh38
rs751928135 Chr.1: 47438482-47438483 on Build GRCh38
rs566956123 Chr.1: 47438106-47438106 on Build GRCh38
rs565241171 Chr.1: 47438506-47438506 on Build GRCh38
rs906501697 Chr.1: 47438338-47438338 on Build GRCh38

View Full Product Details