Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51877153-51877552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF577
Entrez Gene ID
84765
Gene Name
zinc finger protein 577
Gene Aliases
-
UniGene
Hs.723015
Gene Chromosome Location
Chr.19: 51855803-51887976 on Build GRCh38
Transcripts
NM_001135590.1 NM_032679.2 NR_024181.1 XM_006723432.3 XM_006723433.2 XM_011527408.2 XM_017027387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4480444 c.241C>T p.P81S Chr.19: 51877324-51877324 on Build GRCh38
1582422 c.221A>C p.K74T Chr.19: 51877344-51877344 on Build GRCh38
5973119 c.259G>T p.A87S Chr.19: 51877306-51877306 on Build GRCh38
5390537 c.276C>T p.I92I Chr.19: 51877289-51877289 on Build GRCh38
7294881 c.196G>T p.G66C Chr.19: 51877369-51877369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549356287 Chr.19: 51877200-51877200 on Build GRCh38
rs767763946 Chr.19: 51877513-51877513 on Build GRCh38
rs910391038 Chr.19: 51877267-51877267 on Build GRCh38
rs3887892 Chr.19: 51877191-51877191 on Build GRCh38
rs746941003 Chr.19: 51877364-51877364 on Build GRCh38
rs28452854 Chr.19: 51877530-51877530 on Build GRCh38
rs115218037 Chr.19: 51877213-51877213 on Build GRCh38
rs772309230 Chr.19: 51877245-51877245 on Build GRCh38
rs757515159 Chr.19: 51877410-51877410 on Build GRCh38
rs967196635 Chr.19: 51877196-51877196 on Build GRCh38
rs780322262 Chr.19: 51877358-51877358 on Build GRCh38
rs79294328 Chr.19: 51877518-51877518 on Build GRCh38
rs745648813 Chr.19: 51877322-51877323 on Build GRCh38
rs3764540 Chr.19: 51877428-51877428 on Build GRCh38
rs750644592 Chr.19: 51877414-51877414 on Build GRCh38
rs769512704 Chr.19: 51877374-51877375 on Build GRCh38
rs766687180 Chr.19: 51877430-51877430 on Build GRCh38
rs1021074771 Chr.19: 51877187-51877187 on Build GRCh38
rs772539977 Chr.19: 51877352-51877352 on Build GRCh38
rs950514378 Chr.19: 51877492-51877492 on Build GRCh38
rs754404942 Chr.19: 51877409-51877409 on Build GRCh38
rs372353473 Chr.19: 51877370-51877370 on Build GRCh38
rs894055010 Chr.19: 51877345-51877346 on Build GRCh38
rs1017511945 Chr.19: 51877462-51877462 on Build GRCh38
rs973239506 Chr.19: 51877473-51877473 on Build GRCh38
rs768541629 Chr.19: 51877279-51877279 on Build GRCh38
rs111868808 Chr.19: 51877449-51877449 on Build GRCh38
rs757649074 Chr.19: 51877346-51877346 on Build GRCh38
rs963683782 Chr.19: 51877225-51877225 on Build GRCh38
rs531230781 Chr.19: 51877276-51877276 on Build GRCh38
rs370610684 Chr.19: 51877224-51877224 on Build GRCh38
rs562333356 Chr.19: 51877297-51877297 on Build GRCh38
rs534585840 Chr.19: 51877486-51877486 on Build GRCh38
rs753177361 Chr.19: 51877320-51877320 on Build GRCh38
rs137892890 Chr.19: 51877371-51877371 on Build GRCh38
rs149543099 Chr.19: 51877372-51877372 on Build GRCh38
rs760744736 Chr.19: 51877273-51877273 on Build GRCh38
rs150301895 Chr.19: 51877353-51877353 on Build GRCh38
rs778967750 Chr.19: 51877349-51877349 on Build GRCh38
rs1011004192 Chr.19: 51877175-51877175 on Build GRCh38
rs919055360 Chr.19: 51877475-51877475 on Build GRCh38
rs963245912 Chr.19: 51877466-51877466 on Build GRCh38
rs765683151 Chr.19: 51877413-51877413 on Build GRCh38
rs113934016 Chr.19: 51877403-51877403 on Build GRCh38
rs1046077116 Chr.19: 51877316-51877316 on Build GRCh38
rs17856123 Chr.19: 51877314-51877314 on Build GRCh38
rs769492914 Chr.19: 51877344-51877344 on Build GRCh38
rs143182302 Chr.19: 51877397-51877397 on Build GRCh38
rs756652358 Chr.19: 51877326-51877326 on Build GRCh38
rs761512637 Chr.19: 51877305-51877305 on Build GRCh38
rs982231346 Chr.19: 51877506-51877506 on Build GRCh38
rs775520173 Chr.19: 51877269-51877269 on Build GRCh38
rs372025227 Chr.19: 51877241-51877241 on Build GRCh38
rs1030065075 Chr.19: 51877222-51877222 on Build GRCh38
rs375998998 Chr.19: 51877312-51877312 on Build GRCh38
rs879238394 Chr.19: 51877236-51877236 on Build GRCh38
rs778189561 Chr.19: 51877343-51877343 on Build GRCh38
rs1002726110 Chr.19: 51877217-51877217 on Build GRCh38
rs548882617 Chr.19: 51877432-51877432 on Build GRCh38
rs200541521 Chr.19: 51877317-51877317 on Build GRCh38
rs780266673 Chr.19: 51877425-51877425 on Build GRCh38
rs544147587 Chr.19: 51877227-51877227 on Build GRCh38
rs753176681 Chr.19: 51877526-51877526 on Build GRCh38
rs937695335 Chr.19: 51877521-51877521 on Build GRCh38
rs375907992 Chr.19: 51877373-51877373 on Build GRCh38
rs764445353 Chr.19: 51877408-51877408 on Build GRCh38
rs182124583 Chr.19: 51877388-51877388 on Build GRCh38
rs137865319 Chr.19: 51877308-51877308 on Build GRCh38
rs759063455 Chr.19: 51877406-51877406 on Build GRCh38

View Full Product Details