Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53148600-53149115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF347
Entrez Gene ID
84671
Gene Name
zinc finger protein 347
Gene Aliases
ZNF1111
UniGene
Hs.467239
Gene Chromosome Location
Chr.19: 53134935-53159069 on Build GRCh38
Transcripts
NM_001172674.1 NM_001172675.1 NM_032584.2 XM_005259335.4 XM_005259336.4 XM_017027384.1 XM_017027385.1 XM_017027386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423778 c.143-2A>C p.? Chr.19: 53148811-53148811 on Build GRCh38
4624108 c.260C>T p.A87V Chr.19: 53148692-53148692 on Build GRCh38
7399001 c.143-2A>G p.? Chr.19: 53148811-53148811 on Build GRCh38
4154339 c.211A>T p.S71C Chr.19: 53148741-53148741 on Build GRCh38
6339921 c.271+10C>A p.? Chr.19: 53148671-53148671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528034886 Chr.19: 53148863-53148863 on Build GRCh38
rs766436408 Chr.19: 53148735-53148735 on Build GRCh38
rs10405853 Chr.19: 53148640-53148640 on Build GRCh38
rs1001928796 Chr.19: 53148896-53148896 on Build GRCh38
rs560757990 Chr.19: 53148783-53148783 on Build GRCh38
rs867821939 Chr.19: 53148781-53148781 on Build GRCh38
rs10406057 Chr.19: 53148650-53148650 on Build GRCh38
rs759818147 Chr.19: 53148742-53148756 on Build GRCh38
rs77768344 Chr.19: 53148919-53148919 on Build GRCh38
rs982825406 Chr.19: 53148981-53148981 on Build GRCh38
rs764480630 Chr.19: 53148699-53148699 on Build GRCh38
rs899205109 Chr.19: 53148894-53148894 on Build GRCh38
rs547232915 Chr.19: 53149058-53149058 on Build GRCh38
rs943848683 Chr.19: 53148668-53148668 on Build GRCh38
rs201137351 Chr.19: 53148864-53148864 on Build GRCh38
rs951452345 Chr.19: 53148973-53148973 on Build GRCh38
rs754190225 Chr.19: 53148724-53148724 on Build GRCh38
rs80326789 Chr.19: 53149017-53149017 on Build GRCh38
rs555606353 Chr.19: 53148638-53148638 on Build GRCh38
rs780452895 Chr.19: 53148925-53148925 on Build GRCh38
rs1053694887 Chr.19: 53149024-53149024 on Build GRCh38
rs762179257 Chr.19: 53148836-53148836 on Build GRCh38
rs1046042691 Chr.19: 53149078-53149078 on Build GRCh38
rs543465832 Chr.19: 53148732-53148732 on Build GRCh38
rs113887807 Chr.19: 53148898-53148898 on Build GRCh38
rs190209394 Chr.19: 53148767-53148767 on Build GRCh38
rs765748900 Chr.19: 53148680-53148680 on Build GRCh38
rs752762598 Chr.19: 53148809-53148809 on Build GRCh38
rs772660834 Chr.19: 53148665-53148665 on Build GRCh38
rs764169325 Chr.19: 53149006-53149006 on Build GRCh38
rs555522203 Chr.19: 53148747-53148747 on Build GRCh38
rs768694844 Chr.19: 53148768-53148768 on Build GRCh38
rs762463647 Chr.19: 53148795-53148795 on Build GRCh38
rs369881511 Chr.19: 53148759-53148759 on Build GRCh38
rs368137575 Chr.19: 53149020-53149020 on Build GRCh38
rs374326296 Chr.19: 53148821-53148821 on Build GRCh38
rs920230904 Chr.19: 53148944-53148944 on Build GRCh38
rs769641869 Chr.19: 53148782-53148782 on Build GRCh38
rs924791722 Chr.19: 53148726-53148726 on Build GRCh38
rs912570262 Chr.19: 53148996-53148996 on Build GRCh38
rs375618372 Chr.19: 53148696-53148696 on Build GRCh38
rs1039532081 Chr.19: 53148713-53148713 on Build GRCh38
rs765470320 Chr.19: 53148858-53148858 on Build GRCh38
rs756245035 Chr.19: 53148813-53148813 on Build GRCh38
rs970527922 Chr.19: 53148828-53148828 on Build GRCh38
rs753268866 Chr.19: 53148681-53148681 on Build GRCh38
rs974258863 Chr.19: 53148930-53148930 on Build GRCh38
rs987676516 Chr.19: 53148766-53148766 on Build GRCh38
rs765328473 Chr.19: 53148835-53148836 on Build GRCh38
rs199849982 Chr.19: 53148652-53148652 on Build GRCh38
rs781189039 Chr.19: 53148769-53148769 on Build GRCh38
rs770505528 Chr.19: 53148800-53148800 on Build GRCh38
rs921059554 Chr.19: 53149016-53149016 on Build GRCh38
rs771979109 Chr.19: 53148633-53148633 on Build GRCh38
rs756783885 Chr.19: 53148689-53148689 on Build GRCh38
rs762525812 Chr.19: 53148673-53148673 on Build GRCh38
rs529040460 Chr.19: 53149042-53149042 on Build GRCh38
rs761129866 Chr.19: 53148691-53148691 on Build GRCh38
rs75972953 Chr.19: 53149029-53149029 on Build GRCh38
rs1009268080 Chr.19: 53148801-53148801 on Build GRCh38
rs375949478 Chr.19: 53148664-53148664 on Build GRCh38
rs779112687 Chr.19: 53148744-53148744 on Build GRCh38
rs979459674 Chr.19: 53148653-53148653 on Build GRCh38
rs1025132840 Chr.19: 53148807-53148807 on Build GRCh38
rs542475240 Chr.19: 53148776-53148776 on Build GRCh38
rs61331547 Chr.19: 53148676-53148676 on Build GRCh38
rs141509306 Chr.19: 53149003-53149003 on Build GRCh38
rs1039008030 Chr.19: 53148880-53148880 on Build GRCh38
rs565635047 Chr.19: 53149071-53149071 on Build GRCh38
rs892122331 Chr.19: 53148794-53148794 on Build GRCh38
rs776539514 Chr.19: 53148651-53148651 on Build GRCh38
rs758532440 Chr.19: 53148749-53148749 on Build GRCh38
rs142157721 Chr.19: 53148743-53148743 on Build GRCh38
rs145911248 Chr.19: 53148746-53148746 on Build GRCh38
rs757457846 Chr.19: 53149040-53149040 on Build GRCh38
rs930562349 Chr.19: 53148895-53148895 on Build GRCh38
rs1021962745 Chr.19: 53149034-53149034 on Build GRCh38

View Full Product Details