Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53153648-53154157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF347
Entrez Gene ID
84671
Gene Name
zinc finger protein 347
Gene Aliases
ZNF1111
UniGene
Hs.467239
Gene Chromosome Location
Chr.19: 53134935-53159069 on Build GRCh38
Transcripts
NM_001172674.1 NM_001172675.1 NM_032584.2 XM_005259335.4 XM_005259336.4 XM_017027384.1 XM_017027385.1 XM_017027386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
475199 c.15G>C p.Q5H Chr.19: 53153733-53153733 on Build GRCh38
1305045 c.9C>T p.L3L Chr.19: 53153739-53153739 on Build GRCh38
5960160 c.7_8insC p.L3fs*40 Chr.19: 53153740-53153741 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003119063 Chr.19: 53154101-53154101 on Build GRCh38
rs1023537943 Chr.19: 53154105-53154105 on Build GRCh38
rs924719130 Chr.19: 53153962-53153962 on Build GRCh38
rs992062376 Chr.19: 53153891-53153892 on Build GRCh38
rs761247857 Chr.19: 53153728-53153728 on Build GRCh38
rs564455918 Chr.19: 53153798-53153798 on Build GRCh38
rs911759244 Chr.19: 53154005-53154005 on Build GRCh38
rs567917450 Chr.19: 53153834-53153834 on Build GRCh38
rs764745011 Chr.19: 53153785-53153785 on Build GRCh38
rs531393756 Chr.19: 53153821-53153821 on Build GRCh38
rs971597999 Chr.19: 53153813-53153813 on Build GRCh38
rs764187137 Chr.19: 53153785-53153785 on Build GRCh38
rs982103537 Chr.19: 53153818-53153818 on Build GRCh38
rs113788271 Chr.19: 53153966-53153966 on Build GRCh38
rs1020423882 Chr.19: 53153800-53153800 on Build GRCh38
rs1051723412 Chr.19: 53153993-53153993 on Build GRCh38
rs759886232 Chr.19: 53153704-53153704 on Build GRCh38
rs146492650 Chr.19: 53153892-53153893 on Build GRCh38
rs1035907822 Chr.19: 53153890-53153891 on Build GRCh38
rs556343089 Chr.19: 53153911-53153911 on Build GRCh38
rs756857694 Chr.19: 53153845-53153845 on Build GRCh38
rs920030388 Chr.19: 53153880-53153880 on Build GRCh38
rs181351304 Chr.19: 53154127-53154127 on Build GRCh38
rs535326418 Chr.19: 53153874-53153874 on Build GRCh38
rs200775658 Chr.19: 53153684-53153684 on Build GRCh38
rs771226911 Chr.19: 53153683-53153683 on Build GRCh38
rs774824912 Chr.19: 53153703-53153703 on Build GRCh38
rs899020550 Chr.19: 53154033-53154033 on Build GRCh38
rs1034319852 Chr.19: 53153819-53153819 on Build GRCh38
rs1003222830 Chr.19: 53153879-53153879 on Build GRCh38
rs576893004 Chr.19: 53154096-53154096 on Build GRCh38
rs1035053117 Chr.19: 53154108-53154108 on Build GRCh38
rs34187329 Chr.19: 53153905-53153906 on Build GRCh38
rs1007771592 Chr.19: 53153673-53153675 on Build GRCh38
rs188891458 Chr.19: 53153890-53153890 on Build GRCh38
rs919817521 Chr.19: 53153843-53153843 on Build GRCh38
rs764709922 Chr.19: 53153780-53153780 on Build GRCh38
rs943340182 Chr.19: 53154011-53154011 on Build GRCh38
rs147331632 Chr.19: 53154041-53154041 on Build GRCh38
rs950338333 Chr.19: 53153873-53153873 on Build GRCh38
rs750012735 Chr.19: 53154063-53154063 on Build GRCh38
rs775777806 Chr.19: 53153736-53153736 on Build GRCh38
rs147075864 Chr.19: 53153673-53153673 on Build GRCh38
rs200016852 Chr.19: 53153777-53153777 on Build GRCh38
rs958714172 Chr.19: 53154130-53154130 on Build GRCh38
rs1003170419 Chr.19: 53154083-53154083 on Build GRCh38
rs111331117 Chr.19: 53154100-53154100 on Build GRCh38
rs576247733 Chr.19: 53154021-53154021 on Build GRCh38
rs978840638 Chr.19: 53153950-53153950 on Build GRCh38
rs185208388 Chr.19: 53153831-53153831 on Build GRCh38
rs575098992 Chr.19: 53154066-53154066 on Build GRCh38
rs557821346 Chr.19: 53154009-53154009 on Build GRCh38
rs1034730127 Chr.19: 53154094-53154094 on Build GRCh38
rs538440486 Chr.19: 53154038-53154038 on Build GRCh38
rs1032547188 Chr.19: 53153737-53153737 on Build GRCh38
rs138483279 Chr.19: 53153933-53153933 on Build GRCh38
rs974181280 Chr.19: 53153848-53153848 on Build GRCh38
rs760685691 Chr.19: 53153739-53153739 on Build GRCh38
rs772408218 Chr.19: 53153718-53153718 on Build GRCh38
rs936359382 Chr.19: 53154040-53154040 on Build GRCh38

View Full Product Details