Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54594427-54594921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRA1
Entrez Gene ID
11024
Gene Name
leukocyte immunoglobulin like receptor A1
Gene Aliases
CD85I, LIR-6, LIR6
UniGene
Hs.710507
Gene Chromosome Location
Chr.19: 54593576-54602381 on Build GRCh38
Transcripts
NM_001278318.1 NM_001278319.1 NM_006863.3 NR_103501.1 NR_103502.1 NR_103503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3673010 c.323C>T p.S108L Chr.19: 54594917-54594917 on Build GRCh38
3538879 c.168G>A p.Q56Q Chr.19: 54594762-54594762 on Build GRCh38
3538877 c.115A>C p.I39L Chr.19: 54594709-54594709 on Build GRCh38
5097329 c.199G>A p.A67T Chr.19: 54594793-54594793 on Build GRCh38
713665 c.249C>T p.F83F Chr.19: 54594843-54594843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031999998 Chr.19: 54594541-54594541 on Build GRCh38
rs759964535 Chr.19: 54594874-54594874 on Build GRCh38
rs367561488 Chr.19: 54594824-54594824 on Build GRCh38
rs769548699 Chr.19: 54594850-54594850 on Build GRCh38
rs773855809 Chr.19: 54594717-54594717 on Build GRCh38
rs16989288 Chr.19: 54594873-54594873 on Build GRCh38
rs760676793 Chr.19: 54594494-54594494 on Build GRCh38
rs774808259 Chr.19: 54594526-54594526 on Build GRCh38
rs747684580 Chr.19: 54594735-54594735 on Build GRCh38
rs147848714 Chr.19: 54594823-54594823 on Build GRCh38
rs148392870 Chr.19: 54594696-54594696 on Build GRCh38
rs758619951 Chr.19: 54594678-54594678 on Build GRCh38
rs775067092 Chr.19: 54594466-54594466 on Build GRCh38
rs762332897 Chr.19: 54594473-54594473 on Build GRCh38
rs780036728 Chr.19: 54594851-54594851 on Build GRCh38
rs766834992 Chr.19: 54594871-54594871 on Build GRCh38
rs761147156 Chr.19: 54594744-54594744 on Build GRCh38
rs893444156 Chr.19: 54594764-54594764 on Build GRCh38
rs372235251 Chr.19: 54594856-54594856 on Build GRCh38
rs775575815 Chr.19: 54594713-54594713 on Build GRCh38
rs372207012 Chr.19: 54594638-54594638 on Build GRCh38
rs372670314 Chr.19: 54594464-54594464 on Build GRCh38
rs373727113 Chr.19: 54594809-54594809 on Build GRCh38
rs773773782 Chr.19: 54594460-54594460 on Build GRCh38
rs564359802 Chr.19: 54594538-54594538 on Build GRCh38
rs772715948 Chr.19: 54594701-54594701 on Build GRCh38
rs374633256 Chr.19: 54594866-54594866 on Build GRCh38
rs150949431 Chr.19: 54594768-54594768 on Build GRCh38
rs773547373 Chr.19: 54594860-54594860 on Build GRCh38
rs201121747 Chr.19: 54594639-54594639 on Build GRCh38
rs374294332 Chr.19: 54594495-54594495 on Build GRCh38
rs111606492 Chr.19: 54594504-54594504 on Build GRCh38
rs774697016 Chr.19: 54594777-54594777 on Build GRCh38
rs530413054 Chr.19: 54594518-54594518 on Build GRCh38
rs965494161 Chr.19: 54594552-54594552 on Build GRCh38
rs770558939 Chr.19: 54594767-54594767 on Build GRCh38
rs781099508 Chr.19: 54594765-54594765 on Build GRCh38
rs774217344 Chr.19: 54594619-54594619 on Build GRCh38
rs575697897 Chr.19: 54594773-54594773 on Build GRCh38
rs200632994 Chr.19: 54594801-54594801 on Build GRCh38
rs533353815 Chr.19: 54594542-54594542 on Build GRCh38
rs192774454 Chr.19: 54594550-54594550 on Build GRCh38
rs764428603 Chr.19: 54594720-54594720 on Build GRCh38
rs528674997 Chr.19: 54594894-54594894 on Build GRCh38
rs572192724 Chr.19: 54594848-54594848 on Build GRCh38
rs746376728 Chr.19: 54594688-54594688 on Build GRCh38
rs73612456 Chr.19: 54594687-54594687 on Build GRCh38
rs369400190 Chr.19: 54594457-54594457 on Build GRCh38
rs200284559 Chr.19: 54594509-54594509 on Build GRCh38
rs138949486 Chr.19: 54594887-54594887 on Build GRCh38
rs770627759 Chr.19: 54594704-54594704 on Build GRCh38
rs749270821 Chr.19: 54594453-54594453 on Build GRCh38
rs976869424 Chr.19: 54594561-54594561 on Build GRCh38
rs535289226 Chr.19: 54594800-54594800 on Build GRCh38
rs768073987 Chr.19: 54594649-54594650 on Build GRCh38
rs759678804 Chr.19: 54594665-54594665 on Build GRCh38
rs747534144 Chr.19: 54594462-54594462 on Build GRCh38
rs780861027 Chr.19: 54594883-54594883 on Build GRCh38
rs376588218 Chr.19: 54594727-54594727 on Build GRCh38
rs10416264 Chr.19: 54594458-54594458 on Build GRCh38
rs757814151 Chr.19: 54594744-54594744 on Build GRCh38
rs747596999 Chr.19: 54594694-54594694 on Build GRCh38
rs781505304 Chr.19: 54594526-54594526 on Build GRCh38
rs768448003 Chr.19: 54594477-54594477 on Build GRCh38
rs552034786 Chr.19: 54594709-54594709 on Build GRCh38
rs771793774 Chr.19: 54594637-54594637 on Build GRCh38
rs376376360 Chr.19: 54594878-54594878 on Build GRCh38
rs762282430 Chr.19: 54594646-54594646 on Build GRCh38
rs190014598 Chr.19: 54594728-54594728 on Build GRCh38
rs779145927 Chr.19: 54594503-54594503 on Build GRCh38
rs757712125 Chr.19: 54594726-54594726 on Build GRCh38
rs768339802 Chr.19: 54594690-54594690 on Build GRCh38
rs140932075 Chr.19: 54594770-54594770 on Build GRCh38
rs746627684 Chr.19: 54594759-54594759 on Build GRCh38
rs778580384 Chr.19: 54594692-54594692 on Build GRCh38
rs200823340 Chr.19: 54594897-54594897 on Build GRCh38
rs148921901 Chr.19: 54594890-54594890 on Build GRCh38
rs375563330 Chr.19: 54594648-54594648 on Build GRCh38
rs555171577 Chr.19: 54594808-54594808 on Build GRCh38
rs954145470 Chr.19: 54594579-54594580 on Build GRCh38
rs746458602 Chr.19: 54594832-54594832 on Build GRCh38
rs113761418 Chr.19: 54594570-54594570 on Build GRCh38
rs780669485 Chr.19: 54594731-54594731 on Build GRCh38
rs777816117 Chr.19: 54594679-54594679 on Build GRCh38
rs370643444 Chr.19: 54594769-54594769 on Build GRCh38
rs1021184744 Chr.19: 54594794-54594794 on Build GRCh38
rs369930350 Chr.19: 54594479-54594479 on Build GRCh38
rs770476706 Chr.19: 54594891-54594891 on Build GRCh38
rs201715627 Chr.19: 54594895-54594895 on Build GRCh38
rs772394849 Chr.19: 54594459-54594459 on Build GRCh38
rs1001959545 Chr.19: 54594714-54594714 on Build GRCh38
rs754391696 Chr.19: 54594816-54594816 on Build GRCh38
rs768254003 Chr.19: 54594618-54594618 on Build GRCh38
rs747051080 Chr.19: 54594576-54594576 on Build GRCh38
rs527563556 Chr.19: 54594505-54594505 on Build GRCh38
rs777546908 Chr.19: 54594747-54594747 on Build GRCh38
rs142517538 Chr.19: 54594712-54594712 on Build GRCh38
rs762969311 Chr.19: 54594716-54594716 on Build GRCh38
rs761186589 Chr.19: 54594868-54594868 on Build GRCh38
rs182094020 Chr.19: 54594497-54594497 on Build GRCh38
rs879654906 Chr.19: 54594818-54594818 on Build GRCh38
rs746859577 Chr.19: 54594703-54594703 on Build GRCh38
rs761544599 Chr.19: 54594636-54594636 on Build GRCh38
rs758560543 Chr.19: 54594562-54594562 on Build GRCh38
rs10416917 Chr.19: 54594852-54594852 on Build GRCh38
rs748228034 Chr.19: 54594896-54594896 on Build GRCh38
rs764080826 Chr.19: 54594774-54594774 on Build GRCh38
rs76561009 Chr.19: 54594710-54594710 on Build GRCh38
rs149357491 Chr.19: 54594456-54594456 on Build GRCh38
rs761744524 Chr.19: 54594481-54594481 on Build GRCh38
rs767417233 Chr.19: 54594487-54594487 on Build GRCh38
rs780771279 Chr.19: 54594845-54594845 on Build GRCh38
rs958572000 Chr.19: 54594549-54594549 on Build GRCh38
rs186732337 Chr.19: 54594530-54594530 on Build GRCh38
rs748675155 Chr.19: 54594830-54594830 on Build GRCh38
rs375610702 Chr.19: 54594617-54594617 on Build GRCh38
rs756886821 Chr.19: 54594834-54594834 on Build GRCh38
rs141166390 Chr.19: 54594876-54594876 on Build GRCh38
rs111879471 Chr.19: 54594684-54594684 on Build GRCh38
rs529375145 Chr.19: 54594884-54594884 on Build GRCh38
rs148109850 Chr.19: 54594901-54594901 on Build GRCh38
rs141422955 Chr.19: 54594858-54594858 on Build GRCh38
rs200199628 Chr.19: 54594857-54594857 on Build GRCh38
rs113170106 Chr.19: 54594649-54594649 on Build GRCh38
rs765286954 Chr.19: 54594666-54594666 on Build GRCh38
rs372024491 Chr.19: 54594877-54594877 on Build GRCh38
rs1002307347 Chr.19: 54594540-54594540 on Build GRCh38
rs752713074 Chr.19: 54594673-54594673 on Build GRCh38

View Full Product Details