Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52436664-52437163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC11
Entrez Gene ID
23318
Gene Name
zinc finger CCHC-type containing 11
Gene Aliases
PAPD3, TUT4
UniGene
Hs.655407
Gene Chromosome Location
Chr.1: 52423275-52553463 on Build GRCh38
Transcripts
NM_001009881.2 NM_015269.2 XM_005270676.2 XM_005270678.2 XM_005270679.2 XM_005270680.2 XM_005270681.2 XM_005270682.2 XM_005270683.2 XM_006710498.2 XM_006710499.3 XM_006710500.1 XM_011541097.1 XM_011541098.1 XM_011541099.1 XM_017000803.1 XM_017000804.1 XM_017000805.1 XM_017000806.1 XM_017000807.1 XM_017000808.1 XM_017000809.1 XM_017000810.1 XM_017000811.1 XM_017000812.1 XM_017000813.1 XM_017000814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3490667 c.4125G>A p.E1375E Chr.1: 52436792-52436792 on Build GRCh38
6825883 c.4162G>A p.A1388T Chr.1: 52436755-52436755 on Build GRCh38
910672 c.3984G>T p.K1328N Chr.1: 52436933-52436933 on Build GRCh38
1748505 c.4108C>T p.R1370* Chr.1: 52436809-52436809 on Build GRCh38
6413020 c.4139G>T p.R1380L Chr.1: 52436778-52436778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140074425 Chr.1: 52436852-52436852 on Build GRCh38
rs569002401 Chr.1: 52436716-52436716 on Build GRCh38
rs780171701 Chr.1: 52436890-52436890 on Build GRCh38
rs757124815 Chr.1: 52436994-52436994 on Build GRCh38
rs374115184 Chr.1: 52436983-52436983 on Build GRCh38
rs182969283 Chr.1: 52436779-52436779 on Build GRCh38
rs761228437 Chr.1: 52436973-52436973 on Build GRCh38
rs574685601 Chr.1: 52436876-52436876 on Build GRCh38
rs1050676319 Chr.1: 52437039-52437039 on Build GRCh38
rs371549563 Chr.1: 52436831-52436831 on Build GRCh38
rs749579251 Chr.1: 52436893-52436893 on Build GRCh38
rs752326298 Chr.1: 52436749-52436749 on Build GRCh38
rs774694566 Chr.1: 52436821-52436821 on Build GRCh38
rs558547500 Chr.1: 52436824-52436824 on Build GRCh38
rs188412921 Chr.1: 52436868-52436868 on Build GRCh38
rs1018616455 Chr.1: 52436812-52436812 on Build GRCh38
rs1037025979 Chr.1: 52436725-52436725 on Build GRCh38
rs768808125 Chr.1: 52436881-52436883 on Build GRCh38
rs750386336 Chr.1: 52437017-52437017 on Build GRCh38
rs767287233 Chr.1: 52437005-52437005 on Build GRCh38
rs991430248 Chr.1: 52436701-52436701 on Build GRCh38
rs146246594 Chr.1: 52436791-52436791 on Build GRCh38
rs557641096 Chr.1: 52436937-52436939 on Build GRCh38
rs748413020 Chr.1: 52436990-52436990 on Build GRCh38
rs200561885 Chr.1: 52436717-52436717 on Build GRCh38
rs550797984 Chr.1: 52436705-52436705 on Build GRCh38
rs754460618 Chr.1: 52436743-52436743 on Build GRCh38
rs757073625 Chr.1: 52436869-52436869 on Build GRCh38
rs61736686 Chr.1: 52436853-52436853 on Build GRCh38
rs367983064 Chr.1: 52436992-52436992 on Build GRCh38
rs967876832 Chr.1: 52436871-52436871 on Build GRCh38
rs879636613 Chr.1: 52436952-52436952 on Build GRCh38
rs75863166 Chr.1: 52437038-52437038 on Build GRCh38
rs897970567 Chr.1: 52436726-52436726 on Build GRCh38
rs775582914 Chr.1: 52436804-52436804 on Build GRCh38
rs752075573 Chr.1: 52436709-52436709 on Build GRCh38
rs376676575 Chr.1: 52436789-52436789 on Build GRCh38
rs758115885 Chr.1: 52436857-52436857 on Build GRCh38
rs751221593 Chr.1: 52436993-52436993 on Build GRCh38
rs149301397 Chr.1: 52436875-52436875 on Build GRCh38
rs373115070 Chr.1: 52436801-52436801 on Build GRCh38
rs762739631 Chr.1: 52436988-52436988 on Build GRCh38
rs762195574 Chr.1: 52436904-52436904 on Build GRCh38
rs1019429301 Chr.1: 52437126-52437126 on Build GRCh38
rs781025662 Chr.1: 52436870-52436870 on Build GRCh38
rs750211663 Chr.1: 52437037-52437037 on Build GRCh38
rs908303057 Chr.1: 52436966-52436966 on Build GRCh38
rs779737793 Chr.1: 52437087-52437087 on Build GRCh38
rs750901130 Chr.1: 52436838-52436838 on Build GRCh38
rs772412739 Chr.1: 52436927-52436927 on Build GRCh38
rs1022379722 Chr.1: 52436953-52436962 on Build GRCh38
rs374452717 Chr.1: 52436841-52436841 on Build GRCh38
rs753265992 Chr.1: 52436734-52436734 on Build GRCh38
rs774506514 Chr.1: 52436900-52436900 on Build GRCh38
rs1007356813 Chr.1: 52436809-52436809 on Build GRCh38
rs201801992 Chr.1: 52436899-52436899 on Build GRCh38
rs555050333 Chr.1: 52436719-52436719 on Build GRCh38
rs141484128 Chr.1: 52436808-52436808 on Build GRCh38
rs899414666 Chr.1: 52437031-52437031 on Build GRCh38
rs758279485 Chr.1: 52436753-52436753 on Build GRCh38
rs756076545 Chr.1: 52437019-52437019 on Build GRCh38
rs763179972 Chr.1: 52436805-52436805 on Build GRCh38
rs367870528 Chr.1: 52436748-52436748 on Build GRCh38
rs940834604 Chr.1: 52436969-52436969 on Build GRCh38
rs764718607 Chr.1: 52436981-52436981 on Build GRCh38
rs953410754 Chr.1: 52436735-52436735 on Build GRCh38
rs746903493 Chr.1: 52436763-52436765 on Build GRCh38
rs1008917724 Chr.1: 52437106-52437106 on Build GRCh38
rs773604880 Chr.1: 52436946-52436946 on Build GRCh38
rs777747651 Chr.1: 52436864-52436864 on Build GRCh38
rs780028557 Chr.1: 52436734-52436737 on Build GRCh38
rs751607874 Chr.1: 52436763-52436763 on Build GRCh38

View Full Product Details