Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54226818-54227314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSBP3
Entrez Gene ID
23648
Gene Name
single stranded DNA binding protein 3
Gene Aliases
CSDP, SSDP, SSDP1
UniGene
Hs.476706
Gene Chromosome Location
Chr.1: 54225431-54413479 on Build GRCh38
Transcripts
NM_001009955.3 NM_018070.4 NM_145716.3 XM_006710545.3 XM_017000897.1 XM_017000898.1 XM_017000899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7095101 c.1158G>T p.M386I Chr.1: 54227140-54227140 on Build GRCh38
7095102 c.1077G>T p.M359I Chr.1: 54227140-54227140 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs927335257 Chr.1: 54227001-54227001 on Build GRCh38
rs912473017 Chr.1: 54227239-54227239 on Build GRCh38
rs752331280 Chr.1: 54227210-54227210 on Build GRCh38
rs758093263 Chr.1: 54227211-54227211 on Build GRCh38
rs144624285 Chr.1: 54227143-54227143 on Build GRCh38
rs200366318 Chr.1: 54227124-54227124 on Build GRCh38
rs758577615 Chr.1: 54227099-54227099 on Build GRCh38
rs914518802 Chr.1: 54227270-54227270 on Build GRCh38
rs937022982 Chr.1: 54227005-54227005 on Build GRCh38
rs890979326 Chr.1: 54227086-54227086 on Build GRCh38
rs558623961 Chr.1: 54227205-54227205 on Build GRCh38
rs977783383 Chr.1: 54227230-54227230 on Build GRCh38
rs750916256 Chr.1: 54227198-54227198 on Build GRCh38
rs968587952 Chr.1: 54227282-54227282 on Build GRCh38
rs369136306 Chr.1: 54227096-54227096 on Build GRCh38
rs71066908 Chr.1: 54227187-54227188 on Build GRCh38
rs201308890 Chr.1: 54227129-54227129 on Build GRCh38
rs758498750 Chr.1: 54227182-54227182 on Build GRCh38
rs752807208 Chr.1: 54227110-54227110 on Build GRCh38
rs201486426 Chr.1: 54227090-54227090 on Build GRCh38
rs372034987 Chr.1: 54227189-54227190 on Build GRCh38
rs766199642 Chr.1: 54227080-54227080 on Build GRCh38
rs3058961 Chr.1: 54227191-54227192 on Build GRCh38
rs1044440661 Chr.1: 54226848-54226848 on Build GRCh38
rs552124904 Chr.1: 54227077-54227077 on Build GRCh38
rs112173146 Chr.1: 54227188-54227188 on Build GRCh38
rs752110846 Chr.1: 54227184-54227184 on Build GRCh38
rs534119623 Chr.1: 54227181-54227181 on Build GRCh38
rs555288823 Chr.1: 54227023-54227033 on Build GRCh38
rs778380186 Chr.1: 54227111-54227111 on Build GRCh38
rs943883123 Chr.1: 54227295-54227295 on Build GRCh38
rs1057333989 Chr.1: 54226905-54226905 on Build GRCh38
rs981544786 Chr.1: 54226997-54226997 on Build GRCh38
rs775791953 Chr.1: 54227191-54227191 on Build GRCh38
rs954852114 Chr.1: 54227093-54227093 on Build GRCh38
rs545041801 Chr.1: 54227190-54227190 on Build GRCh38
rs566587497 Chr.1: 54227034-54227034 on Build GRCh38
rs1005032207 Chr.1: 54226921-54226921 on Build GRCh38
rs757675152 Chr.1: 54227194-54227194 on Build GRCh38
rs1025534412 Chr.1: 54226915-54226915 on Build GRCh38
rs757500699 Chr.1: 54227113-54227113 on Build GRCh38
rs754487199 Chr.1: 54227188-54227189 on Build GRCh38
rs768405555 Chr.1: 54227125-54227125 on Build GRCh38
rs761062692 Chr.1: 54227185-54227186 on Build GRCh38
rs538650376 Chr.1: 54227071-54227071 on Build GRCh38
rs938425580 Chr.1: 54226893-54226893 on Build GRCh38
rs914465259 Chr.1: 54226845-54226845 on Build GRCh38
rs927024701 Chr.1: 54226999-54226999 on Build GRCh38
rs756452193 Chr.1: 54227170-54227172 on Build GRCh38
rs758852733 Chr.1: 54227181-54227182 on Build GRCh38
rs770938911 Chr.1: 54227189-54227189 on Build GRCh38
rs553040722 Chr.1: 54227187-54227187 on Build GRCh38
rs767777634 Chr.1: 54227103-54227106 on Build GRCh38
rs1037972279 Chr.1: 54226927-54226927 on Build GRCh38
rs765455262 Chr.1: 54227175-54227175 on Build GRCh38
rs752985746 Chr.1: 54227178-54227178 on Build GRCh38
rs927085718 Chr.1: 54226857-54226857 on Build GRCh38
rs575023049 Chr.1: 54227168-54227168 on Build GRCh38
rs59695718 Chr.1: 54227185-54227185 on Build GRCh38
rs60967318 Chr.1: 54226906-54226906 on Build GRCh38
rs141516563 Chr.1: 54227186-54227187 on Build GRCh38
rs769663955 Chr.1: 54227182-54227183 on Build GRCh38
rs764258332 Chr.1: 54227193-54227193 on Build GRCh38
rs967053660 Chr.1: 54227220-54227220 on Build GRCh38
rs886613115 Chr.1: 54226918-54226918 on Build GRCh38
rs779540947 Chr.1: 54227118-54227118 on Build GRCh38
rs914496180 Chr.1: 54227038-54227039 on Build GRCh38
rs770811862 Chr.1: 54227192-54227201 on Build GRCh38
rs780462752 Chr.1: 54227206-54227206 on Build GRCh38
rs781390606 Chr.1: 54227186-54227186 on Build GRCh38
rs755282816 Chr.1: 54227089-54227089 on Build GRCh38
rs759643314 Chr.1: 54227171-54227171 on Build GRCh38
rs764276864 Chr.1: 54227183-54227183 on Build GRCh38
rs753045665 Chr.1: 54227129-54227130 on Build GRCh38
rs547113269 Chr.1: 54226986-54226986 on Build GRCh38
rs185858837 Chr.1: 54226941-54226941 on Build GRCh38
rs759632663 Chr.1: 54227183-54227184 on Build GRCh38
rs970906811 Chr.1: 54226936-54226936 on Build GRCh38
rs766072005 Chr.1: 54227149-54227149 on Build GRCh38
rs989829837 Chr.1: 54227204-54227204 on Build GRCh38
rs372535521 Chr.1: 54227191-54227191 on Build GRCh38
rs199692523 Chr.1: 54227117-54227117 on Build GRCh38
rs775645285 Chr.1: 54227184-54227185 on Build GRCh38
rs989779147 Chr.1: 54227021-54227021 on Build GRCh38
rs1009405508 Chr.1: 54227097-54227097 on Build GRCh38
rs796309778 Chr.1: 54227181-54227182 on Build GRCh38
rs754651175 Chr.1: 54227182-54227182 on Build GRCh38
rs371733997 Chr.1: 54227114-54227114 on Build GRCh38
rs372110874 Chr.1: 54227082-54227082 on Build GRCh38
rs777768042 Chr.1: 54227190-54227191 on Build GRCh38
rs914253240 Chr.1: 54227216-54227217 on Build GRCh38
rs115749686 Chr.1: 54227192-54227192 on Build GRCh38
rs764611487 Chr.1: 54227180-54227180 on Build GRCh38
rs796699424 Chr.1: 54227182-54227183 on Build GRCh38
rs747535779 Chr.1: 54227112-54227112 on Build GRCh38
rs754298483 Chr.1: 54227180-54227181 on Build GRCh38
rs759736876 Chr.1: 54227103-54227104 on Build GRCh38
rs947067720 Chr.1: 54226846-54226846 on Build GRCh38

View Full Product Details