Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54665100-54665596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH7
Entrez Gene ID
374977
Gene Name
maestro heat like repeat family member 7
Gene Aliases
C1orf175, HEATR8
UniGene
Hs.412482
Gene Chromosome Location
Chr.1: 54641740-54710267 on Build GRCh38
Transcripts
NM_001039464.3 NM_001291332.1 NR_026782.2 NR_111931.1 NR_037639.2 NR_037640.2 NR_037641.2

Gene Symbol
MROH7-TTC4
Entrez Gene ID
100527960
Gene Name
MROH7-TTC4 readthrough (NMD candidate)
Gene Aliases
HEATR8-TTC4
UniGene
Gene Chromosome Location
Chr.1: 54641740-54742657 on Build GRCh38
Transcripts
NM_001039464.3 NM_001291332.1 NR_026782.2 NR_111931.1 NR_037639.2 NR_037640.2 NR_037641.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6715214 c.1244A>G p.E415G Chr.1: 54665179-54665179 on Build GRCh38
6285358 c.1301C>T p.A434V Chr.1: 54665236-54665236 on Build GRCh38
6715212 c.1244A>G p.E415G Chr.1: 54665179-54665179 on Build GRCh38
1198420 c.1247T>C p.V416A Chr.1: 54665182-54665182 on Build GRCh38
910905 c.1256G>A p.C419Y Chr.1: 54665191-54665191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548288202 Chr.1: 54665414-54665414 on Build GRCh38
rs974076747 Chr.1: 54665526-54665526 on Build GRCh38
rs774157644 Chr.1: 54665170-54665170 on Build GRCh38
rs549227921 Chr.1: 54665502-54665502 on Build GRCh38
rs984224054 Chr.1: 54665534-54665534 on Build GRCh38
rs748401514 Chr.1: 54665232-54665232 on Build GRCh38
rs952662606 Chr.1: 54665528-54665528 on Build GRCh38
rs757127772 Chr.1: 54665185-54665185 on Build GRCh38
rs534654246 Chr.1: 54665570-54665570 on Build GRCh38
rs770285285 Chr.1: 54665265-54665265 on Build GRCh38
rs376285425 Chr.1: 54665426-54665426 on Build GRCh38
rs565964320 Chr.1: 54665553-54665553 on Build GRCh38
rs909146715 Chr.1: 54665505-54665505 on Build GRCh38
rs981255541 Chr.1: 54665228-54665228 on Build GRCh38
rs565172588 Chr.1: 54665376-54665376 on Build GRCh38
rs756120239 Chr.1: 54665209-54665209 on Build GRCh38
rs566597285 Chr.1: 54665460-54665460 on Build GRCh38
rs957017517 Chr.1: 54665334-54665334 on Build GRCh38
rs777956912 Chr.1: 54665241-54665241 on Build GRCh38
rs996981844 Chr.1: 54665305-54665305 on Build GRCh38
rs529756072 Chr.1: 54665361-54665361 on Build GRCh38
rs1055368713 Chr.1: 54665344-54665344 on Build GRCh38
rs140680909 Chr.1: 54665377-54665377 on Build GRCh38
rs370263661 Chr.1: 54665274-54665274 on Build GRCh38
rs941311413 Chr.1: 54665525-54665525 on Build GRCh38
rs999392831 Chr.1: 54665422-54665422 on Build GRCh38
rs370670189 Chr.1: 54665480-54665480 on Build GRCh38
rs1019901527 Chr.1: 54665439-54665439 on Build GRCh38
rs903352155 Chr.1: 54665386-54665386 on Build GRCh38
rs147673600 Chr.1: 54665330-54665330 on Build GRCh38
rs778953026 Chr.1: 54665141-54665141 on Build GRCh38
rs938399790 Chr.1: 54665337-54665337 on Build GRCh38
rs748168676 Chr.1: 54665151-54665151 on Build GRCh38
rs981260344 Chr.1: 54665421-54665421 on Build GRCh38
rs745912490 Chr.1: 54665255-54665256 on Build GRCh38
rs1011215580 Chr.1: 54665362-54665362 on Build GRCh38
rs184539073 Chr.1: 54665501-54665501 on Build GRCh38
rs761267194 Chr.1: 54665159-54665159 on Build GRCh38
rs144937272 Chr.1: 54665454-54665454 on Build GRCh38
rs191753685 Chr.1: 54665153-54665153 on Build GRCh38
rs753726663 Chr.1: 54665221-54665221 on Build GRCh38
rs200141664 Chr.1: 54665304-54665304 on Build GRCh38
rs1047682049 Chr.1: 54665256-54665256 on Build GRCh38
rs977718517 Chr.1: 54665238-54665238 on Build GRCh38
rs376325842 Chr.1: 54665155-54665155 on Build GRCh38
rs750310367 Chr.1: 54665182-54665182 on Build GRCh38
rs377527312 Chr.1: 54665220-54665220 on Build GRCh38
rs563440250 Chr.1: 54665475-54665475 on Build GRCh38
rs761336036 Chr.1: 54665176-54665176 on Build GRCh38
rs536794118 Chr.1: 54665268-54665268 on Build GRCh38
rs545367399 Chr.1: 54665370-54665370 on Build GRCh38
rs772849679 Chr.1: 54665277-54665277 on Build GRCh38
rs754783332 Chr.1: 54665223-54665223 on Build GRCh38
rs201380455 Chr.1: 54665165-54665165 on Build GRCh38
rs866683172 Chr.1: 54665208-54665208 on Build GRCh38
rs999505594 Chr.1: 54665325-54665325 on Build GRCh38
rs762688471 Chr.1: 54665163-54665163 on Build GRCh38
rs747441442 Chr.1: 54665257-54665257 on Build GRCh38
rs756707476 Chr.1: 54665452-54665452 on Build GRCh38
rs779111741 Chr.1: 54665216-54665217 on Build GRCh38
rs1010678034 Chr.1: 54665126-54665126 on Build GRCh38
rs6703113 Chr.1: 54665273-54665273 on Build GRCh38
rs771177559 Chr.1: 54665161-54665161 on Build GRCh38
rs779042812 Chr.1: 54665230-54665230 on Build GRCh38
rs768936493 Chr.1: 54665127-54665127 on Build GRCh38
rs894256593 Chr.1: 54665347-54665347 on Build GRCh38
rs776627441 Chr.1: 54665259-54665259 on Build GRCh38
rs2078273 Chr.1: 54665258-54665258 on Build GRCh38
rs471724 Chr.1: 54665299-54665299 on Build GRCh38
rs767080704 Chr.1: 54665181-54665181 on Build GRCh38
rs145372615 Chr.1: 54665307-54665307 on Build GRCh38
rs559640702 Chr.1: 54665324-54665324 on Build GRCh38
rs758424599 Chr.1: 54665239-54665239 on Build GRCh38
rs915685324 Chr.1: 54665166-54665166 on Build GRCh38
rs796753710 Chr.1: 54665455-54665455 on Build GRCh38
rs760520223 Chr.1: 54665282-54665282 on Build GRCh38
rs534581167 Chr.1: 54665123-54665123 on Build GRCh38
rs766086136 Chr.1: 54665210-54665210 on Build GRCh38
rs1022642283 Chr.1: 54665372-54665372 on Build GRCh38

View Full Product Details