Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48376005-48376536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN2
Entrez Gene ID
3344
Gene Name
forkhead box N2
Gene Aliases
HTLF
UniGene
Hs.468478
Gene Chromosome Location
Chr.2: 48314640-48379295 on Build GRCh38
Transcripts
NM_002158.3 XM_005264282.2 XM_005264283.2 XM_006712001.3 XM_006712002.3 XM_011532808.2 XM_017003956.1 XM_017003957.1 XM_017003958.1 XM_017003959.1

SNP Detail

SNP ID Chromosome Location
rs971435380 Chr.2: 48376267-48376267 on Build GRCh38
rs1006580134 Chr.2: 48376101-48376101 on Build GRCh38
rs953341517 Chr.2: 48376310-48376310 on Build GRCh38
rs893546227 Chr.2: 48376198-48376198 on Build GRCh38
rs961831247 Chr.2: 48376071-48376071 on Build GRCh38
rs1054665263 Chr.2: 48376176-48376176 on Build GRCh38
rs779632140 Chr.2: 48376306-48376308 on Build GRCh38
rs561585738 Chr.2: 48376361-48376361 on Build GRCh38
rs542810791 Chr.2: 48376165-48376165 on Build GRCh38
rs1013429754 Chr.2: 48376467-48376467 on Build GRCh38
rs943519854 Chr.2: 48376034-48376034 on Build GRCh38
rs545108302 Chr.2: 48376240-48376240 on Build GRCh38
rs772664775 Chr.2: 48376143-48376143 on Build GRCh38
rs1040137693 Chr.2: 48376054-48376054 on Build GRCh38
rs1003182115 Chr.2: 48376255-48376255 on Build GRCh38
rs6705802 Chr.2: 48376344-48376344 on Build GRCh38
rs1023349562 Chr.2: 48376224-48376224 on Build GRCh38
rs948913328 Chr.2: 48376451-48376451 on Build GRCh38
rs1011961790 Chr.2: 48376201-48376201 on Build GRCh38
rs924851207 Chr.2: 48376360-48376360 on Build GRCh38
rs1033584019 Chr.2: 48376110-48376110 on Build GRCh38
rs1013419741 Chr.2: 48376205-48376205 on Build GRCh38
rs1024405530 Chr.2: 48376212-48376212 on Build GRCh38
rs561049500 Chr.2: 48376251-48376251 on Build GRCh38
rs1041966603 Chr.2: 48376163-48376163 on Build GRCh38
rs1027450926 Chr.2: 48376302-48376302 on Build GRCh38
rs943573666 Chr.2: 48376314-48376314 on Build GRCh38
rs906179795 Chr.2: 48376234-48376234 on Build GRCh38
rs1014776859 Chr.2: 48376278-48376278 on Build GRCh38
rs910748088 Chr.2: 48376032-48376032 on Build GRCh38
rs1055121194 Chr.2: 48376410-48376410 on Build GRCh38
rs936307727 Chr.2: 48376369-48376369 on Build GRCh38
rs986622565 Chr.2: 48376127-48376127 on Build GRCh38
rs906316943 Chr.2: 48376493-48376493 on Build GRCh38
rs1055081810 Chr.2: 48376417-48376417 on Build GRCh38
rs577841128 Chr.2: 48376166-48376166 on Build GRCh38
rs901591569 Chr.2: 48376062-48376062 on Build GRCh38
rs753683336 Chr.2: 48376095-48376095 on Build GRCh38
rs538651744 Chr.2: 48376355-48376355 on Build GRCh38
rs944700448 Chr.2: 48376147-48376147 on Build GRCh38
rs911995374 Chr.2: 48376352-48376352 on Build GRCh38
rs962020117 Chr.2: 48376299-48376299 on Build GRCh38
rs560682471 Chr.2: 48376271-48376271 on Build GRCh38
rs767043267 Chr.2: 48376356-48376356 on Build GRCh38
rs150951303 Chr.2: 48376300-48376300 on Build GRCh38
rs767058078 Chr.2: 48376115-48376115 on Build GRCh38
rs1047807071 Chr.2: 48376094-48376094 on Build GRCh38
rs990829579 Chr.2: 48376306-48376306 on Build GRCh38
rs911860169 Chr.2: 48376129-48376129 on Build GRCh38
rs932124661 Chr.2: 48376096-48376096 on Build GRCh38
rs774012478 Chr.2: 48376063-48376063 on Build GRCh38
rs113546529 Chr.2: 48376505-48376505 on Build GRCh38
rs778938125 Chr.2: 48376483-48376483 on Build GRCh38
rs1044769387 Chr.2: 48376476-48376476 on Build GRCh38
rs894611786 Chr.2: 48376423-48376423 on Build GRCh38
rs941704424 Chr.2: 48376414-48376414 on Build GRCh38
rs973289422 Chr.2: 48376089-48376089 on Build GRCh38

View Full Product Details