Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108255483-108255991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT1C3
Entrez Gene ID
442038
Gene Name
sulfotransferase family 1C member 3
Gene Aliases
ST1C3
UniGene
Gene Chromosome Location
Chr.2: 108239992-108265608 on Build GRCh38
Transcripts
NM_001008743.2 NM_001320878.1 XM_017004153.1 XM_017004154.1 XM_017004155.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1647015 c.488A>G p.N163S Chr.2: 108255660-108255660 on Build GRCh38
4084036 c.519C>T p.S173S Chr.2: 108255691-108255691 on Build GRCh38
6466182 c.455A>G p.H152R Chr.2: 108255627-108255627 on Build GRCh38
6792996 c.430T>C p.C144R Chr.2: 108255602-108255602 on Build GRCh38
4940240 c.521G>C p.G174A Chr.2: 108255693-108255693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979652368 Chr.2: 108255653-108255653 on Build GRCh38
rs775090080 Chr.2: 108255766-108255766 on Build GRCh38
rs962374652 Chr.2: 108255799-108255799 on Build GRCh38
rs548933792 Chr.2: 108255806-108255806 on Build GRCh38
rs766509430 Chr.2: 108255861-108255861 on Build GRCh38
rs757774310 Chr.2: 108255677-108255677 on Build GRCh38
rs759153430 Chr.2: 108255647-108255647 on Build GRCh38
rs756386313 Chr.2: 108255690-108255690 on Build GRCh38
rs750829609 Chr.2: 108255689-108255689 on Build GRCh38
rs150826802 Chr.2: 108255657-108255657 on Build GRCh38
rs6716965 Chr.2: 108255665-108255665 on Build GRCh38
rs141507165 Chr.2: 108255727-108255727 on Build GRCh38
rs36039843 Chr.2: 108255732-108255733 on Build GRCh38
rs900274158 Chr.2: 108255820-108255820 on Build GRCh38
rs149752941 Chr.2: 108255676-108255676 on Build GRCh38
rs896205515 Chr.2: 108255753-108255753 on Build GRCh38
rs144109255 Chr.2: 108255692-108255692 on Build GRCh38
rs774542688 Chr.2: 108255575-108255575 on Build GRCh38
rs1028357129 Chr.2: 108255826-108255826 on Build GRCh38
rs538551085 Chr.2: 108255867-108255867 on Build GRCh38
rs567916808 Chr.2: 108255781-108255781 on Build GRCh38
rs772127015 Chr.2: 108255580-108255580 on Build GRCh38
rs766074530 Chr.2: 108255621-108255621 on Build GRCh38
rs929492239 Chr.2: 108255601-108255601 on Build GRCh38
rs753480643 Chr.2: 108255639-108255639 on Build GRCh38
rs944777516 Chr.2: 108255613-108255613 on Build GRCh38
rs17035911 Chr.2: 108255615-108255615 on Build GRCh38
rs866431608 Chr.2: 108255668-108255668 on Build GRCh38
rs531513567 Chr.2: 108255522-108255522 on Build GRCh38
rs1011964592 Chr.2: 108255757-108255757 on Build GRCh38
rs1034862741 Chr.2: 108255739-108255739 on Build GRCh38
rs150089220 Chr.2: 108255570-108255570 on Build GRCh38
rs887651726 Chr.2: 108255865-108255865 on Build GRCh38
rs748185711 Chr.2: 108255704-108255704 on Build GRCh38
rs377144104 Chr.2: 108255532-108255532 on Build GRCh38
rs868519318 Chr.2: 108255685-108255685 on Build GRCh38
rs772143347 Chr.2: 108255709-108255709 on Build GRCh38
rs117877224 Chr.2: 108255940-108255940 on Build GRCh38
rs13392744 Chr.2: 108255556-108255556 on Build GRCh38
rs749410208 Chr.2: 108255561-108255561 on Build GRCh38
rs912021445 Chr.2: 108255607-108255607 on Build GRCh38
rs201807182 Chr.2: 108255691-108255691 on Build GRCh38
rs772986961 Chr.2: 108255712-108255712 on Build GRCh38
rs749113259 Chr.2: 108255926-108255926 on Build GRCh38
rs779195785 Chr.2: 108255699-108255699 on Build GRCh38
rs950911249 Chr.2: 108255840-108255840 on Build GRCh38
rs944746452 Chr.2: 108255925-108255925 on Build GRCh38
rs1004811135 Chr.2: 108255884-108255884 on Build GRCh38
rs781610829 Chr.2: 108255684-108255684 on Build GRCh38
rs746160007 Chr.2: 108255552-108255552 on Build GRCh38
rs149442650 Chr.2: 108255578-108255578 on Build GRCh38
rs764791248 Chr.2: 108255673-108255673 on Build GRCh38
rs757266027 Chr.2: 108255532-108255532 on Build GRCh38
rs996399280 Chr.2: 108255847-108255847 on Build GRCh38
rs556767051 Chr.2: 108255922-108255922 on Build GRCh38
rs1023231321 Chr.2: 108255784-108255784 on Build GRCh38
rs781594561 Chr.2: 108255544-108255544 on Build GRCh38
rs780100833 Chr.2: 108255559-108255559 on Build GRCh38
rs143973041 Chr.2: 108255602-108255602 on Build GRCh38
rs372693842 Chr.2: 108255527-108255527 on Build GRCh38

View Full Product Details