Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112917535-112918035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL37
Entrez Gene ID
27178
Gene Name
interleukin 37
Gene Aliases
FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1
UniGene
Hs.166371
Gene Chromosome Location
Chr.2: 112908889-112918882 on Build GRCh38
Transcripts
NM_014439.3 NM_173202.1 NM_173203.1 NM_173204.1 NM_173205.1 XM_011510963.2 XM_011510964.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685348 c.267G>T p.E89D Chr.2: 112917636-112917636 on Build GRCh38
4544105 c.271G>A p.E91K Chr.2: 112917718-112917718 on Build GRCh38
6874053 c.257G>T p.G86V Chr.2: 112917704-112917704 on Build GRCh38
3040030 c.327G>A p.L109L Chr.2: 112917774-112917774 on Build GRCh38
1743558 c.347G>A p.G116E Chr.2: 112917716-112917716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28947197 Chr.2: 112917878-112917878 on Build GRCh38
rs749751577 Chr.2: 112917732-112917732 on Build GRCh38
rs775151709 Chr.2: 112917659-112917659 on Build GRCh38
rs768327843 Chr.2: 112917755-112917755 on Build GRCh38
rs2723189 Chr.2: 112917851-112917851 on Build GRCh38
rs751615246 Chr.2: 112917801-112917801 on Build GRCh38
rs929533525 Chr.2: 112917609-112917609 on Build GRCh38
rs796113214 Chr.2: 112917809-112917809 on Build GRCh38
rs781579437 Chr.2: 112917619-112917619 on Build GRCh38
rs781317163 Chr.2: 112917701-112917701 on Build GRCh38
rs1055224854 Chr.2: 112917720-112917720 on Build GRCh38
rs771632541 Chr.2: 112917885-112917885 on Build GRCh38
rs747648030 Chr.2: 112917871-112917871 on Build GRCh38
rs759109751 Chr.2: 112917648-112917648 on Build GRCh38
rs971448371 Chr.2: 112917950-112917950 on Build GRCh38
rs116313521 Chr.2: 112917986-112917986 on Build GRCh38
rs750471839 Chr.2: 112917585-112917585 on Build GRCh38
rs751390586 Chr.2: 112917638-112917640 on Build GRCh38
rs773627585 Chr.2: 112917639-112917639 on Build GRCh38
rs368721986 Chr.2: 112917586-112917586 on Build GRCh38
rs773372458 Chr.2: 112917964-112917966 on Build GRCh38
rs372358192 Chr.2: 112917598-112917598 on Build GRCh38
rs200209919 Chr.2: 112917766-112917766 on Build GRCh38
rs761558040 Chr.2: 112917673-112917673 on Build GRCh38
rs374854417 Chr.2: 112917762-112917762 on Build GRCh38
rs763679784 Chr.2: 112917671-112917671 on Build GRCh38
rs1055699695 Chr.2: 112917987-112917987 on Build GRCh38
rs765216544 Chr.2: 112917677-112917677 on Build GRCh38
rs538974324 Chr.2: 112917840-112917840 on Build GRCh38
rs371826701 Chr.2: 112917771-112917771 on Build GRCh38
rs188118882 Chr.2: 112917730-112917730 on Build GRCh38
rs756369502 Chr.2: 112917820-112917820 on Build GRCh38
rs754925615 Chr.2: 112917812-112917813 on Build GRCh38
rs776213205 Chr.2: 112917763-112917763 on Build GRCh38
rs751668154 Chr.2: 112917697-112917697 on Build GRCh38
rs7424641 Chr.2: 112917702-112917702 on Build GRCh38
rs1047026674 Chr.2: 112917666-112917666 on Build GRCh38
rs771763163 Chr.2: 112917654-112917654 on Build GRCh38
rs778214549 Chr.2: 112917822-112917822 on Build GRCh38
rs972418793 Chr.2: 112917921-112917921 on Build GRCh38
rs753648924 Chr.2: 112917672-112917672 on Build GRCh38
rs780243678 Chr.2: 112917595-112917595 on Build GRCh38
rs760366529 Chr.2: 112917670-112917670 on Build GRCh38
rs748515156 Chr.2: 112917706-112917706 on Build GRCh38
rs749694488 Chr.2: 112917824-112917824 on Build GRCh38
rs1029963506 Chr.2: 112917847-112917847 on Build GRCh38
rs770119078 Chr.2: 112917711-112917711 on Build GRCh38
rs766324766 Chr.2: 112917790-112917790 on Build GRCh38
rs909740937 Chr.2: 112917969-112917969 on Build GRCh38
rs550322164 Chr.2: 112917974-112917974 on Build GRCh38
rs867056062 Chr.2: 112917686-112917686 on Build GRCh38
rs2723187 Chr.2: 112917692-112917692 on Build GRCh38
rs138939950 Chr.2: 112917777-112917777 on Build GRCh38
rs771599706 Chr.2: 112917736-112917736 on Build GRCh38
rs553597503 Chr.2: 112917622-112917622 on Build GRCh38
rs193066410 Chr.2: 112917829-112917829 on Build GRCh38
rs770311732 Chr.2: 112917632-112917632 on Build GRCh38
rs564709901 Chr.2: 112917806-112917806 on Build GRCh38
rs182796973 Chr.2: 112917830-112917830 on Build GRCh38
rs2723190 Chr.2: 112917995-112917995 on Build GRCh38
rs372100854 Chr.2: 112917733-112917733 on Build GRCh38
rs768156499 Chr.2: 112917835-112917835 on Build GRCh38
rs755178151 Chr.2: 112917700-112917700 on Build GRCh38
rs36114356 Chr.2: 112917985-112917986 on Build GRCh38
rs972387615 Chr.2: 112917569-112917569 on Build GRCh38
rs1048233912 Chr.2: 112917559-112917559 on Build GRCh38
rs918239957 Chr.2: 112917928-112917928 on Build GRCh38
rs2723188 Chr.2: 112917812-112917812 on Build GRCh38
rs375215039 Chr.2: 112917789-112917789 on Build GRCh38
rs919540531 Chr.2: 112917570-112917570 on Build GRCh38
rs559848172 Chr.2: 112917898-112917898 on Build GRCh38
rs750419461 Chr.2: 112917681-112917681 on Build GRCh38
rs760023330 Chr.2: 112917980-112917980 on Build GRCh38
rs370027195 Chr.2: 112917819-112917819 on Build GRCh38
rs760156316 Chr.2: 112917742-112917742 on Build GRCh38
rs867895700 Chr.2: 112917701-112917702 on Build GRCh38
rs775094506 Chr.2: 112917741-112917741 on Build GRCh38
rs767771385 Chr.2: 112917818-112917818 on Build GRCh38
rs768763628 Chr.2: 112917613-112917613 on Build GRCh38
rs150018952 Chr.2: 112917693-112917693 on Build GRCh38

View Full Product Details