Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46161061-46161569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATC1L
Entrez Gene ID
84221
Gene Name
spermatogenesis and centriole associated 1-like
Gene Aliases
C21orf56
UniGene
Hs.381214
Gene Chromosome Location
Chr.21: 46161148-46185159 on Build GRCh38
Transcripts
NM_001142854.1 NM_032261.4 XM_005261188.4 XM_011529756.2 XM_017028480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1682074 c.974G>C p.C325S Chr.21: 46161428-46161428 on Build GRCh38
1592796 c.904G>A p.A302T Chr.21: 46161498-46161498 on Build GRCh38
3424008 c.406G>A p.D136N Chr.21: 46161534-46161534 on Build GRCh38
6202774 c.535G>T p.D179Y Chr.21: 46161405-46161405 on Build GRCh38
4667524 c.939delC p.K314fs*>27 Chr.21: 46161463-46161463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921161788 Chr.21: 46161128-46161128 on Build GRCh38
rs146895208 Chr.21: 46161521-46161521 on Build GRCh38
rs550244377 Chr.21: 46161526-46161526 on Build GRCh38
rs1020633757 Chr.21: 46161247-46161247 on Build GRCh38
rs758007206 Chr.21: 46161391-46161391 on Build GRCh38
rs751086468 Chr.21: 46161464-46161464 on Build GRCh38
rs918817255 Chr.21: 46161321-46161321 on Build GRCh38
rs773687923 Chr.21: 46161429-46161429 on Build GRCh38
rs770843218 Chr.21: 46161362-46161362 on Build GRCh38
rs570924423 Chr.21: 46161102-46161102 on Build GRCh38
rs767798726 Chr.21: 46161331-46161331 on Build GRCh38
rs371918100 Chr.21: 46161498-46161498 on Build GRCh38
rs539607226 Chr.21: 46161279-46161279 on Build GRCh38
rs1001003017 Chr.21: 46161174-46161174 on Build GRCh38
rs188622442 Chr.21: 46161103-46161103 on Build GRCh38
rs781647948 Chr.21: 46161543-46161543 on Build GRCh38
rs761070479 Chr.21: 46161431-46161431 on Build GRCh38
rs945043923 Chr.21: 46161168-46161168 on Build GRCh38
rs113781247 Chr.21: 46161270-46161270 on Build GRCh38
rs1017434606 Chr.21: 46161276-46161276 on Build GRCh38
rs779045549 Chr.21: 46161532-46161532 on Build GRCh38
rs1021303353 Chr.21: 46161282-46161282 on Build GRCh38
rs780012775 Chr.21: 46161477-46161477 on Build GRCh38
rs763702250 Chr.21: 46161515-46161515 on Build GRCh38
rs10188 Chr.21: 46161305-46161305 on Build GRCh38
rs138897694 Chr.21: 46161535-46161535 on Build GRCh38
rs573580421 Chr.21: 46161364-46161364 on Build GRCh38
rs949597567 Chr.21: 46161307-46161307 on Build GRCh38
rs559670239 Chr.21: 46161387-46161387 on Build GRCh38
rs949625910 Chr.21: 46161213-46161213 on Build GRCh38
rs777844446 Chr.21: 46161537-46161537 on Build GRCh38
rs749648631 Chr.21: 46161526-46161527 on Build GRCh38
rs965981985 Chr.21: 46161154-46161154 on Build GRCh38
rs1013872751 Chr.21: 46161273-46161273 on Build GRCh38
rs995526199 Chr.21: 46161228-46161228 on Build GRCh38
rs763775638 Chr.21: 46161371-46161372 on Build GRCh38
rs1037699285 Chr.21: 46161223-46161223 on Build GRCh38
rs144453848 Chr.21: 46161099-46161099 on Build GRCh38
rs945321819 Chr.21: 46161250-46161250 on Build GRCh38
rs925258545 Chr.21: 46161316-46161316 on Build GRCh38
rs1055610739 Chr.21: 46161327-46161327 on Build GRCh38
rs1017883445 Chr.21: 46161147-46161147 on Build GRCh38
rs750857637 Chr.21: 46161381-46161381 on Build GRCh38
rs145916215 Chr.21: 46161297-46161298 on Build GRCh38
rs553752275 Chr.21: 46161408-46161408 on Build GRCh38
rs754915276 Chr.21: 46161543-46161543 on Build GRCh38
rs931583394 Chr.21: 46161322-46161322 on Build GRCh38
rs775830808 Chr.21: 46161371-46161371 on Build GRCh38
rs376854767 Chr.21: 46161451-46161451 on Build GRCh38
rs763796724 Chr.21: 46161458-46161458 on Build GRCh38
rs959041011 Chr.21: 46161401-46161401 on Build GRCh38
rs1024003309 Chr.21: 46161198-46161198 on Build GRCh38
rs1015315194 Chr.21: 46161283-46161283 on Build GRCh38
rs749539873 Chr.21: 46161412-46161412 on Build GRCh38
rs895291899 Chr.21: 46161273-46161273 on Build GRCh38
rs755224454 Chr.21: 46161492-46161492 on Build GRCh38
rs781174074 Chr.21: 46161405-46161405 on Build GRCh38
rs1042505533 Chr.21: 46161267-46161268 on Build GRCh38
rs372304217 Chr.21: 46161424-46161424 on Build GRCh38
rs762337855 Chr.21: 46161329-46161329 on Build GRCh38
rs568431352 Chr.21: 46161166-46161166 on Build GRCh38
rs777160651 Chr.21: 46161397-46161397 on Build GRCh38
rs767601092 Chr.21: 46161473-46161473 on Build GRCh38
rs754782891 Chr.21: 46161531-46161531 on Build GRCh38
rs780484264 Chr.21: 46161503-46161515 on Build GRCh38
rs749029733 Chr.21: 46161491-46161491 on Build GRCh38
rs752263570 Chr.21: 46161350-46161350 on Build GRCh38
rs749943211 Chr.21: 46161462-46161463 on Build GRCh38
rs543416913 Chr.21: 46161306-46161306 on Build GRCh38
rs767153721 Chr.21: 46161525-46161525 on Build GRCh38
rs771485808 Chr.21: 46161539-46161539 on Build GRCh38
rs912601659 Chr.21: 46161245-46161245 on Build GRCh38
rs979047712 Chr.21: 46161296-46161296 on Build GRCh38
rs138638304 Chr.21: 46161497-46161497 on Build GRCh38
rs1051124058 Chr.21: 46161130-46161130 on Build GRCh38
rs1007014697 Chr.21: 46161267-46161267 on Build GRCh38
rs535343080 Chr.21: 46161226-46161237 on Build GRCh38
rs982484115 Chr.21: 46161096-46161096 on Build GRCh38
rs145806242 Chr.21: 46161409-46161409 on Build GRCh38
rs533139730 Chr.21: 46161087-46161087 on Build GRCh38
rs977990978 Chr.21: 46161534-46161534 on Build GRCh38
rs1041534795 Chr.21: 46161351-46161351 on Build GRCh38
rs780788956 Chr.21: 46161458-46161458 on Build GRCh38
rs966274675 Chr.21: 46161277-46161277 on Build GRCh38
rs764920210 Chr.21: 46161388-46161388 on Build GRCh38
rs890332029 Chr.21: 46161212-46161221 on Build GRCh38
rs751274695 Chr.21: 46161392-46161393 on Build GRCh38
rs777806451 Chr.21: 46161354-46161354 on Build GRCh38
rs533179150 Chr.21: 46161427-46161427 on Build GRCh38
rs932661295 Chr.21: 46161129-46161129 on Build GRCh38
rs75660802 Chr.21: 46161160-46161160 on Build GRCh38
rs554421787 Chr.21: 46161216-46161216 on Build GRCh38
rs946179848 Chr.21: 46161544-46161544 on Build GRCh38
rs14378 Chr.21: 46161509-46161509 on Build GRCh38
rs746950956 Chr.21: 46161357-46161357 on Build GRCh38
rs1025645809 Chr.21: 46161274-46161274 on Build GRCh38
rs766419736 Chr.21: 46161527-46161527 on Build GRCh38
rs559634035 Chr.21: 46161084-46161084 on Build GRCh38
rs762895438 Chr.21: 46161370-46161370 on Build GRCh38
rs966324341 Chr.21: 46161288-46161288 on Build GRCh38
rs754633900 Chr.21: 46161342-46161342 on Build GRCh38
rs904027187 Chr.21: 46161172-46161172 on Build GRCh38
rs750413687 Chr.21: 46161474-46161474 on Build GRCh38
rs932613126 Chr.21: 46161237-46161237 on Build GRCh38
rs959104756 Chr.21: 46161299-46161299 on Build GRCh38
rs889861121 Chr.21: 46161253-46161253 on Build GRCh38
rs1037263876 Chr.21: 46161311-46161311 on Build GRCh38
rs757884346 Chr.21: 46161352-46161352 on Build GRCh38
rs917678714 Chr.21: 46161450-46161450 on Build GRCh38
rs756781686 Chr.21: 46161433-46161434 on Build GRCh38
rs897127587 Chr.21: 46161224-46161224 on Build GRCh38
rs756766800 Chr.21: 46161471-46161471 on Build GRCh38
rs751015041 Chr.21: 46161402-46161402 on Build GRCh38
rs979121455 Chr.21: 46161189-46161189 on Build GRCh38
rs549067112 Chr.21: 46161083-46161083 on Build GRCh38
rs768869313 Chr.21: 46161420-46161420 on Build GRCh38
rs118185461 Chr.21: 46161298-46161298 on Build GRCh38
rs937784933 Chr.21: 46161199-46161199 on Build GRCh38
rs373560114 Chr.21: 46161374-46161374 on Build GRCh38
rs1033946632 Chr.21: 46161178-46161178 on Build GRCh38
rs761329406 Chr.21: 46161522-46161522 on Build GRCh38
rs771359378 Chr.21: 46161503-46161503 on Build GRCh38
rs997408047 Chr.21: 46161281-46161281 on Build GRCh38
rs368311014 Chr.21: 46161403-46161403 on Build GRCh38
rs895357709 Chr.21: 46161373-46161373 on Build GRCh38
rs745652450 Chr.21: 46161365-46161365 on Build GRCh38
rs778535913 Chr.21: 46161344-46161344 on Build GRCh38
rs531739944 Chr.21: 46161111-46161111 on Build GRCh38
rs772515651 Chr.21: 46161425-46161425 on Build GRCh38
rs555111242 Chr.21: 46161361-46161361 on Build GRCh38
rs779186054 Chr.21: 46161496-46161496 on Build GRCh38
rs747086694 Chr.21: 46161538-46161538 on Build GRCh38
rs918120920 Chr.21: 46161206-46161206 on Build GRCh38
rs750745856 Chr.21: 46161341-46161341 on Build GRCh38
rs569190109 Chr.21: 46161167-46161167 on Build GRCh38
rs762516693 Chr.21: 46161453-46161453 on Build GRCh38
rs770195780 Chr.21: 46161368-46161368 on Build GRCh38
rs1055205393 Chr.21: 46161200-46161200 on Build GRCh38
rs774444137 Chr.21: 46161375-46161375 on Build GRCh38
rs535818337 Chr.21: 46161192-46161192 on Build GRCh38
rs774303965 Chr.21: 46161421-46161421 on Build GRCh38
rs745703384 Chr.21: 46161407-46161407 on Build GRCh38
rs747537035 Chr.21: 46161499-46161499 on Build GRCh38
rs375786023 Chr.21: 46161376-46161376 on Build GRCh38
rs537607373 Chr.21: 46161182-46161182 on Build GRCh38
rs756622846 Chr.21: 46161473-46161481 on Build GRCh38
rs1055322033 Chr.21: 46161271-46161271 on Build GRCh38
rs565965239 Chr.21: 46161240-46161240 on Build GRCh38
rs200257589 Chr.21: 46161475-46161475 on Build GRCh38

View Full Product Details