Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46315395-46315899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf58
Entrez Gene ID
54058
Gene Name
chromosome 21 open reading frame 58
Gene Aliases
-
UniGene
Hs.236572
Gene Chromosome Location
Chr.21: 46300426-46323899 on Build GRCh38
Transcripts
NM_001286462.1 NM_001286463.1 NM_001286476.1 NM_001286477.1 NM_058180.4 XM_005261148.4 XM_006724018.3 XM_011529619.2 XM_011529620.2 XM_011529621.2 XM_011529623.2 XM_011529624.2 XM_011529625.2 XM_011529626.2 XM_011529627.2 XM_011529628.1 XM_017028388.1 XM_017028389.1 XM_017028390.1 XM_017028391.1 XM_017028392.1 XM_017028393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6468022 c.168A>G p.R56R Chr.21: 46315501-46315501 on Build GRCh38
1751699 c.411G>A p.L137L Chr.21: 46315507-46315507 on Build GRCh38
4932396 c.391G>A p.D131N Chr.21: 46315527-46315527 on Build GRCh38
6319300 c.141C>A p.D47E Chr.21: 46315528-46315528 on Build GRCh38
6468021 c.417A>G p.R139R Chr.21: 46315501-46315501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780539479 Chr.21: 46315495-46315495 on Build GRCh38
rs199568321 Chr.21: 46315539-46315539 on Build GRCh38
rs750976326 Chr.21: 46315464-46315464 on Build GRCh38
rs780158659 Chr.21: 46315596-46315596 on Build GRCh38
rs1012804284 Chr.21: 46315790-46315790 on Build GRCh38
rs372981482 Chr.21: 46315584-46315584 on Build GRCh38
rs77907804 Chr.21: 46315531-46315531 on Build GRCh38
rs941453481 Chr.21: 46315587-46315587 on Build GRCh38
rs554770311 Chr.21: 46315609-46315609 on Build GRCh38
rs567040784 Chr.21: 46315525-46315525 on Build GRCh38
rs754304468 Chr.21: 46315561-46315561 on Build GRCh38
rs1019994009 Chr.21: 46315524-46315524 on Build GRCh38
rs1011928101 Chr.21: 46315827-46315827 on Build GRCh38
rs1041132238 Chr.21: 46315466-46315466 on Build GRCh38
rs367689378 Chr.21: 46315432-46315432 on Build GRCh38
rs1054153992 Chr.21: 46315808-46315808 on Build GRCh38
rs779598056 Chr.21: 46315499-46315501 on Build GRCh38
rs755621551 Chr.21: 46315595-46315595 on Build GRCh38
rs117451772 Chr.21: 46315555-46315555 on Build GRCh38
rs146598443 Chr.21: 46315535-46315535 on Build GRCh38
rs902929240 Chr.21: 46315668-46315668 on Build GRCh38
rs1032676523 Chr.21: 46315725-46315725 on Build GRCh38
rs770525970 Chr.21: 46315491-46315491 on Build GRCh38
rs545422279 Chr.21: 46315826-46315826 on Build GRCh38
rs563983617 Chr.21: 46315829-46315829 on Build GRCh38
rs1038352600 Chr.21: 46315606-46315606 on Build GRCh38
rs56122748 Chr.21: 46315597-46315597 on Build GRCh38
rs766875269 Chr.21: 46315558-46315558 on Build GRCh38
rs531128272 Chr.21: 46315445-46315445 on Build GRCh38
rs911454257 Chr.21: 46315676-46315676 on Build GRCh38
rs891255423 Chr.21: 46315641-46315641 on Build GRCh38
rs562272074 Chr.21: 46315631-46315631 on Build GRCh38
rs1046059128 Chr.21: 46315844-46315844 on Build GRCh38
rs770696754 Chr.21: 46315485-46315485 on Build GRCh38
rs1041419037 Chr.21: 46315656-46315656 on Build GRCh38
rs548693910 Chr.21: 46315451-46315451 on Build GRCh38
rs1053698392 Chr.21: 46315536-46315536 on Build GRCh38
rs761972596 Chr.21: 46315459-46315459 on Build GRCh38
rs369023065 Chr.21: 46315581-46315581 on Build GRCh38
rs2839210 Chr.21: 46315821-46315821 on Build GRCh38
rs202233624 Chr.21: 46315422-46315422 on Build GRCh38
rs185555820 Chr.21: 46315528-46315528 on Build GRCh38
rs750080257 Chr.21: 46315667-46315667 on Build GRCh38
rs976711260 Chr.21: 46315708-46315708 on Build GRCh38
rs536435946 Chr.21: 46315605-46315605 on Build GRCh38
rs377051341 Chr.21: 46315532-46315532 on Build GRCh38
rs745887032 Chr.21: 46315515-46315515 on Build GRCh38
rs574203343 Chr.21: 46315630-46315630 on Build GRCh38
rs757990683 Chr.21: 46315753-46315753 on Build GRCh38
rs971198010 Chr.21: 46315804-46315804 on Build GRCh38
rs543823697 Chr.21: 46315655-46315655 on Build GRCh38
rs999926796 Chr.21: 46315686-46315686 on Build GRCh38
rs370242656 Chr.21: 46315452-46315452 on Build GRCh38
rs541489854 Chr.21: 46315634-46315634 on Build GRCh38
rs946102456 Chr.21: 46315769-46315769 on Build GRCh38
rs1015458647 Chr.21: 46315813-46315813 on Build GRCh38
rs994294060 Chr.21: 46315870-46315870 on Build GRCh38
rs779787497 Chr.21: 46315589-46315589 on Build GRCh38
rs964052172 Chr.21: 46315699-46315699 on Build GRCh38
rs575376842 Chr.21: 46315582-46315582 on Build GRCh38
rs923801090 Chr.21: 46315794-46315794 on Build GRCh38
rs953003216 Chr.21: 46315635-46315635 on Build GRCh38
rs372055017 Chr.21: 46315504-46315504 on Build GRCh38
rs188324762 Chr.21: 46315545-46315545 on Build GRCh38
rs780815534 Chr.21: 46315583-46315583 on Build GRCh38
rs755358687 Chr.21: 46315565-46315565 on Build GRCh38
rs746593157 Chr.21: 46315488-46315488 on Build GRCh38
rs769845183 Chr.21: 46315521-46315521 on Build GRCh38
rs971229178 Chr.21: 46315574-46315574 on Build GRCh38
rs368720497 Chr.21: 46315471-46315471 on Build GRCh38
rs77475641 Chr.21: 46315848-46315848 on Build GRCh38
rs780452434 Chr.21: 46315475-46315475 on Build GRCh38
rs932908366 Chr.21: 46315610-46315610 on Build GRCh38
rs922694896 Chr.21: 46315726-46315726 on Build GRCh38
rs779127294 Chr.21: 46315576-46315576 on Build GRCh38
rs775425940 Chr.21: 46315437-46315437 on Build GRCh38
rs140707382 Chr.21: 46315476-46315476 on Build GRCh38
rs936500895 Chr.21: 46315802-46315802 on Build GRCh38
rs958650445 Chr.21: 46315780-46315780 on Build GRCh38
rs374616450 Chr.21: 46315460-46315460 on Build GRCh38
rs779640675 Chr.21: 46315789-46315789 on Build GRCh38
rs563767946 Chr.21: 46315416-46315416 on Build GRCh38

View Full Product Details