Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21698195-21698737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL2
Entrez Gene ID
23759
Gene Name
peptidylprolyl isomerase like 2
Gene Aliases
CYC4, CYP60, Cyp-60, UBOX7, hCyP-60
UniGene
Hs.438587
Gene Chromosome Location
Chr.22: 21665984-21698276 on Build GRCh38
Transcripts
XM_005261448.3 XM_011530041.2 XM_011530042.2 XM_011530043.2 XM_011530044.2 XM_011530046.2 XM_011530047.2 XM_011530049.2 XM_011530050.2 XM_011530051.2 XM_017028705.1 XM_017028706.1 NM_013313.4 NR_130910.1

Gene Symbol
YPEL1
Entrez Gene ID
29799
Gene Name
yippee like 1
Gene Aliases
FKSG3
UniGene
Hs.517436
Gene Chromosome Location
Chr.22: 21697536-21735834 on Build GRCh38
Transcripts
XM_005261448.3 XM_011530041.2 XM_011530042.2 XM_011530043.2 XM_011530044.2 XM_011530046.2 XM_011530047.2 XM_011530049.2 XM_011530050.2 XM_011530051.2 XM_017028705.1 XM_017028706.1 NM_013313.4 NR_130910.1

SNP Detail

SNP ID Chromosome Location
rs868029386 Chr.22: 21698624-21698624 on Build GRCh38
rs894270216 Chr.22: 21698694-21698694 on Build GRCh38
rs893989068 Chr.22: 21698672-21698672 on Build GRCh38
rs553065026 Chr.22: 21698589-21698589 on Build GRCh38
rs964263817 Chr.22: 21698592-21698592 on Build GRCh38
rs1025216962 Chr.22: 21698345-21698345 on Build GRCh38
rs766143202 Chr.22: 21698640-21698640 on Build GRCh38
rs1046018472 Chr.22: 21698699-21698699 on Build GRCh38
rs976043168 Chr.22: 21698623-21698623 on Build GRCh38
rs59474281 Chr.22: 21698224-21698225 on Build GRCh38
rs966792555 Chr.22: 21698685-21698685 on Build GRCh38
rs35100577 Chr.22: 21698262-21698262 on Build GRCh38
rs374353985 Chr.22: 21698610-21698610 on Build GRCh38
rs933605208 Chr.22: 21698626-21698626 on Build GRCh38
rs902938336 Chr.22: 21698496-21698496 on Build GRCh38
rs1021083666 Chr.22: 21698684-21698684 on Build GRCh38
rs537338958 Chr.22: 21698223-21698224 on Build GRCh38
rs747091615 Chr.22: 21698484-21698485 on Build GRCh38
rs535901458 Chr.22: 21698559-21698559 on Build GRCh38
rs556538105 Chr.22: 21698545-21698545 on Build GRCh38
rs1005729609 Chr.22: 21698309-21698310 on Build GRCh38
rs185976908 Chr.22: 21698229-21698229 on Build GRCh38
rs571896708 Chr.22: 21698503-21698503 on Build GRCh38
rs984249060 Chr.22: 21698376-21698376 on Build GRCh38
rs982131613 Chr.22: 21698322-21698322 on Build GRCh38
rs1045681 Chr.22: 21698525-21698525 on Build GRCh38
rs775615313 Chr.22: 21698396-21698396 on Build GRCh38
rs1012863751 Chr.22: 21698281-21698281 on Build GRCh38
rs751216679 Chr.22: 21698641-21698641 on Build GRCh38
rs59916934 Chr.22: 21698328-21698328 on Build GRCh38
rs80166899 Chr.22: 21698261-21698261 on Build GRCh38
rs941104733 Chr.22: 21698491-21698491 on Build GRCh38
rs951508543 Chr.22: 21698341-21698341 on Build GRCh38
rs201784522 Chr.22: 21698225-21698226 on Build GRCh38
rs148651668 Chr.22: 21698261-21698261 on Build GRCh38
rs1054276931 Chr.22: 21698509-21698509 on Build GRCh38
rs984201680 Chr.22: 21698574-21698574 on Build GRCh38
rs894332168 Chr.22: 21698518-21698518 on Build GRCh38
rs948587593 Chr.22: 21698695-21698695 on Build GRCh38
rs985287500 Chr.22: 21698448-21698448 on Build GRCh38
rs910088238 Chr.22: 21698388-21698388 on Build GRCh38
rs545301319 Chr.22: 21698693-21698693 on Build GRCh38
rs1036626295 Chr.22: 21698317-21698317 on Build GRCh38
rs970771160 Chr.22: 21698321-21698321 on Build GRCh38
rs371553316 Chr.22: 21698540-21698540 on Build GRCh38
rs753565922 Chr.22: 21698261-21698262 on Build GRCh38
rs192020136 Chr.22: 21698408-21698408 on Build GRCh38
rs564202159 Chr.22: 21698238-21698239 on Build GRCh38
rs573445454 Chr.22: 21698556-21698556 on Build GRCh38
rs942935236 Chr.22: 21698411-21698411 on Build GRCh38
rs764371381 Chr.22: 21698519-21698519 on Build GRCh38
rs932562637 Chr.22: 21698441-21698442 on Build GRCh38
rs935717272 Chr.22: 21698678-21698678 on Build GRCh38
rs754626227 Chr.22: 21698644-21698644 on Build GRCh38
rs1041007343 Chr.22: 21698226-21698226 on Build GRCh38
rs924302318 Chr.22: 21698654-21698654 on Build GRCh38
rs397934469 Chr.22: 21698276-21698277 on Build GRCh38
rs553133162 Chr.22: 21698560-21698560 on Build GRCh38
rs1010993021 Chr.22: 21698677-21698678 on Build GRCh38
rs184186646 Chr.22: 21698428-21698428 on Build GRCh38
rs4821188 Chr.22: 21698260-21698260 on Build GRCh38
rs1002339107 Chr.22: 21698355-21698356 on Build GRCh38
rs763122239 Chr.22: 21698426-21698426 on Build GRCh38
rs757559672 Chr.22: 21698627-21698627 on Build GRCh38
rs1034283130 Chr.22: 21698371-21698371 on Build GRCh38
rs201411822 Chr.22: 21698279-21698279 on Build GRCh38
rs762746860 Chr.22: 21698551-21698551 on Build GRCh38
rs933685398 Chr.22: 21698429-21698429 on Build GRCh38
rs1030094272 Chr.22: 21698712-21698712 on Build GRCh38
rs932534551 Chr.22: 21698292-21698292 on Build GRCh38
rs748034791 Chr.22: 21698329-21698330 on Build GRCh38
rs540858491 Chr.22: 21698497-21698497 on Build GRCh38
rs971397505 Chr.22: 21698349-21698349 on Build GRCh38
rs397780396 Chr.22: 21698232-21698233 on Build GRCh38
rs1054224011 Chr.22: 21698686-21698686 on Build GRCh38
rs529401630 Chr.22: 21698324-21698324 on Build GRCh38
rs1024038116 Chr.22: 21698287-21698287 on Build GRCh38
rs949689656 Chr.22: 21698523-21698523 on Build GRCh38
rs1036763350 Chr.22: 21698500-21698500 on Build GRCh38
rs1050032968 Chr.22: 21698293-21698293 on Build GRCh38
rs909718695 Chr.22: 21698475-21698475 on Build GRCh38
rs910025406 Chr.22: 21698579-21698579 on Build GRCh38
rs572920750 Chr.22: 21698590-21698590 on Build GRCh38
rs200298559 Chr.22: 21698223-21698223 on Build GRCh38
rs556992705 Chr.22: 21698301-21698301 on Build GRCh38
rs1041454463 Chr.22: 21698459-21698459 on Build GRCh38
rs1024237601 Chr.22: 21698537-21698537 on Build GRCh38
rs896852475 Chr.22: 21698326-21698326 on Build GRCh38
rs977082339 Chr.22: 21698653-21698653 on Build GRCh38
rs966739999 Chr.22: 21698412-21698412 on Build GRCh38
rs777951916 Chr.22: 21698471-21698472 on Build GRCh38
rs990001962 Chr.22: 21698400-21698400 on Build GRCh38
rs958120166 Chr.22: 21698378-21698378 on Build GRCh38

View Full Product Details