Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21735511-21736031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL1
Entrez Gene ID
29799
Gene Name
yippee like 1
Gene Aliases
FKSG3
UniGene
Hs.517436
Gene Chromosome Location
Chr.22: 21697536-21735834 on Build GRCh38
Transcripts
NM_013313.4 NR_130910.1

SNP Detail

SNP ID Chromosome Location
rs545185818 Chr.22: 21736004-21736004 on Build GRCh38
rs914082205 Chr.22: 21735565-21735565 on Build GRCh38
rs542788045 Chr.22: 21735898-21735898 on Build GRCh38
rs980127875 Chr.22: 21735598-21735598 on Build GRCh38
rs934109443 Chr.22: 21735964-21735964 on Build GRCh38
rs1016962691 Chr.22: 21735822-21735822 on Build GRCh38
rs937664308 Chr.22: 21735603-21735603 on Build GRCh38
rs898361032 Chr.22: 21735678-21735678 on Build GRCh38
rs962865695 Chr.22: 21735735-21735736 on Build GRCh38
rs1021136936 Chr.22: 21735593-21735593 on Build GRCh38
rs556885197 Chr.22: 21735615-21735615 on Build GRCh38
rs926275193 Chr.22: 21735602-21735602 on Build GRCh38
rs1007000353 Chr.22: 21735975-21735975 on Build GRCh38
rs988256894 Chr.22: 21735560-21735560 on Build GRCh38
rs924103831 Chr.22: 21735961-21735961 on Build GRCh38
rs966156265 Chr.22: 21735546-21735546 on Build GRCh38
rs968327822 Chr.22: 21735580-21735580 on Build GRCh38
rs1036848046 Chr.22: 21735668-21735668 on Build GRCh38
rs981242453 Chr.22: 21735872-21735872 on Build GRCh38
rs998304205 Chr.22: 21735606-21735606 on Build GRCh38
rs998671320 Chr.22: 21735800-21735800 on Build GRCh38
rs972951911 Chr.22: 21735733-21735733 on Build GRCh38
rs927011157 Chr.22: 21735883-21735883 on Build GRCh38
rs917686315 Chr.22: 21735624-21735624 on Build GRCh38
rs1042681108 Chr.22: 21735928-21735928 on Build GRCh38
rs889878848 Chr.22: 21735808-21735808 on Build GRCh38
rs55690117 Chr.22: 21735973-21735973 on Build GRCh38
rs894033872 Chr.22: 21735754-21735754 on Build GRCh38
rs938256862 Chr.22: 21735559-21735559 on Build GRCh38
rs553103779 Chr.22: 21735948-21735948 on Build GRCh38
rs1006865831 Chr.22: 21735809-21735809 on Build GRCh38
rs936967300 Chr.22: 21735751-21735751 on Build GRCh38
rs947007826 Chr.22: 21735755-21735755 on Build GRCh38
rs991189609 Chr.22: 21735857-21735857 on Build GRCh38
rs926958450 Chr.22: 21735750-21735750 on Build GRCh38
rs939780110 Chr.22: 21735664-21735664 on Build GRCh38
rs962669644 Chr.22: 21735726-21735726 on Build GRCh38
rs1050113431 Chr.22: 21735697-21735697 on Build GRCh38
rs1029793691 Chr.22: 21735803-21735803 on Build GRCh38
rs905566010 Chr.22: 21735551-21735551 on Build GRCh38
rs139967375 Chr.22: 21735567-21735568 on Build GRCh38
rs1019006529 Chr.22: 21735531-21735531 on Build GRCh38
rs573533346 Chr.22: 21735843-21735843 on Build GRCh38
rs1016825833 Chr.22: 21735723-21735723 on Build GRCh38
rs962927052 Chr.22: 21735828-21735828 on Build GRCh38
rs981190263 Chr.22: 21735745-21735745 on Build GRCh38
rs989752469 Chr.22: 21735910-21735910 on Build GRCh38
rs910073935 Chr.22: 21735698-21735713 on Build GRCh38
rs949825359 Chr.22: 21735739-21735739 on Build GRCh38
rs995426843 Chr.22: 21735679-21735679 on Build GRCh38
rs1042932095 Chr.22: 21735758-21735758 on Build GRCh38
rs942316780 Chr.22: 21735855-21735855 on Build GRCh38
rs1025519373 Chr.22: 21735998-21735998 on Build GRCh38
rs914267516 Chr.22: 21735916-21735916 on Build GRCh38
rs902712053 Chr.22: 21735760-21735760 on Build GRCh38
rs898530022 Chr.22: 21735985-21735985 on Build GRCh38
rs949420364 Chr.22: 21735981-21735981 on Build GRCh38
rs946945971 Chr.22: 21735926-21735926 on Build GRCh38

View Full Product Details