Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50406556-50407055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP6R2
Entrez Gene ID
9701
Gene Name
protein phosphatase 6 regulatory subunit 2
Gene Aliases
KIAA0685, PP6R2, SAP190, SAPS2
UniGene
Hs.449098
Gene Chromosome Location
Chr.22: 50343317-50445090 on Build GRCh38
Transcripts
NM_001242898.1 NM_001242899.1 NM_001242900.1 NM_014678.4 XM_006724431.2 XM_006724434.1 XM_006724435.2 XM_011530720.2 XM_011530721.2 XM_011530722.2 XM_011530723.2 XM_011530724.2 XM_011530726.2 XM_011530727.2 XM_011530728.2 XM_011530729.2 XM_011530730.2 XM_011530731.1 XM_011530732.2 XM_011530734.2 XM_011530735.2 XM_011530736.2 XM_011530737.2 XM_011530738.2 XM_011530739.2 XM_011530740.2 XM_017029116.1 XM_017029117.1 XM_017029118.1 XM_017029119.1 XM_017029120.1 XM_017029121.1 XM_017029122.1 XM_017029123.1 XM_017029124.1 XM_017029125.1 XM_017029126.1 XM_017029127.1 XM_017029128.1 XM_017029129.1 XM_017029130.1 XM_017029131.1 XM_017029132.1 XM_017029133.1 XM_017029134.1 XM_017029135.1 XM_017029136.1 XM_017029137.1 XM_017029138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3555716 c.360C>T p.L120L Chr.22: 50406821-50406821 on Build GRCh38
6989477 c.360C>T p.L120L Chr.22: 50406821-50406821 on Build GRCh38
4543064 c.330G>A p.L110L Chr.22: 50406791-50406791 on Build GRCh38
3555715 c.334C>T p.H112Y Chr.22: 50406795-50406795 on Build GRCh38
7145824 c.273G>C p.Q91H Chr.22: 50406734-50406734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200665748 Chr.22: 50406749-50406749 on Build GRCh38
rs781341069 Chr.22: 50406869-50406869 on Build GRCh38
rs146238959 Chr.22: 50406844-50406844 on Build GRCh38
rs761892345 Chr.22: 50406669-50406669 on Build GRCh38
rs982606621 Chr.22: 50406896-50406896 on Build GRCh38
rs775531335 Chr.22: 50406893-50406893 on Build GRCh38
rs556612016 Chr.22: 50406731-50406731 on Build GRCh38
rs776394521 Chr.22: 50406913-50406913 on Build GRCh38
rs759430979 Chr.22: 50406776-50406776 on Build GRCh38
rs371735944 Chr.22: 50406730-50406730 on Build GRCh38
rs760525478 Chr.22: 50406787-50406787 on Build GRCh38
rs748415912 Chr.22: 50406870-50406870 on Build GRCh38
rs752363053 Chr.22: 50406725-50406725 on Build GRCh38
rs779515905 Chr.22: 50406741-50406741 on Build GRCh38
rs749400087 Chr.22: 50406884-50406884 on Build GRCh38
rs773346424 Chr.22: 50406880-50406880 on Build GRCh38
rs770533653 Chr.22: 50406972-50406972 on Build GRCh38
rs990587307 Chr.22: 50406772-50406772 on Build GRCh38
rs780346897 Chr.22: 50406747-50406747 on Build GRCh38
rs142637497 Chr.22: 50406860-50406860 on Build GRCh38
rs535431550 Chr.22: 50406794-50406794 on Build GRCh38
rs544650926 Chr.22: 50406596-50406596 on Build GRCh38
rs768773457 Chr.22: 50406651-50406651 on Build GRCh38
rs763801859 Chr.22: 50406788-50406788 on Build GRCh38
rs534177686 Chr.22: 50406975-50406975 on Build GRCh38
rs769883291 Chr.22: 50406759-50406759 on Build GRCh38
rs754402582 Chr.22: 50406592-50406592 on Build GRCh38
rs922543699 Chr.22: 50406609-50406609 on Build GRCh38
rs141219176 Chr.22: 50406822-50406822 on Build GRCh38
rs550603081 Chr.22: 50406802-50406802 on Build GRCh38
rs1033085738 Chr.22: 50406593-50406604 on Build GRCh38
rs1046193536 Chr.22: 50406951-50406951 on Build GRCh38
rs148219827 Chr.22: 50406774-50406774 on Build GRCh38
rs573807877 Chr.22: 50407007-50407007 on Build GRCh38
rs751528552 Chr.22: 50406710-50406710 on Build GRCh38
rs999950792 Chr.22: 50406807-50406807 on Build GRCh38
rs189576394 Chr.22: 50406952-50406952 on Build GRCh38
rs1001065188 Chr.22: 50406998-50406998 on Build GRCh38
rs374838213 Chr.22: 50406922-50406922 on Build GRCh38
rs933835714 Chr.22: 50406628-50406628 on Build GRCh38
rs541685510 Chr.22: 50406758-50406758 on Build GRCh38
rs778435808 Chr.22: 50406605-50406605 on Build GRCh38
rs143049560 Chr.22: 50406744-50406744 on Build GRCh38
rs544156019 Chr.22: 50407011-50407011 on Build GRCh38
rs760340312 Chr.22: 50406894-50406894 on Build GRCh38
rs551922980 Chr.22: 50406635-50406635 on Build GRCh38
rs776551737 Chr.22: 50406821-50406821 on Build GRCh38
rs764033296 Chr.22: 50406898-50406898 on Build GRCh38
rs182043670 Chr.22: 50406968-50406968 on Build GRCh38
rs570640598 Chr.22: 50406665-50406665 on Build GRCh38
rs766234809 Chr.22: 50406677-50406677 on Build GRCh38
rs910483541 Chr.22: 50406845-50406845 on Build GRCh38
rs760850500 Chr.22: 50406640-50406640 on Build GRCh38
rs1003098347 Chr.22: 50406633-50406633 on Build GRCh38
rs759324175 Chr.22: 50406720-50406720 on Build GRCh38
rs558201880 Chr.22: 50406961-50406961 on Build GRCh38
rs941409321 Chr.22: 50406930-50406930 on Build GRCh38
rs12160914 Chr.22: 50406920-50406920 on Build GRCh38
rs373762887 Chr.22: 50407001-50407001 on Build GRCh38
rs764883823 Chr.22: 50406803-50406803 on Build GRCh38
rs770259187 Chr.22: 50406956-50406956 on Build GRCh38
rs777425517 Chr.22: 50406740-50406740 on Build GRCh38
rs1022007451 Chr.22: 50406974-50406974 on Build GRCh38
rs896675099 Chr.22: 50407030-50407030 on Build GRCh38
rs769834075 Chr.22: 50406872-50406872 on Build GRCh38
rs761628277 Chr.22: 50406916-50406916 on Build GRCh38
rs1038409769 Chr.22: 50406946-50406946 on Build GRCh38
rs1011577109 Chr.22: 50406973-50406973 on Build GRCh38
rs768700418 Chr.22: 50406750-50406750 on Build GRCh38
rs752377143 Chr.22: 50406783-50406783 on Build GRCh38
rs967476730 Chr.22: 50406782-50406782 on Build GRCh38
rs897597979 Chr.22: 50406590-50406590 on Build GRCh38
rs555824668 Chr.22: 50407000-50407000 on Build GRCh38
rs769818706 Chr.22: 50406841-50406841 on Build GRCh38
rs907379145 Chr.22: 50406617-50406617 on Build GRCh38
rs371424294 Chr.22: 50406779-50406779 on Build GRCh38
rs1034597279 Chr.22: 50406957-50406957 on Build GRCh38
rs184342780 Chr.22: 50406597-50406597 on Build GRCh38
rs201294162 Chr.22: 50406722-50406722 on Build GRCh38
rs200097055 Chr.22: 50406885-50406885 on Build GRCh38
rs772455624 Chr.22: 50406745-50406745 on Build GRCh38

View Full Product Details