Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74098708-74099211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL2
Entrez Gene ID
2920
Gene Name
C-X-C motif chemokine ligand 2
Gene Aliases
CINC-2a, GRO2, GROb, MGSA-b, MIP-2a, MIP2, MIP2A, SCYB2
UniGene
Hs.75765
Gene Chromosome Location
Chr.4: 74097035-74099280 on Build GRCh38
Transcripts
NM_002089.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6405351 c.55G>T p.A19S Chr.4: 74099066-74099066 on Build GRCh38
6431076 c.94G>T p.A32S Chr.4: 74099027-74099027 on Build GRCh38
6365381 c.33C>A p.S11R Chr.4: 74099088-74099088 on Build GRCh38
6371159 c.5C>T p.A2V Chr.4: 74099116-74099116 on Build GRCh38
5484712 c.216C>G p.T72T Chr.4: 74098807-74098807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779690676 Chr.4: 74098995-74098995 on Build GRCh38
rs774403935 Chr.4: 74098920-74098920 on Build GRCh38
rs575971969 Chr.4: 74098918-74098918 on Build GRCh38
rs561874420 Chr.4: 74098952-74098952 on Build GRCh38
rs911370769 Chr.4: 74098970-74098970 on Build GRCh38
rs370906034 Chr.4: 74098986-74098986 on Build GRCh38
rs200911424 Chr.4: 74098934-74098934 on Build GRCh38
rs186397980 Chr.4: 74099113-74099113 on Build GRCh38
rs201534860 Chr.4: 74098729-74098729 on Build GRCh38
rs907204082 Chr.4: 74099033-74099033 on Build GRCh38
rs780318567 Chr.4: 74099043-74099043 on Build GRCh38
rs200356891 Chr.4: 74098957-74098957 on Build GRCh38
rs762380109 Chr.4: 74098828-74098828 on Build GRCh38
rs200324100 Chr.4: 74099000-74099000 on Build GRCh38
rs199529315 Chr.4: 74098768-74098768 on Build GRCh38
rs758440195 Chr.4: 74098874-74098874 on Build GRCh38
rs754644763 Chr.4: 74099064-74099064 on Build GRCh38
rs918820091 Chr.4: 74098800-74098800 on Build GRCh38
rs201152585 Chr.4: 74099039-74099039 on Build GRCh38
rs1025225411 Chr.4: 74099108-74099108 on Build GRCh38
rs553596447 Chr.4: 74098773-74098773 on Build GRCh38
rs200468668 Chr.4: 74098926-74098926 on Build GRCh38
rs755085970 Chr.4: 74098779-74098779 on Build GRCh38
rs199782155 Chr.4: 74099051-74099051 on Build GRCh38
rs372337678 Chr.4: 74098819-74098819 on Build GRCh38
rs746161349 Chr.4: 74098956-74098956 on Build GRCh38
rs895092701 Chr.4: 74099071-74099071 on Build GRCh38
rs201739633 Chr.4: 74099171-74099171 on Build GRCh38
rs771730846 Chr.4: 74099100-74099100 on Build GRCh38
rs377663383 Chr.4: 74098963-74098963 on Build GRCh38
rs759391615 Chr.4: 74099152-74099152 on Build GRCh38
rs778843350 Chr.4: 74098863-74098863 on Build GRCh38
rs200992030 Chr.4: 74098922-74098922 on Build GRCh38
rs778295766 Chr.4: 74098876-74098876 on Build GRCh38
rs776739890 Chr.4: 74098916-74098916 on Build GRCh38
rs757336342 Chr.4: 74099010-74099010 on Build GRCh38
rs939011729 Chr.4: 74099015-74099015 on Build GRCh38
rs1011969879 Chr.4: 74098940-74098940 on Build GRCh38
rs202120861 Chr.4: 74098924-74098924 on Build GRCh38
rs777759075 Chr.4: 74098941-74098941 on Build GRCh38
rs202016804 Chr.4: 74099136-74099136 on Build GRCh38
rs898242441 Chr.4: 74099097-74099097 on Build GRCh38
rs889147158 Chr.4: 74099125-74099125 on Build GRCh38
rs199906828 Chr.4: 74098865-74098865 on Build GRCh38
rs764022735 Chr.4: 74098829-74098829 on Build GRCh38
rs1045781617 Chr.4: 74098960-74098960 on Build GRCh38
rs775126279 Chr.4: 74098826-74098826 on Build GRCh38
rs1019027622 Chr.4: 74099145-74099145 on Build GRCh38
rs926258893 Chr.4: 74099013-74099013 on Build GRCh38
rs767468602 Chr.4: 74098845-74098845 on Build GRCh38
rs767969307 Chr.4: 74099016-74099016 on Build GRCh38
rs368146276 Chr.4: 74098816-74098816 on Build GRCh38
rs770759328 Chr.4: 74099119-74099119 on Build GRCh38
rs747593102 Chr.4: 74099081-74099081 on Build GRCh38
rs781608529 Chr.4: 74098901-74098901 on Build GRCh38
rs199778373 Chr.4: 74098991-74098991 on Build GRCh38
rs200931429 Chr.4: 74099170-74099170 on Build GRCh38
rs904789861 Chr.4: 74099074-74099074 on Build GRCh38
rs778719789 Chr.4: 74099070-74099070 on Build GRCh38
rs750947723 Chr.4: 74099028-74099028 on Build GRCh38
rs1016805734 Chr.4: 74099088-74099088 on Build GRCh38
rs914624257 Chr.4: 74099018-74099018 on Build GRCh38
rs761826681 Chr.4: 74098975-74098975 on Build GRCh38
rs200663834 Chr.4: 74099133-74099133 on Build GRCh38
rs1003468343 Chr.4: 74098892-74098892 on Build GRCh38
rs199924148 Chr.4: 74099174-74099174 on Build GRCh38
rs748528541 Chr.4: 74099152-74099154 on Build GRCh38
rs201552328 Chr.4: 74098820-74098820 on Build GRCh38
rs892211709 Chr.4: 74099029-74099029 on Build GRCh38
rs374705558 Chr.4: 74098989-74098989 on Build GRCh38
rs773598787 Chr.4: 74098809-74098809 on Build GRCh38
rs775888628 Chr.4: 74099186-74099186 on Build GRCh38
rs748937638 Chr.4: 74098962-74098962 on Build GRCh38
rs934282215 Chr.4: 74098802-74098802 on Build GRCh38
rs371945474 Chr.4: 74098762-74098762 on Build GRCh38
rs564762931 Chr.4: 74099158-74099158 on Build GRCh38
rs765461293 Chr.4: 74098933-74098933 on Build GRCh38
rs200665196 Chr.4: 74098777-74098777 on Build GRCh38
rs200151384 Chr.4: 74099162-74099162 on Build GRCh38
rs148727820 Chr.4: 74098855-74098855 on Build GRCh38
rs972368588 Chr.4: 74098943-74098943 on Build GRCh38
rs201486986 Chr.4: 74098860-74098860 on Build GRCh38
rs779089045 Chr.4: 74098789-74098789 on Build GRCh38
rs971877752 Chr.4: 74099160-74099160 on Build GRCh38
rs946154120 Chr.4: 74099026-74099026 on Build GRCh38
rs769433487 Chr.4: 74098909-74098909 on Build GRCh38
rs757118828 Chr.4: 74098764-74098764 on Build GRCh38
rs751478943 Chr.4: 74098831-74098831 on Build GRCh38
rs142264518 Chr.4: 74098908-74098908 on Build GRCh38
rs760397923 Chr.4: 74098931-74098931 on Build GRCh38
rs775008091 Chr.4: 74098914-74098914 on Build GRCh38
rs1041704227 Chr.4: 74099027-74099027 on Build GRCh38
rs879072796 Chr.4: 74099126-74099126 on Build GRCh38
rs918287928 Chr.4: 74098947-74098947 on Build GRCh38
rs771258965 Chr.4: 74098894-74098894 on Build GRCh38
rs764651890 Chr.4: 74098825-74098825 on Build GRCh38
rs568060950 Chr.4: 74098749-74098749 on Build GRCh38
rs755316953 Chr.4: 74098858-74098858 on Build GRCh38
rs758998062 Chr.4: 74098727-74098727 on Build GRCh38
rs368385936 Chr.4: 74098753-74098753 on Build GRCh38
rs201616184 Chr.4: 74098990-74098990 on Build GRCh38
rs930520676 Chr.4: 74099004-74099004 on Build GRCh38
rs752933462 Chr.4: 74098995-74098995 on Build GRCh38
rs774070216 Chr.4: 74098974-74098974 on Build GRCh38
rs757065479 Chr.4: 74098835-74098835 on Build GRCh38
rs752061063 Chr.4: 74098950-74098950 on Build GRCh38
rs200149133 Chr.4: 74098840-74098840 on Build GRCh38
rs201938440 Chr.4: 74098936-74098936 on Build GRCh38
rs1012475166 Chr.4: 74099080-74099080 on Build GRCh38
rs202144217 Chr.4: 74098886-74098886 on Build GRCh38
rs200606181 Chr.4: 74099112-74099112 on Build GRCh38
rs535488364 Chr.4: 74098759-74098759 on Build GRCh38
rs767415288 Chr.4: 74098923-74098923 on Build GRCh38
rs763155248 Chr.4: 74099002-74099002 on Build GRCh38
rs773261820 Chr.4: 74099107-74099107 on Build GRCh38
rs542273464 Chr.4: 74099008-74099008 on Build GRCh38
rs772692101 Chr.4: 74098793-74098793 on Build GRCh38
rs758430569 Chr.4: 74098757-74098757 on Build GRCh38
rs201218927 Chr.4: 74098879-74098879 on Build GRCh38
rs199521882 Chr.4: 74099114-74099114 on Build GRCh38
rs962514830 Chr.4: 74098942-74098942 on Build GRCh38
rs758008188 Chr.4: 74098956-74098956 on Build GRCh38

View Full Product Details