Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75960896-75961413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDAD1
Entrez Gene ID
55153
Gene Name
SDA1 domain containing 1
Gene Aliases
-
UniGene
Hs.632604
Gene Chromosome Location
Chr.4: 75949906-75990962 on Build GRCh38
Transcripts
NM_001288983.1 NM_001288984.1 NM_018115.3 XM_005263102.4 XM_005263105.4 XM_017008354.1 XM_017008355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7095212 c.1242C>G p.L414L Chr.4: 75961248-75961248 on Build GRCh38
7111149 c.1185C>G p.I395M Chr.4: 75961305-75961305 on Build GRCh38
6100957 c.1323A>G p.T441T Chr.4: 75961061-75961061 on Build GRCh38
6168046 c.1188_1189TG>AT p.N396_A397>KS Chr.4: 75961301-75961302 on Build GRCh38
390226 c.1333C>T p.Q445* Chr.4: 75961051-75961051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013554798 Chr.4: 75961149-75961149 on Build GRCh38
rs764734200 Chr.4: 75961276-75961276 on Build GRCh38
rs902036806 Chr.4: 75961151-75961151 on Build GRCh38
rs201266930 Chr.4: 75961145-75961145 on Build GRCh38
rs369181093 Chr.4: 75961256-75961256 on Build GRCh38
rs780169264 Chr.4: 75961082-75961082 on Build GRCh38
rs568147107 Chr.4: 75961362-75961362 on Build GRCh38
rs567329224 Chr.4: 75961081-75961081 on Build GRCh38
rs772526307 Chr.4: 75961098-75961098 on Build GRCh38
rs749385725 Chr.4: 75961084-75961084 on Build GRCh38
rs747400724 Chr.4: 75961329-75961329 on Build GRCh38
rs774899515 Chr.4: 75961095-75961095 on Build GRCh38
rs758191084 Chr.4: 75961174-75961174 on Build GRCh38
rs762107992 Chr.4: 75961097-75961097 on Build GRCh38
rs754051760 Chr.4: 75961298-75961298 on Build GRCh38
rs1025439377 Chr.4: 75961160-75961160 on Build GRCh38
rs377507179 Chr.4: 75961203-75961203 on Build GRCh38
rs535487001 Chr.4: 75961375-75961375 on Build GRCh38
rs371503784 Chr.4: 75961059-75961059 on Build GRCh38
rs201154587 Chr.4: 75961341-75961341 on Build GRCh38
rs902917530 Chr.4: 75961016-75961016 on Build GRCh38
rs889373137 Chr.4: 75961090-75961090 on Build GRCh38
rs775227985 Chr.4: 75961190-75961191 on Build GRCh38
rs369140120 Chr.4: 75961303-75961303 on Build GRCh38
rs772691754 Chr.4: 75961248-75961248 on Build GRCh38
rs759063974 Chr.4: 75961142-75961142 on Build GRCh38
rs775978530 Chr.4: 75960996-75960996 on Build GRCh38
rs763313000 Chr.4: 75961002-75961002 on Build GRCh38
rs994060624 Chr.4: 75961166-75961166 on Build GRCh38
rs776293957 Chr.4: 75961265-75961265 on Build GRCh38
rs760295028 Chr.4: 75961018-75961018 on Build GRCh38
rs372045634 Chr.4: 75961259-75961259 on Build GRCh38
rs773005147 Chr.4: 75961099-75961099 on Build GRCh38
rs757028091 Chr.4: 75961290-75961290 on Build GRCh38
rs756398904 Chr.4: 75961183-75961183 on Build GRCh38
rs765927426 Chr.4: 75961027-75961027 on Build GRCh38
rs760688855 Chr.4: 75961321-75961324 on Build GRCh38
rs552023073 Chr.4: 75961284-75961284 on Build GRCh38
rs758159904 Chr.4: 75961061-75961061 on Build GRCh38
rs986324711 Chr.4: 75961380-75961380 on Build GRCh38
rs879569986 Chr.4: 75960953-75960954 on Build GRCh38
rs745966356 Chr.4: 75960978-75960978 on Build GRCh38
rs922980239 Chr.4: 75961372-75961372 on Build GRCh38
rs532713299 Chr.4: 75960928-75960928 on Build GRCh38
rs139820906 Chr.4: 75961047-75961047 on Build GRCh38
rs780221956 Chr.4: 75961191-75961191 on Build GRCh38
rs748623734 Chr.4: 75961244-75961244 on Build GRCh38
rs753383821 Chr.4: 75961028-75961028 on Build GRCh38
rs751344802 Chr.4: 75961180-75961180 on Build GRCh38
rs749381831 Chr.4: 75961206-75961206 on Build GRCh38
rs959356889 Chr.4: 75961299-75961299 on Build GRCh38
rs548168022 Chr.4: 75961065-75961065 on Build GRCh38
rs774648563 Chr.4: 75961004-75961004 on Build GRCh38
rs201627257 Chr.4: 75961029-75961029 on Build GRCh38
rs770755760 Chr.4: 75961263-75961263 on Build GRCh38
rs375343526 Chr.4: 75961271-75961271 on Build GRCh38
rs776149205 Chr.4: 75961342-75961342 on Build GRCh38
rs555986297 Chr.4: 75961353-75961353 on Build GRCh38
rs143777133 Chr.4: 75961221-75961221 on Build GRCh38
rs937222289 Chr.4: 75960961-75960961 on Build GRCh38
rs757496622 Chr.4: 75960938-75960938 on Build GRCh38
rs766124253 Chr.4: 75961113-75961113 on Build GRCh38
rs1007848898 Chr.4: 75961122-75961122 on Build GRCh38
rs753327616 Chr.4: 75961141-75961141 on Build GRCh38
rs760460377 Chr.4: 75961105-75961105 on Build GRCh38
rs769315728 Chr.4: 75961348-75961348 on Build GRCh38
rs35978727 Chr.4: 75961072-75961072 on Build GRCh38
rs114496003 Chr.4: 75960948-75960948 on Build GRCh38
rs775134667 Chr.4: 75961351-75961351 on Build GRCh38
rs745708160 Chr.4: 75961009-75961010 on Build GRCh38
rs373422780 Chr.4: 75961339-75961339 on Build GRCh38
rs766532305 Chr.4: 75961296-75961296 on Build GRCh38
rs775300867 Chr.4: 75961279-75961279 on Build GRCh38
rs779226995 Chr.4: 75961007-75961007 on Build GRCh38
rs202155622 Chr.4: 75961285-75961285 on Build GRCh38
rs776210113 Chr.4: 75961351-75961352 on Build GRCh38
rs371779378 Chr.4: 75961066-75961066 on Build GRCh38
rs778878588 Chr.4: 75961030-75961030 on Build GRCh38
rs769088572 Chr.4: 75961094-75961094 on Build GRCh38
rs145396500 Chr.4: 75961009-75961009 on Build GRCh38
rs975777620 Chr.4: 75961370-75961370 on Build GRCh38
rs755018096 Chr.4: 75961302-75961302 on Build GRCh38
rs762938714 Chr.4: 75961280-75961280 on Build GRCh38
rs774540994 Chr.4: 75960992-75960994 on Build GRCh38
rs12509272 Chr.4: 75960921-75960921 on Build GRCh38
rs771247603 Chr.4: 75960978-75960979 on Build GRCh38
rs147216033 Chr.4: 75961222-75961222 on Build GRCh38
rs763884510 Chr.4: 75961336-75961336 on Build GRCh38
rs778402536 Chr.4: 75961325-75961325 on Build GRCh38
rs769196420 Chr.4: 75961003-75961003 on Build GRCh38
rs371056748 Chr.4: 75961324-75961324 on Build GRCh38
rs374533146 Chr.4: 75961014-75961014 on Build GRCh38
rs752636974 Chr.4: 75961173-75961173 on Build GRCh38
rs113244163 Chr.4: 75961272-75961272 on Build GRCh38
rs769879168 Chr.4: 75961075-75961075 on Build GRCh38
rs879386220 Chr.4: 75960948-75960952 on Build GRCh38
rs771826996 Chr.4: 75961012-75961014 on Build GRCh38
rs777484376 Chr.4: 75961176-75961176 on Build GRCh38

View Full Product Details