Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176599341-176599841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRIN1
Entrez Gene ID
114787
Gene Name
G protein regulated inducer of neurite outgrowth 1
Gene Aliases
GRIN1
UniGene
Hs.150549
Gene Chromosome Location
Chr.5: 176595802-176610130 on Build GRCh38
Transcripts
NM_052899.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5868201 c.169C>T p.P57S Chr.5: 176599666-176599666 on Build GRCh38
1066079 c.67C>T p.R23* Chr.5: 176599768-176599768 on Build GRCh38
6877065 c.55C>T p.P19S Chr.5: 176599780-176599780 on Build GRCh38
1066078 c.159C>T p.S53S Chr.5: 176599676-176599676 on Build GRCh38
4500614 c.56C>G p.P19R Chr.5: 176599779-176599779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775160273 Chr.5: 176599461-176599461 on Build GRCh38
rs773762496 Chr.5: 176599538-176599538 on Build GRCh38
rs35847475 Chr.5: 176599490-176599490 on Build GRCh38
rs563554836 Chr.5: 176599432-176599432 on Build GRCh38
rs745720208 Chr.5: 176599423-176599423 on Build GRCh38
rs566009007 Chr.5: 176599601-176599601 on Build GRCh38
rs369705495 Chr.5: 176599392-176599392 on Build GRCh38
rs773941381 Chr.5: 176599427-176599427 on Build GRCh38
rs774967158 Chr.5: 176599648-176599648 on Build GRCh38
rs369617306 Chr.5: 176599533-176599533 on Build GRCh38
rs964331552 Chr.5: 176599609-176599609 on Build GRCh38
rs781403775 Chr.5: 176599467-176599467 on Build GRCh38
rs771797551 Chr.5: 176599643-176599643 on Build GRCh38
rs984896441 Chr.5: 176599575-176599575 on Build GRCh38
rs764270167 Chr.5: 176599776-176599776 on Build GRCh38
rs749219015 Chr.5: 176599674-176599674 on Build GRCh38
rs1002993014 Chr.5: 176599472-176599472 on Build GRCh38
rs1006567497 Chr.5: 176599592-176599592 on Build GRCh38
rs747118885 Chr.5: 176599368-176599368 on Build GRCh38
rs373735554 Chr.5: 176599655-176599655 on Build GRCh38
rs375003836 Chr.5: 176599441-176599441 on Build GRCh38
rs779759901 Chr.5: 176599716-176599716 on Build GRCh38
rs746196366 Chr.5: 176599631-176599631 on Build GRCh38
rs747625918 Chr.5: 176599752-176599752 on Build GRCh38
rs778209257 Chr.5: 176599507-176599507 on Build GRCh38
rs752798153 Chr.5: 176599676-176599676 on Build GRCh38
rs753537681 Chr.5: 176599375-176599375 on Build GRCh38
rs757802572 Chr.5: 176599695-176599695 on Build GRCh38
rs764124095 Chr.5: 176599566-176599566 on Build GRCh38
rs371681993 Chr.5: 176599372-176599372 on Build GRCh38
rs768225305 Chr.5: 176599652-176599652 on Build GRCh38
rs530875829 Chr.5: 176599433-176599433 on Build GRCh38
rs775115831 Chr.5: 176599556-176599556 on Build GRCh38
rs779307241 Chr.5: 176599629-176599629 on Build GRCh38
rs17854765 Chr.5: 176599474-176599474 on Build GRCh38
rs773670993 Chr.5: 176599403-176599403 on Build GRCh38
rs764218735 Chr.5: 176599685-176599685 on Build GRCh38
rs146009772 Chr.5: 176599817-176599817 on Build GRCh38
rs149491456 Chr.5: 176599393-176599393 on Build GRCh38
rs755020095 Chr.5: 176599462-176599462 on Build GRCh38
rs760874794 Chr.5: 176599775-176599775 on Build GRCh38
rs558770081 Chr.5: 176599814-176599814 on Build GRCh38
rs141810829 Chr.5: 176599521-176599521 on Build GRCh38
rs762814613 Chr.5: 176599760-176599760 on Build GRCh38
rs868494121 Chr.5: 176599492-176599492 on Build GRCh38
rs753726320 Chr.5: 176599451-176599451 on Build GRCh38
rs139277418 Chr.5: 176599664-176599664 on Build GRCh38
rs763269493 Chr.5: 176599418-176599418 on Build GRCh38
rs952075385 Chr.5: 176599568-176599568 on Build GRCh38
rs753911157 Chr.5: 176599692-176599692 on Build GRCh38
rs369279565 Chr.5: 176599460-176599460 on Build GRCh38
rs200984378 Chr.5: 176599793-176599793 on Build GRCh38
rs201254907 Chr.5: 176599576-176599576 on Build GRCh38
rs114390685 Chr.5: 176599437-176599437 on Build GRCh38
rs780338746 Chr.5: 176599812-176599812 on Build GRCh38
rs768407134 Chr.5: 176599741-176599741 on Build GRCh38
rs913452373 Chr.5: 176599391-176599391 on Build GRCh38
rs752522649 Chr.5: 176599776-176599776 on Build GRCh38
rs767600020 Chr.5: 176599590-176599590 on Build GRCh38
rs776427796 Chr.5: 176599377-176599377 on Build GRCh38
rs750821485 Chr.5: 176599714-176599714 on Build GRCh38
rs201692802 Chr.5: 176599715-176599715 on Build GRCh38
rs774184931 Chr.5: 176599768-176599768 on Build GRCh38
rs755290532 Chr.5: 176599587-176599587 on Build GRCh38
rs142576280 Chr.5: 176599666-176599666 on Build GRCh38
rs199789374 Chr.5: 176599697-176599697 on Build GRCh38
rs374324333 Chr.5: 176599786-176599786 on Build GRCh38
rs771610826 Chr.5: 176599547-176599547 on Build GRCh38
rs750799201 Chr.5: 176599767-176599767 on Build GRCh38
rs749726553 Chr.5: 176599611-176599611 on Build GRCh38
rs773995820 Chr.5: 176599535-176599535 on Build GRCh38
rs756234375 Chr.5: 176599682-176599682 on Build GRCh38
rs747696664 Chr.5: 176599815-176599815 on Build GRCh38
rs61738845 Chr.5: 176599383-176599383 on Build GRCh38
rs765009644 Chr.5: 176599664-176599664 on Build GRCh38
rs146274822 Chr.5: 176599579-176599579 on Build GRCh38
rs765257738 Chr.5: 176599665-176599665 on Build GRCh38
rs757755833 Chr.5: 176599627-176599627 on Build GRCh38
rs913411055 Chr.5: 176599689-176599689 on Build GRCh38
rs910561160 Chr.5: 176599362-176599362 on Build GRCh38
rs923509213 Chr.5: 176599639-176599639 on Build GRCh38
rs780912831 Chr.5: 176599742-176599742 on Build GRCh38
rs561026169 Chr.5: 176599459-176599459 on Build GRCh38
rs758801044 Chr.5: 176599808-176599808 on Build GRCh38
rs746553159 Chr.5: 176599649-176599649 on Build GRCh38
rs536587245 Chr.5: 176599632-176599632 on Build GRCh38
rs761716186 Chr.5: 176599577-176599577 on Build GRCh38
rs754629710 Chr.5: 176599423-176599424 on Build GRCh38
rs531996464 Chr.5: 176599541-176599541 on Build GRCh38
rs1048027500 Chr.5: 176599494-176599494 on Build GRCh38
rs749673252 Chr.5: 176599514-176599514 on Build GRCh38
rs748625659 Chr.5: 176599386-176599386 on Build GRCh38
rs746582726 Chr.5: 176599726-176599726 on Build GRCh38
rs756095865 Chr.5: 176599595-176599595 on Build GRCh38
rs778264164 Chr.5: 176599597-176599597 on Build GRCh38
rs769879609 Chr.5: 176599753-176599753 on Build GRCh38
rs370708910 Chr.5: 176599754-176599754 on Build GRCh38

View Full Product Details