Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176868013-176868505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC5A
Entrez Gene ID
90249
Gene Name
unc-5 netrin receptor A
Gene Aliases
UNC5H1
UniGene
Hs.33191
Gene Chromosome Location
Chr.5: 176810559-176880898 on Build GRCh38
Transcripts
NM_133369.2 XM_006714927.1 XM_006714928.1 XM_011534686.1 XM_011534687.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6170561 c.258C>A p.C86* Chr.5: 176868215-176868215 on Build GRCh38
4347055 c.193C>T p.R65C Chr.5: 176868150-176868150 on Build GRCh38
4739329 c.355C>A p.L119M Chr.5: 176868192-176868192 on Build GRCh38
3340608 c.313C>T p.R105C Chr.5: 176868150-176868150 on Build GRCh38
3340607 c.312C>T p.V104V Chr.5: 176868149-176868149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002523453 Chr.5: 176868396-176868396 on Build GRCh38
rs757793939 Chr.5: 176868263-176868263 on Build GRCh38
rs772731294 Chr.5: 176868271-176868271 on Build GRCh38
rs780569422 Chr.5: 176868216-176868216 on Build GRCh38
rs375197591 Chr.5: 176868092-176868092 on Build GRCh38
rs200845386 Chr.5: 176868058-176868058 on Build GRCh38
rs776211040 Chr.5: 176868232-176868232 on Build GRCh38
rs367946633 Chr.5: 176868322-176868322 on Build GRCh38
rs761286091 Chr.5: 176868121-176868121 on Build GRCh38
rs746382338 Chr.5: 176868206-176868206 on Build GRCh38
rs183051657 Chr.5: 176868363-176868363 on Build GRCh38
rs375118336 Chr.5: 176868312-176868312 on Build GRCh38
rs1028209618 Chr.5: 176868377-176868377 on Build GRCh38
rs267600562 Chr.5: 176868197-176868197 on Build GRCh38
rs577653637 Chr.5: 176868419-176868419 on Build GRCh38
rs1022183469 Chr.5: 176868109-176868109 on Build GRCh38
rs747315272 Chr.5: 176868227-176868227 on Build GRCh38
rs777914371 Chr.5: 176868185-176868185 on Build GRCh38
rs769124232 Chr.5: 176868228-176868228 on Build GRCh38
rs577727748 Chr.5: 176868475-176868475 on Build GRCh38
rs145214251 Chr.5: 176868176-176868176 on Build GRCh38
rs1009375752 Chr.5: 176868358-176868358 on Build GRCh38
rs769240002 Chr.5: 176868265-176868265 on Build GRCh38
rs371476595 Chr.5: 176868303-176868303 on Build GRCh38
rs267600563 Chr.5: 176868198-176868198 on Build GRCh38
rs960656078 Chr.5: 176868481-176868481 on Build GRCh38
rs549600836 Chr.5: 176868083-176868083 on Build GRCh38
rs377325867 Chr.5: 176868308-176868308 on Build GRCh38
rs776291145 Chr.5: 176868105-176868105 on Build GRCh38
rs754080042 Chr.5: 176868190-176868190 on Build GRCh38
rs73341814 Chr.5: 176868311-176868311 on Build GRCh38
rs997993941 Chr.5: 176868042-176868042 on Build GRCh38
rs999643930 Chr.5: 176868061-176868061 on Build GRCh38
rs1029482697 Chr.5: 176868043-176868043 on Build GRCh38
rs544721124 Chr.5: 176868474-176868474 on Build GRCh38
rs377070410 Chr.5: 176868177-176868177 on Build GRCh38
rs571689500 Chr.5: 176868192-176868192 on Build GRCh38
rs985827475 Chr.5: 176868272-176868272 on Build GRCh38
rs762929019 Chr.5: 176868158-176868158 on Build GRCh38
rs772667180 Chr.5: 176868215-176868215 on Build GRCh38
rs980489104 Chr.5: 176868142-176868142 on Build GRCh38
rs867714470 Chr.5: 176868418-176868418 on Build GRCh38
rs754054378 Chr.5: 176868306-176868306 on Build GRCh38
rs906543236 Chr.5: 176868340-176868340 on Build GRCh38
rs1033391010 Chr.5: 176868397-176868397 on Build GRCh38
rs750449170 Chr.5: 176868309-176868309 on Build GRCh38
rs573470131 Chr.5: 176868359-176868359 on Build GRCh38
rs760434121 Chr.5: 176868088-176868088 on Build GRCh38
rs891853957 Chr.5: 176868349-176868349 on Build GRCh38
rs201652528 Chr.5: 176868122-176868122 on Build GRCh38
rs761492284 Chr.5: 176868233-176868233 on Build GRCh38
rs192826758 Chr.5: 176868328-176868328 on Build GRCh38
rs762914199 Chr.5: 176868300-176868300 on Build GRCh38
rs113171620 Chr.5: 176868305-176868305 on Build GRCh38
rs368865182 Chr.5: 176868324-176868324 on Build GRCh38
rs538689245 Chr.5: 176868304-176868304 on Build GRCh38
rs555155450 Chr.5: 176868333-176868333 on Build GRCh38
rs868676462 Chr.5: 176868313-176868313 on Build GRCh38
rs750371659 Chr.5: 176868150-176868150 on Build GRCh38
rs770897391 Chr.5: 176868094-176868094 on Build GRCh38
rs867627987 Chr.5: 176868219-176868219 on Build GRCh38
rs373576664 Chr.5: 176868162-176868162 on Build GRCh38
rs892296161 Chr.5: 176868062-176868062 on Build GRCh38
rs953164401 Chr.5: 176868425-176868425 on Build GRCh38
rs879073769 Chr.5: 176868261-176868261 on Build GRCh38

View Full Product Details