Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176877349-176877842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC5A
Entrez Gene ID
90249
Gene Name
unc-5 netrin receptor A
Gene Aliases
UNC5H1
UniGene
Hs.33191
Gene Chromosome Location
Chr.5: 176810559-176880898 on Build GRCh38
Transcripts
NM_133369.2 XM_006714927.1 XM_006714928.1 XM_011534686.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6220829 c.1507G>A p.V503I Chr.5: 176877575-176877575 on Build GRCh38
4739338 c.1497G>T p.S499S Chr.5: 176877685-176877685 on Build GRCh38
5868205 c.1628G>A p.S543N Chr.5: 176877696-176877696 on Build GRCh38
5815648 c.1557T>A p.A519A Chr.5: 176877625-176877625 on Build GRCh38
3919728 c.1593G>A p.W531* Chr.5: 176877661-176877661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2304517 Chr.5: 176877398-176877398 on Build GRCh38
rs757748040 Chr.5: 176877722-176877722 on Build GRCh38
rs987694465 Chr.5: 176877397-176877397 on Build GRCh38
rs1055843526 Chr.5: 176877763-176877763 on Build GRCh38
rs887180297 Chr.5: 176877780-176877780 on Build GRCh38
rs145966608 Chr.5: 176877792-176877792 on Build GRCh38
rs771734603 Chr.5: 176877636-176877636 on Build GRCh38
rs749730825 Chr.5: 176877608-176877608 on Build GRCh38
rs770930810 Chr.5: 176877485-176877490 on Build GRCh38
rs757028328 Chr.5: 176877558-176877558 on Build GRCh38
rs773587251 Chr.5: 176877494-176877494 on Build GRCh38
rs369884419 Chr.5: 176877726-176877726 on Build GRCh38
rs558742323 Chr.5: 176877788-176877788 on Build GRCh38
rs749784301 Chr.5: 176877720-176877720 on Build GRCh38
rs546781214 Chr.5: 176877384-176877384 on Build GRCh38
rs748632995 Chr.5: 176877601-176877601 on Build GRCh38
rs71601324 Chr.5: 176877786-176877786 on Build GRCh38
rs926942908 Chr.5: 176877446-176877446 on Build GRCh38
rs766650524 Chr.5: 176877560-176877560 on Build GRCh38
rs143892682 Chr.5: 176877605-176877605 on Build GRCh38
rs368534307 Chr.5: 176877425-176877425 on Build GRCh38
rs866656111 Chr.5: 176877376-176877376 on Build GRCh38
rs752628468 Chr.5: 176877531-176877531 on Build GRCh38
rs774190473 Chr.5: 176877610-176877610 on Build GRCh38
rs147274124 Chr.5: 176877628-176877628 on Build GRCh38
rs779375748 Chr.5: 176877723-176877723 on Build GRCh38
rs979775740 Chr.5: 176877433-176877433 on Build GRCh38
rs774995388 Chr.5: 176877643-176877643 on Build GRCh38
rs774987864 Chr.5: 176877739-176877739 on Build GRCh38
rs752764039 Chr.5: 176877690-176877690 on Build GRCh38
rs936230640 Chr.5: 176877759-176877759 on Build GRCh38
rs1046250780 Chr.5: 176877775-176877775 on Build GRCh38
rs767108833 Chr.5: 176877527-176877527 on Build GRCh38
rs1010371411 Chr.5: 176877632-176877632 on Build GRCh38
rs1036144782 Chr.5: 176877793-176877793 on Build GRCh38
rs926170860 Chr.5: 176877557-176877557 on Build GRCh38
rs1031021101 Chr.5: 176877626-176877626 on Build GRCh38
rs373702466 Chr.5: 176877595-176877595 on Build GRCh38
rs760538769 Chr.5: 176877644-176877644 on Build GRCh38
rs953123296 Chr.5: 176877396-176877396 on Build GRCh38
rs938639377 Chr.5: 176877472-176877472 on Build GRCh38
rs751749790 Chr.5: 176877574-176877574 on Build GRCh38
rs764148327 Chr.5: 176877658-176877658 on Build GRCh38
rs567157966 Chr.5: 176877757-176877757 on Build GRCh38
rs748664592 Chr.5: 176877487-176877487 on Build GRCh38
rs550723381 Chr.5: 176877506-176877506 on Build GRCh38
rs755099198 Chr.5: 176877575-176877575 on Build GRCh38
rs770075919 Chr.5: 176877489-176877489 on Build GRCh38
rs915916990 Chr.5: 176877770-176877770 on Build GRCh38
rs909710008 Chr.5: 176877769-176877769 on Build GRCh38
rs776424930 Chr.5: 176877668-176877668 on Build GRCh38
rs532264598 Chr.5: 176877431-176877431 on Build GRCh38
rs796380681 Chr.5: 176877464-176877465 on Build GRCh38
rs114202839 Chr.5: 176877755-176877755 on Build GRCh38
rs768017637 Chr.5: 176877744-176877744 on Build GRCh38
rs771421798 Chr.5: 176877609-176877609 on Build GRCh38
rs367736981 Chr.5: 176877802-176877802 on Build GRCh38
rs368092339 Chr.5: 176877684-176877684 on Build GRCh38
rs767500072 Chr.5: 176877689-176877689 on Build GRCh38
rs371948820 Chr.5: 176877685-176877685 on Build GRCh38
rs776501009 Chr.5: 176877742-176877742 on Build GRCh38
rs755462801 Chr.5: 176877486-176877486 on Build GRCh38
rs774135248 Chr.5: 176877514-176877514 on Build GRCh38
rs377126119 Chr.5: 176877712-176877712 on Build GRCh38
rs374314240 Chr.5: 176877740-176877740 on Build GRCh38
rs921712326 Chr.5: 176877385-176877385 on Build GRCh38
rs933331562 Chr.5: 176877785-176877785 on Build GRCh38
rs899126255 Chr.5: 176877800-176877800 on Build GRCh38
rs377728723 Chr.5: 176877754-176877754 on Build GRCh38
rs771693438 Chr.5: 176877735-176877735 on Build GRCh38
rs760615400 Chr.5: 176877543-176877543 on Build GRCh38
rs138151702 Chr.5: 176877548-176877548 on Build GRCh38
rs73804275 Chr.5: 176877817-176877817 on Build GRCh38
rs758650462 Chr.5: 176877747-176877747 on Build GRCh38
rs769375615 Chr.5: 176877540-176877540 on Build GRCh38
rs762518988 Chr.5: 176877502-176877502 on Build GRCh38
rs756136862 Chr.5: 176877691-176877691 on Build GRCh38
rs755152279 Chr.5: 176877688-176877688 on Build GRCh38
rs1004694375 Chr.5: 176877630-176877630 on Build GRCh38
rs115634193 Chr.5: 176877669-176877669 on Build GRCh38
rs528325915 Chr.5: 176877377-176877377 on Build GRCh38
rs759187611 Chr.5: 176877521-176877521 on Build GRCh38
rs116480303 Chr.5: 176877774-176877774 on Build GRCh38
rs781282336 Chr.5: 176877596-176877596 on Build GRCh38

View Full Product Details