Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106520654-106521165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM1
Entrez Gene ID
202
Gene Name
absent in melanoma 1
Gene Aliases
CRYBG1, ST4
UniGene
Hs.724262
Gene Chromosome Location
Chr.6: 106360808-106570449 on Build GRCh38
Transcripts
NM_001624.3 XM_005266839.3 XM_017010333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
450273 c.2239A>T p.K747* Chr.6: 106520671-106520671 on Build GRCh38
739786 c.2590A>G p.T864A Chr.6: 106521022-106521022 on Build GRCh38
1195098 c.2350G>A p.A784T Chr.6: 106520782-106520782 on Build GRCh38
1071770 c.2688G>A p.L896L Chr.6: 106521120-106521120 on Build GRCh38
6547537 c.2692G>T p.D898Y Chr.6: 106521124-106521124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755546436 Chr.6: 106520687-106520687 on Build GRCh38
rs752201884 Chr.6: 106520702-106520702 on Build GRCh38
rs200461385 Chr.6: 106521123-106521123 on Build GRCh38
rs746936367 Chr.6: 106520815-106520815 on Build GRCh38
rs780076876 Chr.6: 106521099-106521099 on Build GRCh38
rs151094069 Chr.6: 106521006-106521006 on Build GRCh38
rs369857107 Chr.6: 106521012-106521012 on Build GRCh38
rs777224294 Chr.6: 106521042-106521042 on Build GRCh38
rs754064957 Chr.6: 106520735-106520735 on Build GRCh38
rs763220526 Chr.6: 106520757-106520757 on Build GRCh38
rs768892501 Chr.6: 106521115-106521115 on Build GRCh38
rs372150269 Chr.6: 106520916-106520916 on Build GRCh38
rs759828725 Chr.6: 106520859-106520859 on Build GRCh38
rs780650499 Chr.6: 106520700-106520700 on Build GRCh38
rs751257287 Chr.6: 106521080-106521080 on Build GRCh38
rs574757027 Chr.6: 106520779-106520779 on Build GRCh38
rs374238219 Chr.6: 106521128-106521128 on Build GRCh38
rs774587945 Chr.6: 106520858-106520858 on Build GRCh38
rs779978674 Chr.6: 106520814-106520814 on Build GRCh38
rs143246482 Chr.6: 106520725-106520725 on Build GRCh38
rs767971279 Chr.6: 106520778-106520778 on Build GRCh38
rs764855382 Chr.6: 106520955-106520955 on Build GRCh38
rs775754813 Chr.6: 106520925-106520925 on Build GRCh38
rs148858033 Chr.6: 106520845-106520845 on Build GRCh38
rs778938706 Chr.6: 106521086-106521086 on Build GRCh38
rs755609344 Chr.6: 106520719-106520719 on Build GRCh38
rs766167838 Chr.6: 106520971-106520971 on Build GRCh38
rs762581577 Chr.6: 106520967-106520967 on Build GRCh38
rs775315490 Chr.6: 106520729-106520729 on Build GRCh38
rs747292072 Chr.6: 106521034-106521034 on Build GRCh38
rs773427826 Chr.6: 106520754-106520754 on Build GRCh38
rs758212291 Chr.6: 106520883-106520883 on Build GRCh38
rs1050137826 Chr.6: 106520709-106520709 on Build GRCh38
rs772228590 Chr.6: 106521028-106521028 on Build GRCh38
rs376221228 Chr.6: 106521011-106521011 on Build GRCh38
rs370122396 Chr.6: 106521122-106521122 on Build GRCh38
rs368874479 Chr.6: 106520887-106520887 on Build GRCh38
rs773232706 Chr.6: 106520843-106520843 on Build GRCh38
rs776938579 Chr.6: 106520920-106520921 on Build GRCh38
rs747271353 Chr.6: 106521110-106521110 on Build GRCh38
rs771003350 Chr.6: 106520905-106520905 on Build GRCh38
rs777415653 Chr.6: 106520721-106520721 on Build GRCh38
rs142612305 Chr.6: 106521010-106521010 on Build GRCh38
rs767784436 Chr.6: 106520866-106520866 on Build GRCh38
rs770475403 Chr.6: 106521124-106521124 on Build GRCh38
rs1008237006 Chr.6: 106520946-106520946 on Build GRCh38
rs61739383 Chr.6: 106520796-106520796 on Build GRCh38
rs200098719 Chr.6: 106520727-106520727 on Build GRCh38
rs756845318 Chr.6: 106520875-106520875 on Build GRCh38
rs373317464 Chr.6: 106521077-106521077 on Build GRCh38
rs777312523 Chr.6: 106520945-106520945 on Build GRCh38
rs779772953 Chr.6: 106520884-106520884 on Build GRCh38
rs201716413 Chr.6: 106520909-106520909 on Build GRCh38
rs772891918 Chr.6: 106521061-106521061 on Build GRCh38
rs745764944 Chr.6: 106520726-106520726 on Build GRCh38
rs765412508 Chr.6: 106521132-106521133 on Build GRCh38
rs761149534 Chr.6: 106520868-106520868 on Build GRCh38
rs761344006 Chr.6: 106520949-106520949 on Build GRCh38
rs139814213 Chr.6: 106520783-106520783 on Build GRCh38
rs1048919143 Chr.6: 106520819-106520819 on Build GRCh38
rs764455511 Chr.6: 106520930-106520930 on Build GRCh38
rs761218414 Chr.6: 106520928-106520928 on Build GRCh38
rs750088904 Chr.6: 106520962-106520962 on Build GRCh38
rs372732472 Chr.6: 106520869-106520869 on Build GRCh38
rs770833441 Chr.6: 106520724-106520724 on Build GRCh38
rs759774505 Chr.6: 106520950-106520952 on Build GRCh38
rs544194019 Chr.6: 106520913-106520913 on Build GRCh38
rs769906749 Chr.6: 106520831-106520831 on Build GRCh38
rs61739377 Chr.6: 106520777-106520777 on Build GRCh38
rs755591572 Chr.6: 106520780-106520780 on Build GRCh38
rs978721575 Chr.6: 106520965-106520965 on Build GRCh38
rs200028219 Chr.6: 106520975-106520975 on Build GRCh38
rs762472106 Chr.6: 106521059-106521059 on Build GRCh38
rs747613157 Chr.6: 106520980-106520980 on Build GRCh38
rs201230039 Chr.6: 106521029-106521029 on Build GRCh38
rs866622614 Chr.6: 106520834-106520834 on Build GRCh38
rs756083117 Chr.6: 106521085-106521085 on Build GRCh38
rs762330813 Chr.6: 106521136-106521136 on Build GRCh38
rs368637688 Chr.6: 106520800-106520800 on Build GRCh38
rs368573019 Chr.6: 106520773-106520773 on Build GRCh38
rs771191766 Chr.6: 106520847-106520847 on Build GRCh38
rs769770576 Chr.6: 106520737-106520737 on Build GRCh38
rs765900077 Chr.6: 106521069-106521069 on Build GRCh38
rs748904708 Chr.6: 106520723-106520723 on Build GRCh38
rs967006145 Chr.6: 106520952-106520952 on Build GRCh38
rs749318058 Chr.6: 106520903-106520903 on Build GRCh38
rs777887660 Chr.6: 106520890-106520890 on Build GRCh38
rs776050416 Chr.6: 106520867-106520867 on Build GRCh38
rs199971648 Chr.6: 106521084-106521084 on Build GRCh38
rs369153076 Chr.6: 106521001-106521001 on Build GRCh38
rs766727729 Chr.6: 106520763-106520763 on Build GRCh38
rs1417354 Chr.6: 106520799-106520799 on Build GRCh38
rs780341653 Chr.6: 106521033-106521033 on Build GRCh38
rs143755688 Chr.6: 106520886-106520886 on Build GRCh38
rs949074343 Chr.6: 106521082-106521082 on Build GRCh38
rs772477743 Chr.6: 106520917-106520917 on Build GRCh38
rs545257394 Chr.6: 106520882-106520882 on Build GRCh38
rs780923769 Chr.6: 106521002-106521002 on Build GRCh38
rs769426664 Chr.6: 106521060-106521060 on Build GRCh38
rs769073910 Chr.6: 106521038-106521038 on Build GRCh38
rs199754054 Chr.6: 106520872-106520872 on Build GRCh38
rs201285247 Chr.6: 106521138-106521138 on Build GRCh38
rs202235693 Chr.6: 106521083-106521083 on Build GRCh38
rs566268314 Chr.6: 106521097-106521097 on Build GRCh38
rs146905899 Chr.6: 106521068-106521068 on Build GRCh38
rs778824046 Chr.6: 106520797-106520797 on Build GRCh38
rs375857730 Chr.6: 106520880-106520880 on Build GRCh38

View Full Product Details