Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107152861-107153378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDSS2
Entrez Gene ID
57107
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Gene Aliases
C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1
UniGene
Hs.745008
Gene Chromosome Location
Chr.6: 107152557-107459683 on Build GRCh38
Transcripts
NM_020381.3 XM_011535956.2 XM_011535957.2 XM_011535958.2 XM_011535959.2 XM_011535960.2 XM_011535961.2 XM_011535962.2

SNP Detail

SNP ID Chromosome Location
rs897645748 Chr.6: 107153353-107153353 on Build GRCh38
rs944268514 Chr.6: 107153107-107153107 on Build GRCh38
rs73763734 Chr.6: 107153009-107153009 on Build GRCh38
rs555815596 Chr.6: 107153155-107153155 on Build GRCh38
rs541334001 Chr.6: 107153254-107153254 on Build GRCh38
rs886060918 Chr.6: 107152966-107152966 on Build GRCh38
rs572009730 Chr.6: 107153247-107153247 on Build GRCh38
rs977034934 Chr.6: 107153117-107153117 on Build GRCh38
rs576592297 Chr.6: 107153024-107153024 on Build GRCh38
rs116118550 Chr.6: 107153284-107153284 on Build GRCh38
rs782206175 Chr.6: 107153148-107153148 on Build GRCh38
rs935507706 Chr.6: 107153130-107153130 on Build GRCh38
rs748980679 Chr.6: 107152928-107152928 on Build GRCh38
rs1045159 Chr.6: 107152933-107152933 on Build GRCh38
rs11548249 Chr.6: 107153033-107153033 on Build GRCh38
rs148040761 Chr.6: 107153354-107153354 on Build GRCh38
rs555022569 Chr.6: 107153240-107153240 on Build GRCh38
rs1014709929 Chr.6: 107152921-107152921 on Build GRCh38
rs79647284 Chr.6: 107153186-107153186 on Build GRCh38
rs924091173 Chr.6: 107153118-107153118 on Build GRCh38
rs183305059 Chr.6: 107153312-107153312 on Build GRCh38
rs533550071 Chr.6: 107153306-107153306 on Build GRCh38
rs1003409942 Chr.6: 107153305-107153305 on Build GRCh38
rs911549716 Chr.6: 107153102-107153102 on Build GRCh38
rs973114499 Chr.6: 107152999-107152999 on Build GRCh38
rs533806575 Chr.6: 107152948-107152948 on Build GRCh38
rs748596791 Chr.6: 107153299-107153299 on Build GRCh38
rs1054379299 Chr.6: 107153185-107153185 on Build GRCh38
rs782446668 Chr.6: 107153285-107153285 on Build GRCh38
rs190728862 Chr.6: 107153031-107153031 on Build GRCh38
rs782285947 Chr.6: 107153344-107153344 on Build GRCh38
rs541800250 Chr.6: 107153187-107153187 on Build GRCh38

View Full Product Details