Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108994551-108995099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SESN1
Entrez Gene ID
27244
Gene Name
sestrin 1
Gene Aliases
PA26, SEST1
UniGene
Hs.591336
Gene Chromosome Location
Chr.6: 108986437-109094505 on Build GRCh38
Transcripts
NM_001199933.1 NM_001199934.1 NM_014454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6213648 c.811T>A p.F271I Chr.6: 108994594-108994594 on Build GRCh38
6213647 c.988T>A p.F330I Chr.6: 108994594-108994594 on Build GRCh38
1071946 c.983C>T p.S328F Chr.6: 108994599-108994599 on Build GRCh38
1071945 c.987C>A p.F329L Chr.6: 108994595-108994595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032097962 Chr.6: 108994926-108994926 on Build GRCh38
rs776612917 Chr.6: 108994580-108994580 on Build GRCh38
rs753042998 Chr.6: 108994632-108994632 on Build GRCh38
rs11395949 Chr.6: 108994699-108994700 on Build GRCh38
rs545131089 Chr.6: 108995033-108995033 on Build GRCh38
rs554057047 Chr.6: 108994952-108994952 on Build GRCh38
rs958191694 Chr.6: 108994576-108994576 on Build GRCh38
rs535710671 Chr.6: 108994608-108994608 on Build GRCh38
rs918321637 Chr.6: 108994729-108994729 on Build GRCh38
rs756658378 Chr.6: 108994656-108994656 on Build GRCh38
rs777018214 Chr.6: 108994698-108994699 on Build GRCh38
rs12529409 Chr.6: 108994681-108994681 on Build GRCh38
rs1019781898 Chr.6: 108994970-108994970 on Build GRCh38
rs1057160221 Chr.6: 108994828-108994828 on Build GRCh38
rs779740983 Chr.6: 108994864-108994864 on Build GRCh38
rs909510944 Chr.6: 108994955-108994955 on Build GRCh38
rs983880550 Chr.6: 108994935-108994935 on Build GRCh38
rs956434916 Chr.6: 108994763-108994763 on Build GRCh38
rs150652269 Chr.6: 108994879-108994879 on Build GRCh38
rs1009306795 Chr.6: 108994956-108994956 on Build GRCh38
rs921499936 Chr.6: 108994803-108994803 on Build GRCh38
rs537872455 Chr.6: 108994804-108994804 on Build GRCh38
rs191993227 Chr.6: 108994768-108994768 on Build GRCh38
rs550523323 Chr.6: 108994860-108994860 on Build GRCh38
rs987770561 Chr.6: 108994764-108994764 on Build GRCh38
rs558631260 Chr.6: 108994771-108994771 on Build GRCh38
rs896393987 Chr.6: 108994638-108994638 on Build GRCh38
rs909554158 Chr.6: 108994770-108994770 on Build GRCh38
rs751877811 Chr.6: 108994611-108994611 on Build GRCh38
rs1000673511 Chr.6: 108994920-108994920 on Build GRCh38
rs768435847 Chr.6: 108994904-108994904 on Build GRCh38
rs554545515 Chr.6: 108994806-108994806 on Build GRCh38
rs1048563575 Chr.6: 108994819-108994819 on Build GRCh38
rs112563275 Chr.6: 108994606-108994606 on Build GRCh38
rs577113709 Chr.6: 108994984-108994984 on Build GRCh38
rs965409336 Chr.6: 108994790-108994790 on Build GRCh38
rs998005754 Chr.6: 108994866-108994866 on Build GRCh38
rs566218683 Chr.6: 108994755-108994755 on Build GRCh38
rs113277194 Chr.6: 108994753-108994753 on Build GRCh38
rs1056064906 Chr.6: 108994776-108994776 on Build GRCh38
rs905095577 Chr.6: 108994894-108994894 on Build GRCh38
rs9486985 Chr.6: 108994796-108994796 on Build GRCh38
rs1013466257 Chr.6: 108994649-108994649 on Build GRCh38
rs574820380 Chr.6: 108994820-108994820 on Build GRCh38
rs779935597 Chr.6: 108994592-108994595 on Build GRCh38
rs965251998 Chr.6: 108994699-108994705 on Build GRCh38
rs200255805 Chr.6: 108994605-108994605 on Build GRCh38
rs976474815 Chr.6: 108994699-108994709 on Build GRCh38
rs917497051 Chr.6: 108995023-108995023 on Build GRCh38
rs1057129965 Chr.6: 108994581-108994581 on Build GRCh38
rs1000635746 Chr.6: 108994731-108994731 on Build GRCh38
rs375991942 Chr.6: 108994598-108994598 on Build GRCh38
rs368448588 Chr.6: 108994847-108994847 on Build GRCh38
rs755200320 Chr.6: 108994615-108994615 on Build GRCh38
rs1000277167 Chr.6: 108994812-108994812 on Build GRCh38
rs950204702 Chr.6: 108994791-108994791 on Build GRCh38
rs969287890 Chr.6: 108994688-108994688 on Build GRCh38
rs940045681 Chr.6: 108994827-108994827 on Build GRCh38
rs895850567 Chr.6: 108994832-108994832 on Build GRCh38
rs565393467 Chr.6: 108995071-108995071 on Build GRCh38
rs895949184 Chr.6: 108994777-108994777 on Build GRCh38
rs768024850 Chr.6: 108994627-108994627 on Build GRCh38
rs763876525 Chr.6: 108994574-108994574 on Build GRCh38

View Full Product Details