Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109440711-109441198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL6
Entrez Gene ID
285755
Gene Name
peptidylprolyl isomerase like 6
Gene Aliases
PPIase, RSPH12, bA425D10.6, dJ919F19.1
UniGene
Hs.32234
Gene Chromosome Location
Chr.6: 109390215-109441171 on Build GRCh38
Transcripts
NM_001111298.2 NM_001286360.1 NM_001286361.1 NM_173672.4 NR_104429.1 NM_003080.2

Gene Symbol
SMPD2
Entrez Gene ID
6610
Gene Name
sphingomyelin phosphodiesterase 2
Gene Aliases
ISC1, NSMASE, NSMASE1
UniGene
Hs.55235
Gene Chromosome Location
Chr.6: 109440646-109443922 on Build GRCh38
Transcripts
NM_001111298.2 NM_001286360.1 NM_001286361.1 NM_173672.4 NR_104429.1 NM_003080.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6785630 c.2T>C p.M1T Chr.6: 109441123-109441123 on Build GRCh38
4160112 c.8C>T p.P3L Chr.6: 109441129-109441129 on Build GRCh38
7391124 c.12C>G p.N4K Chr.6: 109441133-109441133 on Build GRCh38
5014631 c.38A>G p.N13S Chr.6: 109441159-109441159 on Build GRCh38
4767150 c.28C>T p.R10W Chr.6: 109441149-109441149 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs181183799 Chr.6: 109440763-109440763 on Build GRCh38
rs751177773 Chr.6: 109441132-109441132 on Build GRCh38
rs1041640968 Chr.6: 109440905-109440905 on Build GRCh38
rs985465076 Chr.6: 109441065-109441065 on Build GRCh38
rs913791510 Chr.6: 109441054-109441054 on Build GRCh38
rs779974923 Chr.6: 109441171-109441171 on Build GRCh38
rs150479732 Chr.6: 109441140-109441140 on Build GRCh38
rs1049766814 Chr.6: 109440870-109440870 on Build GRCh38
rs942180857 Chr.6: 109441141-109441141 on Build GRCh38
rs186985241 Chr.6: 109440908-109440908 on Build GRCh38
rs529756695 Chr.6: 109440773-109440773 on Build GRCh38
rs913141498 Chr.6: 109440841-109440841 on Build GRCh38
rs1048691785 Chr.6: 109440926-109440926 on Build GRCh38
rs866461699 Chr.6: 109440965-109440965 on Build GRCh38
rs1026677283 Chr.6: 109440777-109440777 on Build GRCh38
rs889276437 Chr.6: 109440923-109440923 on Build GRCh38
rs1020562071 Chr.6: 109441024-109441024 on Build GRCh38
rs201131563 Chr.6: 109441172-109441172 on Build GRCh38
rs901904382 Chr.6: 109440887-109440887 on Build GRCh38
rs762476552 Chr.6: 109441119-109441119 on Build GRCh38
rs568198378 Chr.6: 109440959-109440959 on Build GRCh38
rs1048197 Chr.6: 109441129-109441129 on Build GRCh38
rs1050581609 Chr.6: 109440909-109440909 on Build GRCh38
rs375876394 Chr.6: 109440737-109440737 on Build GRCh38
rs1048210 Chr.6: 109440889-109440889 on Build GRCh38
rs767344988 Chr.6: 109441143-109441143 on Build GRCh38
rs926515094 Chr.6: 109441011-109441011 on Build GRCh38
rs572510624 Chr.6: 109441155-109441155 on Build GRCh38
rs115963563 Chr.6: 109440864-109440864 on Build GRCh38
rs747355230 Chr.6: 109441092-109441092 on Build GRCh38
rs1048203 Chr.6: 109441033-109441033 on Build GRCh38
rs758452934 Chr.6: 109441166-109441166 on Build GRCh38
rs970982818 Chr.6: 109440829-109440829 on Build GRCh38
rs546635102 Chr.6: 109440793-109440793 on Build GRCh38
rs1024699332 Chr.6: 109440960-109440960 on Build GRCh38
rs111369755 Chr.6: 109440824-109440824 on Build GRCh38
rs767090557 Chr.6: 109441080-109441080 on Build GRCh38
rs865798957 Chr.6: 109441123-109441123 on Build GRCh38
rs960094391 Chr.6: 109441009-109441009 on Build GRCh38
rs936687711 Chr.6: 109440865-109440865 on Build GRCh38
rs768919421 Chr.6: 109441158-109441158 on Build GRCh38
rs752471965 Chr.6: 109441144-109441144 on Build GRCh38
rs961718551 Chr.6: 109440952-109440952 on Build GRCh38
rs752300310 Chr.6: 109441100-109441100 on Build GRCh38
rs945984722 Chr.6: 109440842-109440842 on Build GRCh38
rs553477533 Chr.6: 109441055-109441055 on Build GRCh38
rs970465956 Chr.6: 109440978-109440978 on Build GRCh38
rs756055563 Chr.6: 109441149-109441149 on Build GRCh38
rs150353891 Chr.6: 109440804-109440804 on Build GRCh38
rs1041836725 Chr.6: 109440880-109440880 on Build GRCh38
rs774554964 Chr.6: 109441169-109441170 on Build GRCh38
rs971812074 Chr.6: 109440794-109440794 on Build GRCh38
rs957991503 Chr.6: 109441064-109441064 on Build GRCh38
rs372015625 Chr.6: 109441120-109441120 on Build GRCh38
rs1054107986 Chr.6: 109440857-109440857 on Build GRCh38
rs568794743 Chr.6: 109440940-109440940 on Build GRCh38
rs989424451 Chr.6: 109441045-109441045 on Build GRCh38
rs745963003 Chr.6: 109441164-109441164 on Build GRCh38
rs9827 Chr.6: 109440942-109440942 on Build GRCh38
rs367857542 Chr.6: 109441097-109441097 on Build GRCh38
rs772863478 Chr.6: 109441122-109441122 on Build GRCh38
rs554461501 Chr.6: 109440955-109440955 on Build GRCh38
rs749518374 Chr.6: 109441138-109441138 on Build GRCh38
rs944215738 Chr.6: 109440899-109440899 on Build GRCh38
rs200126871 Chr.6: 109441086-109441086 on Build GRCh38
rs779018459 Chr.6: 109441071-109441071 on Build GRCh38
rs980920021 Chr.6: 109441006-109441006 on Build GRCh38
rs374122924 Chr.6: 109441077-109441077 on Build GRCh38
rs1019823291 Chr.6: 109440735-109440735 on Build GRCh38
rs889870540 Chr.6: 109440876-109440876 on Build GRCh38
rs539166395 Chr.6: 109441062-109441062 on Build GRCh38
rs1014076489 Chr.6: 109441023-109441023 on Build GRCh38
rs750859422 Chr.6: 109440892-109440892 on Build GRCh38
rs936634172 Chr.6: 109440856-109440856 on Build GRCh38
rs952420207 Chr.6: 109440787-109440787 on Build GRCh38
rs745886037 Chr.6: 109441074-109441074 on Build GRCh38
rs926537171 Chr.6: 109440837-109440837 on Build GRCh38
rs946049239 Chr.6: 109441057-109441057 on Build GRCh38
rs768987055 Chr.6: 109441173-109441173 on Build GRCh38
rs980608634 Chr.6: 109440833-109440833 on Build GRCh38
rs914597017 Chr.6: 109440859-109440859 on Build GRCh38
rs978619621 Chr.6: 109441053-109441053 on Build GRCh38
rs762460465 Chr.6: 109441124-109441124 on Build GRCh38
rs777103986 Chr.6: 109441102-109441102 on Build GRCh38
rs376341583 Chr.6: 109441154-109441154 on Build GRCh38
rs897438085 Chr.6: 109440754-109440754 on Build GRCh38
rs755479968 Chr.6: 109440750-109440750 on Build GRCh38

View Full Product Details