Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5481262-5481757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL18
Entrez Gene ID
80028
Gene Name
F-box and leucine rich repeat protein 18
Gene Aliases
Fbl18
UniGene
Hs.623974
Gene Chromosome Location
Chr.7: 5454448-5513813 on Build GRCh38
Transcripts
NM_024963.5 XM_011515531.2

SNP Detail

SNP ID Chromosome Location
rs1052027656 Chr.7: 5481348-5481348 on Build GRCh38
rs569752019 Chr.7: 5481379-5481379 on Build GRCh38
rs767571434 Chr.7: 5481668-5481668 on Build GRCh38
rs1023243903 Chr.7: 5481566-5481566 on Build GRCh38
rs933261746 Chr.7: 5481336-5481336 on Build GRCh38
rs781042738 Chr.7: 5481315-5481315 on Build GRCh38
rs888935685 Chr.7: 5481504-5481504 on Build GRCh38
rs895456643 Chr.7: 5481684-5481684 on Build GRCh38
rs984204897 Chr.7: 5481412-5481412 on Build GRCh38
rs978061025 Chr.7: 5481472-5481472 on Build GRCh38
rs994462077 Chr.7: 5481404-5481404 on Build GRCh38
rs139102528 Chr.7: 5481567-5481567 on Build GRCh38
rs61447150 Chr.7: 5481359-5481359 on Build GRCh38
rs925996407 Chr.7: 5481457-5481457 on Build GRCh38
rs376163857 Chr.7: 5481651-5481651 on Build GRCh38
rs183842235 Chr.7: 5481624-5481624 on Build GRCh38
rs569402291 Chr.7: 5481717-5481717 on Build GRCh38
rs543079662 Chr.7: 5481573-5481573 on Build GRCh38
rs1051711530 Chr.7: 5481493-5481493 on Build GRCh38
rs192306991 Chr.7: 5481718-5481718 on Build GRCh38
rs551310427 Chr.7: 5481672-5481672 on Build GRCh38
rs936312144 Chr.7: 5481509-5481509 on Build GRCh38
rs10253801 Chr.7: 5481406-5481406 on Build GRCh38
rs923643208 Chr.7: 5481473-5481473 on Build GRCh38
rs796363002 Chr.7: 5481421-5481421 on Build GRCh38
rs561230642 Chr.7: 5481583-5481583 on Build GRCh38
rs895012993 Chr.7: 5481324-5481324 on Build GRCh38
rs189622133 Chr.7: 5481632-5481632 on Build GRCh38
rs1025513571 Chr.7: 5481557-5481557 on Build GRCh38
rs117256601 Chr.7: 5481723-5481723 on Build GRCh38
rs552840872 Chr.7: 5481508-5481508 on Build GRCh38
rs1000643500 Chr.7: 5481314-5481314 on Build GRCh38
rs967950301 Chr.7: 5481585-5481585 on Build GRCh38
rs929807400 Chr.7: 5481686-5481686 on Build GRCh38
rs1046841437 Chr.7: 5481716-5481716 on Build GRCh38
rs1014901927 Chr.7: 5481560-5481560 on Build GRCh38
rs769906499 Chr.7: 5481571-5481571 on Build GRCh38
rs1033178717 Chr.7: 5481574-5481574 on Build GRCh38
rs549994437 Chr.7: 5481631-5481631 on Build GRCh38
rs1015996311 Chr.7: 5481368-5481368 on Build GRCh38
rs1044470992 Chr.7: 5481715-5481715 on Build GRCh38
rs10046575 Chr.7: 5481298-5481298 on Build GRCh38
rs898932682 Chr.7: 5481389-5481389 on Build GRCh38
rs1013892144 Chr.7: 5481714-5481714 on Build GRCh38
rs369591063 Chr.7: 5481303-5481303 on Build GRCh38
rs546716548 Chr.7: 5481448-5481448 on Build GRCh38
rs10046576 Chr.7: 5481588-5481588 on Build GRCh38
rs934747978 Chr.7: 5481462-5481462 on Build GRCh38
rs1012093061 Chr.7: 5481337-5481337 on Build GRCh38
rs947857845 Chr.7: 5481634-5481634 on Build GRCh38
rs577565044 Chr.7: 5481525-5481525 on Build GRCh38
rs948807479 Chr.7: 5481302-5481302 on Build GRCh38
rs1046241277 Chr.7: 5481538-5481538 on Build GRCh38
rs898880277 Chr.7: 5481530-5481530 on Build GRCh38
rs899369355 Chr.7: 5481431-5481431 on Build GRCh38
rs539231805 Chr.7: 5481318-5481318 on Build GRCh38
rs561403043 Chr.7: 5481607-5481607 on Build GRCh38
rs920462759 Chr.7: 5481656-5481656 on Build GRCh38
rs964059886 Chr.7: 5481434-5481434 on Build GRCh38
rs537113006 Chr.7: 5481391-5481391 on Build GRCh38
rs911316073 Chr.7: 5481433-5481433 on Build GRCh38
rs111347877 Chr.7: 5481406-5481406 on Build GRCh38
rs1029856384 Chr.7: 5481584-5481584 on Build GRCh38
rs550871977 Chr.7: 5481635-5481635 on Build GRCh38
rs373348676 Chr.7: 5481357-5481362 on Build GRCh38
rs947770772 Chr.7: 5481528-5481528 on Build GRCh38
rs775991160 Chr.7: 5481670-5481670 on Build GRCh38
rs757029843 Chr.7: 5481728-5481728 on Build GRCh38
rs1053414172 Chr.7: 5481515-5481515 on Build GRCh38
rs933233146 Chr.7: 5481482-5481482 on Build GRCh38
rs964111492 Chr.7: 5481578-5481578 on Build GRCh38
rs929054144 Chr.7: 5481660-5481660 on Build GRCh38
rs749423541 Chr.7: 5481617-5481617 on Build GRCh38
rs761015489 Chr.7: 5481691-5481691 on Build GRCh38
rs970687756 Chr.7: 5481362-5481362 on Build GRCh38
rs569173848 Chr.7: 5481690-5481690 on Build GRCh38
rs976966799 Chr.7: 5481446-5481446 on Build GRCh38
rs146201606 Chr.7: 5481550-5481550 on Build GRCh38
rs1025927647 Chr.7: 5481549-5481549 on Build GRCh38
rs571613813 Chr.7: 5481456-5481456 on Build GRCh38
rs367852114 Chr.7: 5481623-5481623 on Build GRCh38
rs926198518 Chr.7: 5481606-5481606 on Build GRCh38
rs143172313 Chr.7: 5481405-5481405 on Build GRCh38
rs906344538 Chr.7: 5481542-5481542 on Build GRCh38
rs544785483 Chr.7: 5481529-5481529 on Build GRCh38

View Full Product Details