Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6163162-6163665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH3
Entrez Gene ID
9265
Gene Name
cytohesin 3
Gene Aliases
ARNO3, GRP1, PSCD3
UniGene
Hs.487479
Gene Chromosome Location
Chr.7: 6161779-6272611 on Build GRCh38
Transcripts
NM_004227.3

SNP Detail

SNP ID Chromosome Location
rs377336829 Chr.7: 6163403-6163403 on Build GRCh38
rs995814517 Chr.7: 6163580-6163580 on Build GRCh38
rs78314028 Chr.7: 6163445-6163445 on Build GRCh38
rs1027384461 Chr.7: 6163581-6163581 on Build GRCh38
rs892399744 Chr.7: 6163260-6163260 on Build GRCh38
rs893913837 Chr.7: 6163310-6163310 on Build GRCh38
rs916882774 Chr.7: 6163405-6163405 on Build GRCh38
rs1043405836 Chr.7: 6163277-6163277 on Build GRCh38
rs1037422116 Chr.7: 6163195-6163195 on Build GRCh38
rs925204615 Chr.7: 6163638-6163638 on Build GRCh38
rs755371969 Chr.7: 6163334-6163334 on Build GRCh38
rs961647021 Chr.7: 6163193-6163193 on Build GRCh38
rs937731387 Chr.7: 6163484-6163484 on Build GRCh38
rs1001993618 Chr.7: 6163308-6163308 on Build GRCh38
rs6953241 Chr.7: 6163600-6163600 on Build GRCh38
rs745406029 Chr.7: 6163478-6163478 on Build GRCh38
rs955128425 Chr.7: 6163361-6163361 on Build GRCh38
rs563678157 Chr.7: 6163503-6163503 on Build GRCh38
rs910762467 Chr.7: 6163194-6163194 on Build GRCh38
rs999997333 Chr.7: 6163296-6163296 on Build GRCh38
rs945881085 Chr.7: 6163641-6163641 on Build GRCh38
rs771958089 Chr.7: 6163479-6163479 on Build GRCh38
rs182134012 Chr.7: 6163329-6163329 on Build GRCh38
rs985535579 Chr.7: 6163392-6163392 on Build GRCh38
rs1014210767 Chr.7: 6163242-6163242 on Build GRCh38
rs1040237870 Chr.7: 6163258-6163258 on Build GRCh38
rs935955434 Chr.7: 6163611-6163611 on Build GRCh38
rs756213765 Chr.7: 6163602-6163602 on Build GRCh38
rs780734769 Chr.7: 6163276-6163276 on Build GRCh38
rs987704226 Chr.7: 6163361-6163361 on Build GRCh38
rs140418877 Chr.7: 6163636-6163636 on Build GRCh38
rs1043781121 Chr.7: 6163460-6163460 on Build GRCh38
rs577986827 Chr.7: 6163470-6163470 on Build GRCh38
rs913772871 Chr.7: 6163454-6163454 on Build GRCh38
rs370549340 Chr.7: 6163244-6163244 on Build GRCh38
rs949721037 Chr.7: 6163592-6163592 on Build GRCh38
rs1033830451 Chr.7: 6163319-6163319 on Build GRCh38
rs545463471 Chr.7: 6163493-6163493 on Build GRCh38
rs767141918 Chr.7: 6163216-6163223 on Build GRCh38
rs1044707098 Chr.7: 6163244-6163244 on Build GRCh38
rs929073449 Chr.7: 6163399-6163399 on Build GRCh38
rs480919 Chr.7: 6163495-6163495 on Build GRCh38
rs954128665 Chr.7: 6163379-6163379 on Build GRCh38
rs373340636 Chr.7: 6163295-6163295 on Build GRCh38
rs773678639 Chr.7: 6163467-6163467 on Build GRCh38
rs983520194 Chr.7: 6163401-6163401 on Build GRCh38
rs1013724401 Chr.7: 6163498-6163498 on Build GRCh38
rs151216611 Chr.7: 6163374-6163374 on Build GRCh38
rs920329203 Chr.7: 6163383-6163383 on Build GRCh38
rs964982951 Chr.7: 6163557-6163557 on Build GRCh38
rs1054248099 Chr.7: 6163475-6163475 on Build GRCh38
rs928932756 Chr.7: 6163219-6163219 on Build GRCh38
rs35044788 Chr.7: 6163644-6163645 on Build GRCh38
rs913774574 Chr.7: 6163615-6163615 on Build GRCh38
rs781663000 Chr.7: 6163340-6163340 on Build GRCh38
rs965439743 Chr.7: 6163607-6163607 on Build GRCh38
rs895714195 Chr.7: 6163225-6163225 on Build GRCh38
rs1027863183 Chr.7: 6163575-6163575 on Build GRCh38
rs537291713 Chr.7: 6163264-6163264 on Build GRCh38
rs748886534 Chr.7: 6163369-6163369 on Build GRCh38
rs954015549 Chr.7: 6163599-6163599 on Build GRCh38
rs976312720 Chr.7: 6163610-6163610 on Build GRCh38
rs370197217 Chr.7: 6163204-6163204 on Build GRCh38
rs754817756 Chr.7: 6163427-6163427 on Build GRCh38
rs1047418823 Chr.7: 6163223-6163223 on Build GRCh38
rs974448284 Chr.7: 6163365-6163365 on Build GRCh38
rs961653881 Chr.7: 6163364-6163364 on Build GRCh38
rs1050214187 Chr.7: 6163220-6163220 on Build GRCh38
rs897394018 Chr.7: 6163579-6163579 on Build GRCh38
rs949757880 Chr.7: 6163441-6163441 on Build GRCh38
rs973558881 Chr.7: 6163572-6163572 on Build GRCh38
rs910407296 Chr.7: 6163235-6163235 on Build GRCh38
rs919967946 Chr.7: 6163208-6163208 on Build GRCh38
rs529176298 Chr.7: 6163620-6163620 on Build GRCh38
rs896256427 Chr.7: 6163494-6163494 on Build GRCh38
rs893885928 Chr.7: 6163483-6163483 on Build GRCh38
rs775375596 Chr.7: 6163555-6163555 on Build GRCh38
rs916996881 Chr.7: 6163590-6163590 on Build GRCh38
rs142184977 Chr.7: 6163212-6163212 on Build GRCh38
rs1015773506 Chr.7: 6163526-6163526 on Build GRCh38
rs377599404 Chr.7: 6163643-6163643 on Build GRCh38
rs932748657 Chr.7: 6163210-6163210 on Build GRCh38
rs1017833022 Chr.7: 6163523-6163523 on Build GRCh38
rs561869437 Chr.7: 6163606-6163606 on Build GRCh38
rs186074177 Chr.7: 6163551-6163551 on Build GRCh38
rs944610260 Chr.7: 6163256-6163256 on Build GRCh38
rs1016237559 Chr.7: 6163349-6163349 on Build GRCh38
rs945347002 Chr.7: 6163459-6163459 on Build GRCh38

View Full Product Details