Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 15626692-15627228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEOX2
Entrez Gene ID
4223
Gene Name
mesenchyme homeobox 2
Gene Aliases
GAX, MOX2
UniGene
Hs.170355
Gene Chromosome Location
Chr.7: 15611212-15686683 on Build GRCh38
Transcripts
NM_005924.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6033719 c.649C>T p.R217* Chr.7: 15626787-15626787 on Build GRCh38
4701747 c.634C>A p.L212I Chr.7: 15626802-15626802 on Build GRCh38
1449650 c.524A>T p.Q175L Chr.7: 15626912-15626912 on Build GRCh38
6711462 c.640A>C p.R214R Chr.7: 15626796-15626796 on Build GRCh38
4701748 c.537C>G p.Y179* Chr.7: 15626899-15626899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989377131 Chr.7: 15626951-15626951 on Build GRCh38
rs944447392 Chr.7: 15627040-15627040 on Build GRCh38
rs779485604 Chr.7: 15626889-15626889 on Build GRCh38
rs769307951 Chr.7: 15626721-15626721 on Build GRCh38
rs765654736 Chr.7: 15626969-15626969 on Build GRCh38
rs908300227 Chr.7: 15627078-15627078 on Build GRCh38
rs915199366 Chr.7: 15626964-15626964 on Build GRCh38
rs544600165 Chr.7: 15627063-15627063 on Build GRCh38
rs569517497 Chr.7: 15626886-15626886 on Build GRCh38
rs1033755345 Chr.7: 15627181-15627181 on Build GRCh38
rs1024904763 Chr.7: 15626756-15626756 on Build GRCh38
rs113001727 Chr.7: 15627126-15627127 on Build GRCh38
rs1054471680 Chr.7: 15627155-15627159 on Build GRCh38
rs759063285 Chr.7: 15626926-15626926 on Build GRCh38
rs540917999 Chr.7: 15626718-15626718 on Build GRCh38
rs762865770 Chr.7: 15626816-15626816 on Build GRCh38
rs779105084 Chr.7: 15626723-15626723 on Build GRCh38
rs574754777 Chr.7: 15627182-15627182 on Build GRCh38
rs769606593 Chr.7: 15626927-15626927 on Build GRCh38
rs529659583 Chr.7: 15627071-15627071 on Build GRCh38
rs752379059 Chr.7: 15626815-15626815 on Build GRCh38
rs763998346 Chr.7: 15626942-15626942 on Build GRCh38
rs560701668 Chr.7: 15626999-15626999 on Build GRCh38
rs926571321 Chr.7: 15626973-15626973 on Build GRCh38
rs931162818 Chr.7: 15627036-15627036 on Build GRCh38
rs146520307 Chr.7: 15627015-15627015 on Build GRCh38
rs772551159 Chr.7: 15626893-15626893 on Build GRCh38
rs141068618 Chr.7: 15627154-15627154 on Build GRCh38
rs547309136 Chr.7: 15627030-15627030 on Build GRCh38
rs1027334382 Chr.7: 15627202-15627202 on Build GRCh38
rs745306412 Chr.7: 15627061-15627061 on Build GRCh38
rs751422050 Chr.7: 15626947-15626947 on Build GRCh38
rs563767319 Chr.7: 15626976-15626976 on Build GRCh38
rs1046869528 Chr.7: 15627076-15627076 on Build GRCh38
rs777256420 Chr.7: 15626902-15626902 on Build GRCh38
rs541993792 Chr.7: 15626956-15626956 on Build GRCh38
rs982117613 Chr.7: 15626968-15626968 on Build GRCh38
rs900099787 Chr.7: 15626735-15626735 on Build GRCh38
rs531030875 Chr.7: 15626977-15626977 on Build GRCh38
rs1038772111 Chr.7: 15627057-15627057 on Build GRCh38
rs776144357 Chr.7: 15626791-15626791 on Build GRCh38
rs867233014 Chr.7: 15626906-15626906 on Build GRCh38
rs766711223 Chr.7: 15626958-15626958 on Build GRCh38
rs765228491 Chr.7: 15626814-15626814 on Build GRCh38
rs548002691 Chr.7: 15627072-15627072 on Build GRCh38
rs754001486 Chr.7: 15626875-15626875 on Build GRCh38
rs780232088 Chr.7: 15626865-15626865 on Build GRCh38
rs370166690 Chr.7: 15626922-15626922 on Build GRCh38
rs771564049 Chr.7: 15626737-15626737 on Build GRCh38
rs7800593 Chr.7: 15627124-15627124 on Build GRCh38
rs1010164328 Chr.7: 15627166-15627166 on Build GRCh38
rs181631911 Chr.7: 15626733-15626733 on Build GRCh38
rs886660071 Chr.7: 15627101-15627101 on Build GRCh38
rs1021089466 Chr.7: 15627173-15627173 on Build GRCh38
rs753910321 Chr.7: 15626961-15626961 on Build GRCh38
rs911716696 Chr.7: 15627027-15627027 on Build GRCh38
rs756240195 Chr.7: 15626845-15626845 on Build GRCh38
rs113782262 Chr.7: 15627125-15627125 on Build GRCh38
rs894035963 Chr.7: 15627156-15627156 on Build GRCh38
rs762624873 Chr.7: 15626941-15626941 on Build GRCh38
rs1003669572 Chr.7: 15627180-15627180 on Build GRCh38
rs563641807 Chr.7: 15626782-15626782 on Build GRCh38
rs973409075 Chr.7: 15627190-15627190 on Build GRCh38
rs1024455242 Chr.7: 15627106-15627106 on Build GRCh38
rs570313531 Chr.7: 15627153-15627153 on Build GRCh38
rs775566088 Chr.7: 15626932-15626932 on Build GRCh38
rs371557289 Chr.7: 15626891-15626891 on Build GRCh38
rs569686467 Chr.7: 15627073-15627073 on Build GRCh38
rs7781465 Chr.7: 15627126-15627126 on Build GRCh38
rs113309389 Chr.7: 15627124-15627124 on Build GRCh38
rs552834542 Chr.7: 15627179-15627179 on Build GRCh38
rs564779153 Chr.7: 15627018-15627018 on Build GRCh38
rs755141457 Chr.7: 15626963-15626963 on Build GRCh38
rs375199918 Chr.7: 15626825-15626825 on Build GRCh38
rs746619188 Chr.7: 15626911-15626911 on Build GRCh38
rs759433211 Chr.7: 15626809-15626809 on Build GRCh38
rs773104935 Chr.7: 15626727-15626727 on Build GRCh38
rs202182874 Chr.7: 15627019-15627019 on Build GRCh38
rs760169461 Chr.7: 15626786-15626786 on Build GRCh38
rs560428266 Chr.7: 15627193-15627193 on Build GRCh38
rs542176643 Chr.7: 15627203-15627203 on Build GRCh38
rs574755420 Chr.7: 15626788-15626788 on Build GRCh38
rs776056051 Chr.7: 15626916-15626916 on Build GRCh38
rs959358257 Chr.7: 15626975-15626975 on Build GRCh38
rs755468003 Chr.7: 15626877-15626877 on Build GRCh38
rs565566970 Chr.7: 15627060-15627060 on Build GRCh38
rs71525655 Chr.7: 15627124-15627126 on Build GRCh38
rs760674308 Chr.7: 15626921-15626922 on Build GRCh38
rs966613224 Chr.7: 15627197-15627197 on Build GRCh38
rs112262227 Chr.7: 15626833-15626833 on Build GRCh38
rs770604890 Chr.7: 15626912-15626912 on Build GRCh38
rs1005547944 Chr.7: 15627102-15627102 on Build GRCh38
rs113051023 Chr.7: 15627062-15627062 on Build GRCh38
rs990181315 Chr.7: 15627176-15627176 on Build GRCh38
rs750714411 Chr.7: 15626842-15626842 on Build GRCh38
rs957353784 Chr.7: 15627167-15627167 on Build GRCh38

View Full Product Details