Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81967504-81967953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D1
Entrez Gene ID
781
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
Gene Aliases
CACNA2, CACNL2A, CCHL2A, LINC01112, lncRNA-N3
UniGene
Hs.282151
Gene Chromosome Location
Chr.7: 81946444-82443921 on Build GRCh38
Transcripts
NM_000722.3 XM_005250570.2 XM_005250572.2 XM_005250573.2 XM_005250574.2 XM_006716118.2 XM_006716119.3 XM_006716120.3 XM_006716121.3 XM_011516570.2 XM_011516571.2 XM_011516572.2 XM_017012588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4480869 c.2461C>T p.P821S Chr.7: 81967598-81967598 on Build GRCh38
6248753 c.2454C>A p.I818I Chr.7: 81967605-81967605 on Build GRCh38
5413916 c.2453T>C p.V818A Chr.7: 81967642-81967642 on Build GRCh38
5413915 c.2417T>C p.V806A Chr.7: 81967642-81967642 on Build GRCh38
7205144 c.2454A>G p.V818V Chr.7: 81967641-81967641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781351390 Chr.7: 81967565-81967565 on Build GRCh38
rs755356586 Chr.7: 81967563-81967563 on Build GRCh38
rs978366900 Chr.7: 81967901-81967901 on Build GRCh38
rs559533450 Chr.7: 81967550-81967550 on Build GRCh38
rs1005281458 Chr.7: 81967532-81967533 on Build GRCh38
rs139526006 Chr.7: 81967756-81967756 on Build GRCh38
rs761540201 Chr.7: 81967679-81967679 on Build GRCh38
rs776613213 Chr.7: 81967675-81967675 on Build GRCh38
rs576164504 Chr.7: 81967786-81967789 on Build GRCh38
rs1012319105 Chr.7: 81967910-81967910 on Build GRCh38
rs889520047 Chr.7: 81967530-81967530 on Build GRCh38
rs749832863 Chr.7: 81967713-81967713 on Build GRCh38
rs896963742 Chr.7: 81967860-81967860 on Build GRCh38
rs894840717 Chr.7: 81967905-81967905 on Build GRCh38
rs1048155369 Chr.7: 81967897-81967897 on Build GRCh38
rs943932875 Chr.7: 81967853-81967853 on Build GRCh38
rs990168565 Chr.7: 81967629-81967629 on Build GRCh38
rs1026564694 Chr.7: 81967620-81967620 on Build GRCh38
rs1036875225 Chr.7: 81967855-81967855 on Build GRCh38
rs749391834 Chr.7: 81967582-81967582 on Build GRCh38
rs775523046 Chr.7: 81967609-81967609 on Build GRCh38
rs773503339 Chr.7: 81967560-81967561 on Build GRCh38
rs985426642 Chr.7: 81967805-81967805 on Build GRCh38
rs565224102 Chr.7: 81967570-81967570 on Build GRCh38
rs536011074 Chr.7: 81967851-81967851 on Build GRCh38
rs982019965 Chr.7: 81967560-81967560 on Build GRCh38
rs562538998 Chr.7: 81967539-81967539 on Build GRCh38
rs749669650 Chr.7: 81967792-81967795 on Build GRCh38
rs1023626350 Chr.7: 81967543-81967543 on Build GRCh38
rs1022512982 Chr.7: 81967824-81967824 on Build GRCh38
rs747704505 Chr.7: 81967701-81967701 on Build GRCh38
rs1033706230 Chr.7: 81967727-81967728 on Build GRCh38
rs932628430 Chr.7: 81967748-81967748 on Build GRCh38
rs191213592 Chr.7: 81967662-81967662 on Build GRCh38
rs769205599 Chr.7: 81967862-81967862 on Build GRCh38
rs989579882 Chr.7: 81967803-81967803 on Build GRCh38
rs551354977 Chr.7: 81967551-81967551 on Build GRCh38
rs200135341 Chr.7: 81967574-81967574 on Build GRCh38
rs771850068 Chr.7: 81967583-81967583 on Build GRCh38
rs748338081 Chr.7: 81967567-81967567 on Build GRCh38
rs765004029 Chr.7: 81967704-81967704 on Build GRCh38
rs760509803 Chr.7: 81967612-81967612 on Build GRCh38
rs912468834 Chr.7: 81967836-81967836 on Build GRCh38
rs1024604030 Chr.7: 81967930-81967930 on Build GRCh38

View Full Product Details