Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134531803-134532309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1B10
Entrez Gene ID
57016
Gene Name
aldo-keto reductase family 1 member B10
Gene Aliases
AKR1B11, AKR1B12, ALDRLn, ARL-1, ARL1, HIS, HSI
UniGene
Hs.116724
Gene Chromosome Location
Chr.7: 134527592-134541414 on Build GRCh38
Transcripts
NM_020299.4 XM_011516416.1 XM_011516417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6176622 c.241C>T p.P81S Chr.7: 134531914-134531914 on Build GRCh38
6597943 c.269G>T p.R90M Chr.7: 134531942-134531942 on Build GRCh38
1244789 c.331C>A p.H111N Chr.7: 134532004-134532004 on Build GRCh38
3633566 c.289C>T p.L97F Chr.7: 134531962-134531962 on Build GRCh38
6176621 c.339A>G p.P113P Chr.7: 134532012-134532012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563657501 Chr.7: 134531989-134531989 on Build GRCh38
rs535644853 Chr.7: 134532102-134532102 on Build GRCh38
rs1646765 Chr.7: 134532100-134532100 on Build GRCh38
rs760115087 Chr.7: 134532061-134532061 on Build GRCh38
rs985383343 Chr.7: 134532142-134532142 on Build GRCh38
rs1020660699 Chr.7: 134531841-134531841 on Build GRCh38
rs912439927 Chr.7: 134532145-134532145 on Build GRCh38
rs547344212 Chr.7: 134532236-134532236 on Build GRCh38
rs146306673 Chr.7: 134532016-134532016 on Build GRCh38
rs747618966 Chr.7: 134532039-134532039 on Build GRCh38
rs775424720 Chr.7: 134531916-134531916 on Build GRCh38
rs138234669 Chr.7: 134532005-134532005 on Build GRCh38
rs1013379179 Chr.7: 134531828-134531828 on Build GRCh38
rs150425053 Chr.7: 134531982-134531982 on Build GRCh38
rs372587864 Chr.7: 134531992-134531992 on Build GRCh38
rs555974466 Chr.7: 134532170-134532170 on Build GRCh38
rs760962667 Chr.7: 134531965-134531965 on Build GRCh38
rs773560659 Chr.7: 134531961-134531961 on Build GRCh38
rs2303312 Chr.7: 134531932-134531932 on Build GRCh38
rs766861648 Chr.7: 134531967-134531967 on Build GRCh38
rs143396171 Chr.7: 134531908-134531908 on Build GRCh38
rs752761942 Chr.7: 134532072-134532072 on Build GRCh38
rs151300112 Chr.7: 134531930-134531930 on Build GRCh38
rs201621939 Chr.7: 134532039-134532040 on Build GRCh38
rs140738774 Chr.7: 134532013-134532013 on Build GRCh38
rs772762928 Chr.7: 134532044-134532044 on Build GRCh38
rs755888697 Chr.7: 134532001-134532001 on Build GRCh38
rs751248667 Chr.7: 134531860-134531860 on Build GRCh38
rs754365410 Chr.7: 134531968-134531968 on Build GRCh38
rs202083279 Chr.7: 134532012-134532012 on Build GRCh38
rs931443084 Chr.7: 134532211-134532211 on Build GRCh38
rs530880920 Chr.7: 134532060-134532060 on Build GRCh38
rs775428242 Chr.7: 134531956-134531956 on Build GRCh38
rs560588394 Chr.7: 134531882-134531882 on Build GRCh38
rs953989764 Chr.7: 134531913-134531913 on Build GRCh38
rs559318632 Chr.7: 134532160-134532160 on Build GRCh38
rs374158443 Chr.7: 134531934-134531934 on Build GRCh38
rs769711685 Chr.7: 134531915-134531915 on Build GRCh38
rs1048948576 Chr.7: 134532219-134532219 on Build GRCh38
rs546243178 Chr.7: 134532096-134532096 on Build GRCh38
rs953708632 Chr.7: 134532129-134532129 on Build GRCh38
rs960186900 Chr.7: 134531824-134531824 on Build GRCh38
rs568395530 Chr.7: 134531970-134531970 on Build GRCh38
rs868026095 Chr.7: 134532035-134532036 on Build GRCh38
rs753912533 Chr.7: 134531895-134531895 on Build GRCh38
rs569429917 Chr.7: 134532178-134532178 on Build GRCh38
rs755590380 Chr.7: 134531975-134531975 on Build GRCh38
rs142873661 Chr.7: 134532066-134532066 on Build GRCh38
rs766828447 Chr.7: 134532057-134532057 on Build GRCh38
rs765392907 Chr.7: 134531976-134531976 on Build GRCh38
rs771037167 Chr.7: 134531874-134531874 on Build GRCh38
rs745653060 Chr.7: 134531914-134531914 on Build GRCh38
rs755130976 Chr.7: 134531906-134531906 on Build GRCh38
rs927317242 Chr.7: 134532222-134532222 on Build GRCh38
rs189552034 Chr.7: 134532078-134532078 on Build GRCh38
rs748752793 Chr.7: 134531921-134531921 on Build GRCh38
rs937283416 Chr.7: 134532244-134532244 on Build GRCh38
rs761060865 Chr.7: 134532049-134532049 on Build GRCh38
rs769653900 Chr.7: 134531931-134531932 on Build GRCh38
rs763675781 Chr.7: 134532068-134532069 on Build GRCh38
rs758605056 Chr.7: 134531909-134531909 on Build GRCh38
rs779869150 Chr.7: 134532004-134532004 on Build GRCh38
rs111241649 Chr.7: 134532252-134532252 on Build GRCh38
rs1001820298 Chr.7: 134532024-134532024 on Build GRCh38
rs745623300 Chr.7: 134531999-134531999 on Build GRCh38
rs201546370 Chr.7: 134532085-134532085 on Build GRCh38
rs758577784 Chr.7: 134531991-134531991 on Build GRCh38
rs766802093 Chr.7: 134531893-134531893 on Build GRCh38
rs918397172 Chr.7: 134532200-134532200 on Build GRCh38
rs761497720 Chr.7: 134531862-134531862 on Build GRCh38
rs767904769 Chr.7: 134531875-134531875 on Build GRCh38
rs756641916 Chr.7: 134531883-134531883 on Build GRCh38
rs781034480 Chr.7: 134531911-134531911 on Build GRCh38
rs574846405 Chr.7: 134531857-134531857 on Build GRCh38

View Full Product Details