Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134646701-134647209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPGM
Entrez Gene ID
669
Gene Name
bisphosphoglycerate mutase
Gene Aliases
DPGM
UniGene
Hs.198365
Gene Chromosome Location
Chr.7: 134646779-134679816 on Build GRCh38
Transcripts
NM_001293085.1 NM_001724.4 NM_199186.2

SNP Detail

SNP ID Chromosome Location
rs1039989586 Chr.7: 134647045-134647045 on Build GRCh38
rs750272258 Chr.7: 134646979-134646979 on Build GRCh38
rs1028607682 Chr.7: 134647128-134647128 on Build GRCh38
rs552405463 Chr.7: 134647017-134647017 on Build GRCh38
rs767275673 Chr.7: 134646962-134646962 on Build GRCh38
rs1036671358 Chr.7: 134646819-134646819 on Build GRCh38
rs1034767312 Chr.7: 134646859-134646859 on Build GRCh38
rs984320624 Chr.7: 134646883-134646883 on Build GRCh38
rs546082308 Chr.7: 134646880-134646880 on Build GRCh38
rs1052670338 Chr.7: 134646823-134646823 on Build GRCh38
rs528791424 Chr.7: 134646885-134646885 on Build GRCh38
rs892616567 Chr.7: 134646863-134646863 on Build GRCh38
rs551130034 Chr.7: 134646971-134646971 on Build GRCh38
rs79065225 Chr.7: 134646920-134646920 on Build GRCh38
rs946566228 Chr.7: 134646844-134646844 on Build GRCh38
rs953019248 Chr.7: 134646754-134646754 on Build GRCh38
rs543139686 Chr.7: 134646836-134646836 on Build GRCh38
rs926285867 Chr.7: 134646945-134646945 on Build GRCh38
rs988226156 Chr.7: 134646917-134646917 on Build GRCh38
rs541822114 Chr.7: 134646886-134646886 on Build GRCh38
rs930581931 Chr.7: 134646920-134646921 on Build GRCh38
rs568080168 Chr.7: 134647085-134647085 on Build GRCh38
rs559749184 Chr.7: 134646881-134646881 on Build GRCh38
rs140878347 Chr.7: 134646994-134646994 on Build GRCh38
rs969597733 Chr.7: 134647181-134647181 on Build GRCh38
rs151046022 Chr.7: 134646915-134646915 on Build GRCh38
rs927998308 Chr.7: 134646805-134646805 on Build GRCh38
rs911719350 Chr.7: 134646807-134646807 on Build GRCh38
rs930275651 Chr.7: 134646839-134646839 on Build GRCh38
rs920473516 Chr.7: 134646898-134646909 on Build GRCh38
rs113326034 Chr.7: 134646871-134646871 on Build GRCh38
rs763035396 Chr.7: 134646846-134646846 on Build GRCh38
rs897470194 Chr.7: 134647048-134647048 on Build GRCh38
rs572930259 Chr.7: 134646896-134646897 on Build GRCh38
rs1027395483 Chr.7: 134647064-134647064 on Build GRCh38
rs935359494 Chr.7: 134646900-134646900 on Build GRCh38
rs764105628 Chr.7: 134646869-134646869 on Build GRCh38
rs144930222 Chr.7: 134647074-134647074 on Build GRCh38
rs775580896 Chr.7: 134646831-134646831 on Build GRCh38
rs955391060 Chr.7: 134646889-134646889 on Build GRCh38
rs982084617 Chr.7: 134646888-134646888 on Build GRCh38
rs564127004 Chr.7: 134646897-134646898 on Build GRCh38
rs760001766 Chr.7: 134646900-134646901 on Build GRCh38
rs993606757 Chr.7: 134647053-134647053 on Build GRCh38
rs961493073 Chr.7: 134647185-134647185 on Build GRCh38
rs377558203 Chr.7: 134646976-134646976 on Build GRCh38
rs939050670 Chr.7: 134646961-134646961 on Build GRCh38
rs986868786 Chr.7: 134646894-134646895 on Build GRCh38
rs982115957 Chr.7: 134646804-134646804 on Build GRCh38
rs927998581 Chr.7: 134646897-134646897 on Build GRCh38
rs997093104 Chr.7: 134647092-134647092 on Build GRCh38
rs181961890 Chr.7: 134647018-134647018 on Build GRCh38
rs916871694 Chr.7: 134646758-134646758 on Build GRCh38
rs547467045 Chr.7: 134646864-134646864 on Build GRCh38
rs1044718629 Chr.7: 134646842-134646842 on Build GRCh38
rs373411794 Chr.7: 134646847-134646847 on Build GRCh38
rs935446815 Chr.7: 134646809-134646809 on Build GRCh38
rs755418090 Chr.7: 134646786-134646786 on Build GRCh38
rs754460251 Chr.7: 134647167-134647167 on Build GRCh38
rs748906685 Chr.7: 134647122-134647122 on Build GRCh38
rs765622973 Chr.7: 134646755-134646755 on Build GRCh38
rs531886661 Chr.7: 134646740-134646740 on Build GRCh38
rs1008485967 Chr.7: 134646858-134646858 on Build GRCh38
rs955339154 Chr.7: 134647164-134647164 on Build GRCh38
rs1018570204 Chr.7: 134646862-134646862 on Build GRCh38
rs2303314 Chr.7: 134646875-134646875 on Build GRCh38
rs952905092 Chr.7: 134646877-134646877 on Build GRCh38
rs147174635 Chr.7: 134646898-134646900 on Build GRCh38
rs754285073 Chr.7: 134646778-134646778 on Build GRCh38
rs986922578 Chr.7: 134646770-134646770 on Build GRCh38
rs534934708 Chr.7: 134647026-134647026 on Build GRCh38

View Full Product Details