Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139341433-139341936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf55-LUC7L2
Entrez Gene ID
100996928
Gene Name
C7orf55-LUC7L2 readthrough
Gene Aliases
-
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139341132-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_197964.4 NR_073058.1 NR_073059.1 NR_073060.1 NM_001244585.1

Gene Symbol
FMC1
Entrez Gene ID
154791
Gene Name
formation of mitochondrial complex V assembly factor 1 homolog
Gene Aliases
C7orf55, HSPC268
UniGene
Hs.718441
Gene Chromosome Location
Chr.7: 139340450-139346319 on Build GRCh38
Transcripts
NM_001244584.2 NM_197964.4 NR_073058.1 NR_073059.1 NR_073060.1 NM_001244585.1

Gene Symbol
LUC7L2
Entrez Gene ID
51631
Gene Name
LUC7 like 2, pre-mRNA splicing factor
Gene Aliases
CGI-59, CGI-74, LUC7B2
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139340359-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_197964.4 NR_073058.1 NR_073059.1 NR_073060.1 NM_001244585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580461 c.110A>G p.Y37C Chr.7: 139341494-139341494 on Build GRCh38
1312731 c.106C>G p.R36G Chr.7: 139341490-139341490 on Build GRCh38
6515666 c.56G>A p.R19H Chr.7: 139341440-139341440 on Build GRCh38
4389824 c.72C>T p.A24A Chr.7: 139341456-139341456 on Build GRCh38
745407 c.61C>G p.L21V Chr.7: 139341445-139341445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745404460 Chr.7: 139341527-139341527 on Build GRCh38
rs561687926 Chr.7: 139341720-139341720 on Build GRCh38
rs138945070 Chr.7: 139341768-139341768 on Build GRCh38
rs571616041 Chr.7: 139341500-139341500 on Build GRCh38
rs756177561 Chr.7: 139341508-139341508 on Build GRCh38
rs928347397 Chr.7: 139341666-139341666 on Build GRCh38
rs762406780 Chr.7: 139341488-139341488 on Build GRCh38
rs775057665 Chr.7: 139341484-139341484 on Build GRCh38
rs540943665 Chr.7: 139341700-139341700 on Build GRCh38
rs758182186 Chr.7: 139341512-139341512 on Build GRCh38
rs1020674576 Chr.7: 139341749-139341749 on Build GRCh38
rs546646905 Chr.7: 139341470-139341470 on Build GRCh38
rs967688009 Chr.7: 139341767-139341767 on Build GRCh38
rs759231210 Chr.7: 139341452-139341452 on Build GRCh38
rs763234210 Chr.7: 139341480-139341480 on Build GRCh38
rs573544149 Chr.7: 139341688-139341688 on Build GRCh38
rs775020710 Chr.7: 139341531-139341531 on Build GRCh38
rs773788281 Chr.7: 139341556-139341556 on Build GRCh38
rs781063369 Chr.7: 139341526-139341526 on Build GRCh38
rs746984637 Chr.7: 139341520-139341520 on Build GRCh38
rs6947215 Chr.7: 139341892-139341892 on Build GRCh38
rs1021113768 Chr.7: 139341630-139341630 on Build GRCh38
rs146144422 Chr.7: 139341503-139341503 on Build GRCh38
rs769050200 Chr.7: 139341467-139341467 on Build GRCh38
rs1050919943 Chr.7: 139341662-139341662 on Build GRCh38
rs544723571 Chr.7: 139341721-139341721 on Build GRCh38
rs897822560 Chr.7: 139341704-139341704 on Build GRCh38
rs913537995 Chr.7: 139341591-139341591 on Build GRCh38
rs6965212 Chr.7: 139341719-139341719 on Build GRCh38
rs766790260 Chr.7: 139341812-139341812 on Build GRCh38
rs544455569 Chr.7: 139341490-139341490 on Build GRCh38
rs546373889 Chr.7: 139341608-139341608 on Build GRCh38
rs1008896773 Chr.7: 139341672-139341672 on Build GRCh38
rs890557520 Chr.7: 139341665-139341665 on Build GRCh38
rs552649389 Chr.7: 139341717-139341717 on Build GRCh38
rs369452273 Chr.7: 139341459-139341459 on Build GRCh38
rs542379332 Chr.7: 139341837-139341837 on Build GRCh38
rs768169920 Chr.7: 139341552-139341552 on Build GRCh38
rs754031356 Chr.7: 139341511-139341511 on Build GRCh38
rs759713020 Chr.7: 139341566-139341566 on Build GRCh38
rs977152425 Chr.7: 139341645-139341645 on Build GRCh38
rs748958951 Chr.7: 139341535-139341535 on Build GRCh38
rs774826732 Chr.7: 139341548-139341548 on Build GRCh38
rs967559849 Chr.7: 139341640-139341640 on Build GRCh38
rs1003686202 Chr.7: 139341769-139341769 on Build GRCh38
rs111891482 Chr.7: 139341718-139341719 on Build GRCh38
rs1049027286 Chr.7: 139341894-139341894 on Build GRCh38
rs912359790 Chr.7: 139341824-139341824 on Build GRCh38
rs747721447 Chr.7: 139341690-139341690 on Build GRCh38
rs762459972 Chr.7: 139341550-139341550 on Build GRCh38
rs1011142709 Chr.7: 139341756-139341756 on Build GRCh38
rs1021563032 Chr.7: 139341758-139341758 on Build GRCh38
rs915816838 Chr.7: 139341699-139341699 on Build GRCh38
rs759591054 Chr.7: 139341716-139341717 on Build GRCh38
rs777537487 Chr.7: 139341514-139341514 on Build GRCh38
rs938343020 Chr.7: 139341670-139341670 on Build GRCh38
rs980567869 Chr.7: 139341506-139341506 on Build GRCh38
rs915572637 Chr.7: 139341823-139341823 on Build GRCh38
rs372855450 Chr.7: 139341717-139341717 on Build GRCh38
rs908426083 Chr.7: 139341864-139341864 on Build GRCh38
rs775171261 Chr.7: 139341458-139341458 on Build GRCh38
rs769358474 Chr.7: 139341456-139341456 on Build GRCh38
rs930624017 Chr.7: 139341893-139341893 on Build GRCh38
rs116523410 Chr.7: 139341797-139341797 on Build GRCh38
rs987040396 Chr.7: 139341821-139341821 on Build GRCh38
rs901231476 Chr.7: 139341723-139341723 on Build GRCh38
rs776281314 Chr.7: 139341575-139341575 on Build GRCh38
rs952768248 Chr.7: 139341848-139341848 on Build GRCh38
rs894082054 Chr.7: 139341750-139341750 on Build GRCh38
rs575856755 Chr.7: 139341606-139341606 on Build GRCh38
rs75799520 Chr.7: 139341629-139341629 on Build GRCh38
rs6965143 Chr.7: 139341862-139341862 on Build GRCh38
rs996805783 Chr.7: 139341724-139341724 on Build GRCh38
rs1025546188 Chr.7: 139341799-139341799 on Build GRCh38
rs771699835 Chr.7: 139341523-139341523 on Build GRCh38
rs766513651 Chr.7: 139341510-139341510 on Build GRCh38
rs371664261 Chr.7: 139341601-139341601 on Build GRCh38
rs905460122 Chr.7: 139341896-139341896 on Build GRCh38
rs555238900 Chr.7: 139341686-139341686 on Build GRCh38
rs765410748 Chr.7: 139341791-139341791 on Build GRCh38
rs760682035 Chr.7: 139341574-139341574 on Build GRCh38
rs776807965 Chr.7: 139341556-139341556 on Build GRCh38
rs991192469 Chr.7: 139341820-139341820 on Build GRCh38
rs1028320332 Chr.7: 139341740-139341740 on Build GRCh38
rs764463286 Chr.7: 139341482-139341482 on Build GRCh38
rs769211858 Chr.7: 139341545-139341545 on Build GRCh38
rs769410289 Chr.7: 139341718-139341718 on Build GRCh38

View Full Product Details