Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140403902-140404427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB19
Entrez Gene ID
401409
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
RAB19B
UniGene
Hs.583545
Gene Chromosome Location
Chr.7: 140403740-140426881 on Build GRCh38
Transcripts
NM_001008749.2 XM_005249996.3 XM_005249997.3 XM_005249998.3 XM_011516626.2 XM_011516627.2 XM_017012712.1 XM_017012713.1 XM_017012714.1 XM_017012715.1 XM_017012716.1 XM_017012717.1 XM_017012718.1

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
-
UniGene
Hs.446021
Gene Chromosome Location
Chr.7: 140333752-140404352 on Build GRCh38
Transcripts
NM_001008749.2 XM_005249996.3 XM_005249997.3 XM_005249998.3 XM_011516626.2 XM_011516627.2 XM_017012712.1 XM_017012713.1 XM_017012714.1 XM_017012715.1 XM_017012716.1 XM_017012717.1 XM_017012718.1

SNP Detail

SNP ID Chromosome Location
rs961197484 Chr.7: 140404217-140404217 on Build GRCh38
rs764317844 Chr.7: 140404238-140404238 on Build GRCh38
rs950373655 Chr.7: 140404264-140404264 on Build GRCh38
rs747381609 Chr.7: 140403955-140403964 on Build GRCh38
rs765270515 Chr.7: 140403964-140403964 on Build GRCh38
rs73163251 Chr.7: 140404262-140404262 on Build GRCh38
rs925916927 Chr.7: 140404107-140404107 on Build GRCh38
rs888201851 Chr.7: 140404364-140404365 on Build GRCh38
rs987221537 Chr.7: 140404020-140404020 on Build GRCh38
rs750159895 Chr.7: 140404141-140404141 on Build GRCh38
rs5887976 Chr.7: 140403964-140403965 on Build GRCh38
rs936658086 Chr.7: 140404135-140404135 on Build GRCh38
rs1053828807 Chr.7: 140404144-140404144 on Build GRCh38
rs925858498 Chr.7: 140404337-140404337 on Build GRCh38
rs906687043 Chr.7: 140404193-140404193 on Build GRCh38
rs757138066 Chr.7: 140404053-140404053 on Build GRCh38
rs943198685 Chr.7: 140404057-140404057 on Build GRCh38
rs932669294 Chr.7: 140404224-140404224 on Build GRCh38
rs1014418113 Chr.7: 140404031-140404031 on Build GRCh38
rs763184572 Chr.7: 140404103-140404103 on Build GRCh38
rs575593386 Chr.7: 140404270-140404270 on Build GRCh38
rs1046432715 Chr.7: 140404277-140404277 on Build GRCh38
rs532635108 Chr.7: 140404376-140404376 on Build GRCh38
rs908336978 Chr.7: 140404183-140404183 on Build GRCh38
rs184327164 Chr.7: 140403927-140403927 on Build GRCh38
rs569622887 Chr.7: 140403997-140403997 on Build GRCh38
rs573169850 Chr.7: 140404322-140404322 on Build GRCh38
rs987192018 Chr.7: 140404293-140404293 on Build GRCh38
rs952579069 Chr.7: 140404282-140404282 on Build GRCh38
rs1009688130 Chr.7: 140403935-140403935 on Build GRCh38
rs1041054689 Chr.7: 140403937-140403937 on Build GRCh38
rs530682996 Chr.7: 140404399-140404399 on Build GRCh38
rs5887975 Chr.7: 140403947-140403948 on Build GRCh38
rs577766624 Chr.7: 140404178-140404178 on Build GRCh38
rs189611337 Chr.7: 140403947-140403947 on Build GRCh38
rs145091502 Chr.7: 140404263-140404263 on Build GRCh38
rs985456185 Chr.7: 140404253-140404253 on Build GRCh38
rs1002398296 Chr.7: 140404205-140404205 on Build GRCh38
rs536693078 Chr.7: 140404019-140404019 on Build GRCh38
rs1018725780 Chr.7: 140404296-140404296 on Build GRCh38
rs758211449 Chr.7: 140404243-140404243 on Build GRCh38
rs935892490 Chr.7: 140404356-140404356 on Build GRCh38
rs553175452 Chr.7: 140404174-140404174 on Build GRCh38
rs899449620 Chr.7: 140404260-140404260 on Build GRCh38
rs561233774 Chr.7: 140404317-140404317 on Build GRCh38
rs565156595 Chr.7: 140403983-140403983 on Build GRCh38
rs565094852 Chr.7: 140404365-140404365 on Build GRCh38
rs73163252 Chr.7: 140404332-140404332 on Build GRCh38
rs199865308 Chr.7: 140403952-140403952 on Build GRCh38
rs542687377 Chr.7: 140404309-140404309 on Build GRCh38
rs181191478 Chr.7: 140403987-140403987 on Build GRCh38
rs78738033 Chr.7: 140403986-140403986 on Build GRCh38
rs779640096 Chr.7: 140404261-140404261 on Build GRCh38
rs1025059469 Chr.7: 140403938-140403938 on Build GRCh38
rs201498079 Chr.7: 140403969-140403970 on Build GRCh38
rs528642787 Chr.7: 140403954-140403954 on Build GRCh38
rs977110758 Chr.7: 140404222-140404222 on Build GRCh38
rs397963116 Chr.7: 140403951-140403952 on Build GRCh38
rs567599834 Chr.7: 140404148-140404148 on Build GRCh38
rs117586496 Chr.7: 140404166-140404166 on Build GRCh38
rs543299523 Chr.7: 140403942-140403942 on Build GRCh38
rs376384506 Chr.7: 140404080-140404080 on Build GRCh38
rs5021205 Chr.7: 140403956-140403956 on Build GRCh38

View Full Product Details