Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140696236-140696764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFB2
Entrez Gene ID
4708
Gene Name
NADH:ubiquinone oxidoreductase subunit B2
Gene Aliases
AGGG, CI-AGGG
UniGene
Hs.655788
Gene Chromosome Location
Chr.7: 140696681-140706646 on Build GRCh38
Transcripts
NM_004546.2 NR_024454.1

Gene Symbol
NDUFB2-AS1
Entrez Gene ID
100134713
Gene Name
NDUFB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.534141
Gene Chromosome Location
Chr.7: 140695336-140697077 on Build GRCh38
Transcripts
NM_004546.2 NR_024454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397331 c.16C>T p.R6W Chr.7: 140696760-140696760 on Build GRCh38
4601913 c.7G>T p.A3S Chr.7: 140696751-140696751 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs983975801 Chr.7: 140696458-140696458 on Build GRCh38
rs578075508 Chr.7: 140696547-140696547 on Build GRCh38
rs1041263120 Chr.7: 140696489-140696489 on Build GRCh38
rs925221853 Chr.7: 140696565-140696566 on Build GRCh38
rs528674067 Chr.7: 140696646-140696646 on Build GRCh38
rs1011601002 Chr.7: 140696592-140696592 on Build GRCh38
rs368646482 Chr.7: 140696463-140696463 on Build GRCh38
rs200505172 Chr.7: 140696731-140696731 on Build GRCh38
rs181108921 Chr.7: 140696684-140696684 on Build GRCh38
rs921560406 Chr.7: 140696527-140696527 on Build GRCh38
rs554687940 Chr.7: 140696695-140696695 on Build GRCh38
rs1027224375 Chr.7: 140696638-140696638 on Build GRCh38
rs952388917 Chr.7: 140696427-140696427 on Build GRCh38
rs780048068 Chr.7: 140696595-140696595 on Build GRCh38
rs565396803 Chr.7: 140696685-140696685 on Build GRCh38
rs999409016 Chr.7: 140696294-140696294 on Build GRCh38
rs531262766 Chr.7: 140696612-140696612 on Build GRCh38
rs1045128275 Chr.7: 140696622-140696622 on Build GRCh38
rs1028782085 Chr.7: 140696727-140696727 on Build GRCh38
rs908486233 Chr.7: 140696465-140696465 on Build GRCh38
rs967513468 Chr.7: 140696289-140696289 on Build GRCh38
rs939964745 Chr.7: 140696479-140696479 on Build GRCh38
rs559885417 Chr.7: 140696545-140696545 on Build GRCh38
rs767268673 Chr.7: 140696516-140696516 on Build GRCh38
rs889090420 Chr.7: 140696578-140696578 on Build GRCh38
rs761216905 Chr.7: 140696334-140696334 on Build GRCh38
rs975827102 Chr.7: 140696725-140696725 on Build GRCh38
rs188286824 Chr.7: 140696560-140696560 on Build GRCh38
rs983733927 Chr.7: 140696658-140696658 on Build GRCh38
rs999151359 Chr.7: 140696623-140696623 on Build GRCh38
rs887379260 Chr.7: 140696644-140696644 on Build GRCh38
rs775799734 Chr.7: 140696735-140696735 on Build GRCh38
rs565848472 Chr.7: 140696641-140696641 on Build GRCh38
rs770543489 Chr.7: 140696722-140696722 on Build GRCh38
rs915537857 Chr.7: 140696329-140696329 on Build GRCh38
rs1027687386 Chr.7: 140696330-140696330 on Build GRCh38
rs983383317 Chr.7: 140696455-140696455 on Build GRCh38
rs553888021 Chr.7: 140696327-140696327 on Build GRCh38
rs752478738 Chr.7: 140696735-140696736 on Build GRCh38
rs949435087 Chr.7: 140696619-140696619 on Build GRCh38
rs539062761 Chr.7: 140696688-140696688 on Build GRCh38
rs78640923 Chr.7: 140696409-140696409 on Build GRCh38
rs1021897022 Chr.7: 140696268-140696268 on Build GRCh38
rs952359412 Chr.7: 140696653-140696653 on Build GRCh38
rs935162206 Chr.7: 140696593-140696594 on Build GRCh38
rs746876697 Chr.7: 140696718-140696718 on Build GRCh38
rs979366178 Chr.7: 140696555-140696555 on Build GRCh38
rs561569966 Chr.7: 140696631-140696631 on Build GRCh38
rs922496808 Chr.7: 140696496-140696496 on Build GRCh38
rs931680749 Chr.7: 140696535-140696535 on Build GRCh38
rs748122801 Chr.7: 140696701-140696701 on Build GRCh38
rs370927500 Chr.7: 140696409-140696409 on Build GRCh38
rs2930314 Chr.7: 140696706-140696706 on Build GRCh38
rs796423279 Chr.7: 140696410-140696412 on Build GRCh38
rs563990803 Chr.7: 140696571-140696571 on Build GRCh38
rs1013047097 Chr.7: 140696642-140696642 on Build GRCh38
rs776316983 Chr.7: 140696726-140696726 on Build GRCh38
rs61366549 Chr.7: 140696427-140696427 on Build GRCh38
rs751094137 Chr.7: 140696259-140696259 on Build GRCh38
rs763498668 Chr.7: 140696741-140696741 on Build GRCh38
rs908245431 Chr.7: 140696660-140696660 on Build GRCh38
rs1031701754 Chr.7: 140696428-140696428 on Build GRCh38
rs1036265662 Chr.7: 140696630-140696630 on Build GRCh38
rs903420482 Chr.7: 140696603-140696603 on Build GRCh38
rs879757902 Chr.7: 140696403-140696403 on Build GRCh38
rs1014662900 Chr.7: 140696466-140696466 on Build GRCh38
rs933400224 Chr.7: 140696505-140696505 on Build GRCh38
rs57717083 Chr.7: 140696672-140696673 on Build GRCh38
rs16882339 Chr.7: 140696702-140696702 on Build GRCh38
rs865818264 Chr.7: 140696491-140696491 on Build GRCh38
rs397689769 Chr.7: 140696675-140696676 on Build GRCh38
rs765265990 Chr.7: 140696703-140696703 on Build GRCh38
rs549394565 Chr.7: 140696627-140696627 on Build GRCh38
rs1035889404 Chr.7: 140696300-140696300 on Build GRCh38
rs758231214 Chr.7: 140696335-140696335 on Build GRCh38
rs968165915 Chr.7: 140696305-140696305 on Build GRCh38
rs1023565476 Chr.7: 140696296-140696296 on Build GRCh38
rs960050682 Chr.7: 140696306-140696306 on Build GRCh38

View Full Product Details