Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1565605-1566117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP2
Entrez Gene ID
9228
Gene Name
DLG associated protein 2
Gene Aliases
DAP2, SAPAP2
UniGene
Hs.113287
Gene Chromosome Location
Chr.8: 1501366-1708476 on Build GRCh38
Transcripts
NM_001277161.1 NM_004745.4 NR_103863.1

Gene Symbol
DLGAP2-AS1
Entrez Gene ID
100507435
Gene Name
DLGAP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.667133
Gene Chromosome Location
Chr.8: 1565509-1621664 on Build GRCh38
Transcripts
NM_001277161.1 NM_004745.4 NR_103863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3898400 c.1134G>C p.G378G Chr.8: 1565760-1565760 on Build GRCh38
5910619 c.1234G>A p.G412R Chr.8: 1565794-1565794 on Build GRCh38
4677859 c.1185G>C p.R395S Chr.8: 1565745-1565745 on Build GRCh38
3646724 c.1187G>A p.R396K Chr.8: 1565747-1565747 on Build GRCh38
1203740 c.1139A>G p.Y380C Chr.8: 1565765-1565765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749460084 Chr.8: 1565814-1565814 on Build GRCh38
rs762157201 Chr.8: 1566032-1566032 on Build GRCh38
rs918658352 Chr.8: 1565820-1565820 on Build GRCh38
rs953692864 Chr.8: 1565836-1565836 on Build GRCh38
rs558830103 Chr.8: 1565819-1565819 on Build GRCh38
rs553712210 Chr.8: 1565840-1565840 on Build GRCh38
rs777504934 Chr.8: 1565944-1565944 on Build GRCh38
rs1021177216 Chr.8: 1565782-1565782 on Build GRCh38
rs1055013818 Chr.8: 1565921-1565921 on Build GRCh38
rs3815235 Chr.8: 1565648-1565648 on Build GRCh38
rs756698008 Chr.8: 1565849-1565849 on Build GRCh38
rs781601177 Chr.8: 1565799-1565799 on Build GRCh38
rs755669113 Chr.8: 1565749-1565749 on Build GRCh38
rs1002194195 Chr.8: 1565660-1565660 on Build GRCh38
rs769019798 Chr.8: 1565816-1565816 on Build GRCh38
rs767385484 Chr.8: 1565730-1565730 on Build GRCh38
rs184775948 Chr.8: 1566028-1566028 on Build GRCh38
rs372579035 Chr.8: 1565887-1565887 on Build GRCh38
rs780692996 Chr.8: 1565669-1565669 on Build GRCh38
rs368597899 Chr.8: 1565676-1565676 on Build GRCh38
rs965097973 Chr.8: 1566036-1566036 on Build GRCh38
rs540625837 Chr.8: 1565829-1565829 on Build GRCh38
rs902328993 Chr.8: 1565964-1565964 on Build GRCh38
rs763133160 Chr.8: 1565850-1565850 on Build GRCh38
rs756646716 Chr.8: 1565911-1565911 on Build GRCh38
rs368905053 Chr.8: 1565945-1565945 on Build GRCh38
rs748250935 Chr.8: 1565940-1565940 on Build GRCh38
rs775916633 Chr.8: 1565710-1565710 on Build GRCh38
rs761552008 Chr.8: 1565822-1565822 on Build GRCh38
rs780743371 Chr.8: 1565912-1565912 on Build GRCh38
rs916531660 Chr.8: 1565968-1565968 on Build GRCh38
rs747102860 Chr.8: 1565909-1565909 on Build GRCh38
rs754587148 Chr.8: 1565833-1565833 on Build GRCh38
rs375184000 Chr.8: 1565793-1565793 on Build GRCh38
rs545834007 Chr.8: 1566048-1566048 on Build GRCh38
rs758569109 Chr.8: 1565727-1565727 on Build GRCh38
rs759098418 Chr.8: 1565880-1565880 on Build GRCh38
rs773593481 Chr.8: 1566044-1566044 on Build GRCh38
rs371842482 Chr.8: 1565784-1565784 on Build GRCh38
rs548099226 Chr.8: 1565965-1565965 on Build GRCh38
rs935187068 Chr.8: 1566022-1566022 on Build GRCh38
rs778152747 Chr.8: 1565790-1565790 on Build GRCh38
rs2301964 Chr.8: 1566004-1566004 on Build GRCh38
rs567377471 Chr.8: 1565638-1565638 on Build GRCh38
rs538849643 Chr.8: 1566051-1566051 on Build GRCh38
rs377204953 Chr.8: 1565665-1565665 on Build GRCh38
rs770069125 Chr.8: 1565810-1565810 on Build GRCh38
rs754638338 Chr.8: 1565785-1565785 on Build GRCh38
rs764839301 Chr.8: 1565842-1565842 on Build GRCh38
rs562623267 Chr.8: 1565848-1565848 on Build GRCh38
rs779190281 Chr.8: 1565939-1565939 on Build GRCh38
rs757624140 Chr.8: 1565844-1565844 on Build GRCh38
rs180753029 Chr.8: 1565658-1565658 on Build GRCh38
rs775353108 Chr.8: 1565867-1565867 on Build GRCh38
rs984884357 Chr.8: 1566086-1566086 on Build GRCh38
rs746307204 Chr.8: 1565847-1565847 on Build GRCh38
rs377473670 Chr.8: 1565642-1565642 on Build GRCh38
rs542762184 Chr.8: 1565846-1565846 on Build GRCh38
rs367568530 Chr.8: 1565716-1565716 on Build GRCh38
rs776089084 Chr.8: 1565877-1565877 on Build GRCh38
rs762079099 Chr.8: 1566000-1566000 on Build GRCh38
rs377735485 Chr.8: 1565671-1565671 on Build GRCh38
rs972779366 Chr.8: 1565766-1565766 on Build GRCh38
rs950946934 Chr.8: 1566075-1566075 on Build GRCh38
rs947982715 Chr.8: 1565976-1565976 on Build GRCh38
rs778116892 Chr.8: 1565685-1565685 on Build GRCh38
rs569893378 Chr.8: 1565664-1565664 on Build GRCh38
rs896497121 Chr.8: 1565926-1565926 on Build GRCh38
rs745977771 Chr.8: 1565708-1565708 on Build GRCh38
rs767712714 Chr.8: 1565894-1565894 on Build GRCh38
rs762893654 Chr.8: 1565657-1565657 on Build GRCh38
rs764931897 Chr.8: 1565884-1565884 on Build GRCh38
rs773757160 Chr.8: 1565635-1565635 on Build GRCh38
rs370722015 Chr.8: 1565673-1565673 on Build GRCh38
rs954336931 Chr.8: 1565920-1565920 on Build GRCh38
rs565976341 Chr.8: 1566090-1566090 on Build GRCh38
rs1001002371 Chr.8: 1565812-1565812 on Build GRCh38
rs753944157 Chr.8: 1565916-1565916 on Build GRCh38
rs545241585 Chr.8: 1565866-1565866 on Build GRCh38
rs753369744 Chr.8: 1565832-1565832 on Build GRCh38
rs549937034 Chr.8: 1566023-1566023 on Build GRCh38
rs199886485 Chr.8: 1565942-1565942 on Build GRCh38
rs757755202 Chr.8: 1565794-1565794 on Build GRCh38
rs773081201 Chr.8: 1565824-1565824 on Build GRCh38
rs770888838 Chr.8: 1565876-1565876 on Build GRCh38
rs771365827 Chr.8: 1565699-1565699 on Build GRCh38
rs116119098 Chr.8: 1565823-1565823 on Build GRCh38
rs368098115 Chr.8: 1565807-1565807 on Build GRCh38
rs371150576 Chr.8: 1565709-1565709 on Build GRCh38
rs1032181645 Chr.8: 1565969-1565969 on Build GRCh38
rs375315939 Chr.8: 1565675-1565675 on Build GRCh38
rs747493177 Chr.8: 1565694-1565694 on Build GRCh38
rs918709248 Chr.8: 1565962-1565962 on Build GRCh38
rs1046334437 Chr.8: 1565988-1565988 on Build GRCh38
rs868461092 Chr.8: 1565731-1565731 on Build GRCh38
rs188379340 Chr.8: 1566061-1566061 on Build GRCh38
rs761757961 Chr.8: 1565726-1565726 on Build GRCh38
rs895078018 Chr.8: 1565835-1565835 on Build GRCh38
rs775712545 Chr.8: 1565811-1565811 on Build GRCh38
rs200888342 Chr.8: 1565919-1565919 on Build GRCh38
rs772537326 Chr.8: 1565634-1565634 on Build GRCh38
rs984158602 Chr.8: 1565966-1565966 on Build GRCh38
rs749915594 Chr.8: 1565733-1565733 on Build GRCh38
rs762675179 Chr.8: 1565889-1565889 on Build GRCh38
rs372181339 Chr.8: 1566054-1566054 on Build GRCh38
rs779051609 Chr.8: 1565854-1565854 on Build GRCh38
rs765818847 Chr.8: 1565831-1565831 on Build GRCh38
rs2301963 Chr.8: 1565843-1565843 on Build GRCh38
rs776735846 Chr.8: 1565706-1565706 on Build GRCh38
rs972878795 Chr.8: 1565956-1565956 on Build GRCh38
rs374538901 Chr.8: 1565715-1565715 on Build GRCh38
rs768652516 Chr.8: 1565918-1565918 on Build GRCh38
rs117510071 Chr.8: 1565986-1565986 on Build GRCh38
rs909392083 Chr.8: 1566091-1566091 on Build GRCh38
rs889507962 Chr.8: 1565972-1565972 on Build GRCh38
rs765589691 Chr.8: 1565656-1565656 on Build GRCh38
rs201868215 Chr.8: 1565865-1565865 on Build GRCh38
rs747961521 Chr.8: 1565863-1565863 on Build GRCh38

View Full Product Details