Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 55523203-55523706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XKR4
Entrez Gene ID
114786
Gene Name
XK related 4
Gene Aliases
XRG4
UniGene
Hs.130197
Gene Chromosome Location
Chr.8: 55102457-55526155 on Build GRCh38
Transcripts
NM_052898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5007361 c.1198T>C p.Y400H Chr.8: 55523472-55523472 on Build GRCh38
5821409 c.1394T>G p.L465R Chr.8: 55523668-55523668 on Build GRCh38
230150 c.1399G>A p.E467K Chr.8: 55523673-55523673 on Build GRCh38
5044344 c.1270_1271TG>AA p.C424N Chr.8: 55523544-55523545 on Build GRCh38
3951733 c.1263A>G p.T421T Chr.8: 55523537-55523537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144592369 Chr.8: 55523393-55523393 on Build GRCh38
rs766227461 Chr.8: 55523574-55523574 on Build GRCh38
rs550251863 Chr.8: 55523253-55523253 on Build GRCh38
rs567768011 Chr.8: 55523678-55523678 on Build GRCh38
rs771157343 Chr.8: 55523230-55523230 on Build GRCh38
rs769582378 Chr.8: 55523540-55523540 on Build GRCh38
rs368185066 Chr.8: 55523282-55523282 on Build GRCh38
rs776073591 Chr.8: 55523517-55523517 on Build GRCh38
rs7839072 Chr.8: 55523336-55523336 on Build GRCh38
rs750598777 Chr.8: 55523636-55523636 on Build GRCh38
rs200138646 Chr.8: 55523683-55523683 on Build GRCh38
rs781681328 Chr.8: 55523664-55523664 on Build GRCh38
rs780322724 Chr.8: 55523650-55523650 on Build GRCh38
rs940206853 Chr.8: 55523225-55523226 on Build GRCh38
rs753881169 Chr.8: 55523596-55523596 on Build GRCh38
rs267601955 Chr.8: 55523525-55523525 on Build GRCh38
rs747519190 Chr.8: 55523489-55523489 on Build GRCh38
rs772390690 Chr.8: 55523481-55523481 on Build GRCh38
rs868239659 Chr.8: 55523486-55523486 on Build GRCh38
rs149121707 Chr.8: 55523594-55523594 on Build GRCh38
rs761412913 Chr.8: 55523391-55523391 on Build GRCh38
rs905944447 Chr.8: 55523353-55523353 on Build GRCh38
rs1031467095 Chr.8: 55523606-55523606 on Build GRCh38
rs373647980 Chr.8: 55523364-55523364 on Build GRCh38
rs774733517 Chr.8: 55523348-55523348 on Build GRCh38
rs775795877 Chr.8: 55523360-55523360 on Build GRCh38
rs769103038 Chr.8: 55523661-55523661 on Build GRCh38
rs1018855468 Chr.8: 55523441-55523441 on Build GRCh38
rs768003479 Chr.8: 55523379-55523379 on Build GRCh38
rs751411350 Chr.8: 55523427-55523427 on Build GRCh38
rs758597574 Chr.8: 55523643-55523643 on Build GRCh38
rs4263738 Chr.8: 55523315-55523315 on Build GRCh38
rs763814319 Chr.8: 55523269-55523269 on Build GRCh38
rs767977383 Chr.8: 55523256-55523256 on Build GRCh38
rs140731209 Chr.8: 55523356-55523356 on Build GRCh38
rs769175965 Chr.8: 55523490-55523490 on Build GRCh38
rs147261528 Chr.8: 55523291-55523291 on Build GRCh38
rs753395129 Chr.8: 55523276-55523276 on Build GRCh38
rs374111882 Chr.8: 55523294-55523294 on Build GRCh38
rs776185325 Chr.8: 55523389-55523389 on Build GRCh38
rs772584686 Chr.8: 55523357-55523357 on Build GRCh38
rs774859783 Chr.8: 55523250-55523250 on Build GRCh38
rs757539151 Chr.8: 55523471-55523471 on Build GRCh38
rs750301125 Chr.8: 55523409-55523409 on Build GRCh38
rs777760757 Chr.8: 55523468-55523468 on Build GRCh38
rs753921591 Chr.8: 55523469-55523469 on Build GRCh38
rs779053061 Chr.8: 55523477-55523477 on Build GRCh38
rs867658481 Chr.8: 55523666-55523666 on Build GRCh38
rs368680085 Chr.8: 55523576-55523576 on Build GRCh38
rs749418970 Chr.8: 55523343-55523343 on Build GRCh38
rs147871706 Chr.8: 55523547-55523547 on Build GRCh38
rs1011655361 Chr.8: 55523665-55523665 on Build GRCh38
rs747397907 Chr.8: 55523660-55523660 on Build GRCh38
rs762778438 Chr.8: 55523417-55523417 on Build GRCh38
rs772606173 Chr.8: 55523505-55523505 on Build GRCh38
rs757263321 Chr.8: 55523631-55523631 on Build GRCh38
rs748566342 Chr.8: 55523424-55523424 on Build GRCh38
rs761395206 Chr.8: 55523523-55523523 on Build GRCh38
rs771115839 Chr.8: 55523347-55523347 on Build GRCh38
rs746238764 Chr.8: 55523480-55523480 on Build GRCh38
rs775043728 Chr.8: 55523263-55523263 on Build GRCh38
rs759429003 Chr.8: 55523583-55523583 on Build GRCh38
rs756236708 Chr.8: 55523450-55523450 on Build GRCh38
rs766421902 Chr.8: 55523420-55523420 on Build GRCh38
rs551126259 Chr.8: 55523426-55523426 on Build GRCh38
rs756080010 Chr.8: 55523317-55523317 on Build GRCh38
rs748272373 Chr.8: 55523304-55523304 on Build GRCh38
rs372636201 Chr.8: 55523634-55523634 on Build GRCh38
rs886369114 Chr.8: 55523388-55523388 on Build GRCh38
rs751343278 Chr.8: 55523580-55523580 on Build GRCh38
rs760266028 Chr.8: 55523373-55523373 on Build GRCh38
rs375137052 Chr.8: 55523264-55523264 on Build GRCh38
rs762871165 Chr.8: 55523552-55523552 on Build GRCh38
rs867767654 Chr.8: 55523355-55523355 on Build GRCh38
rs765175807 Chr.8: 55523290-55523290 on Build GRCh38

View Full Product Details