Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56113262-56113767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOS
Entrez Gene ID
4342
Gene Name
v-mos Moloney murine sarcoma viral oncogene homolog
Gene Aliases
MSV
UniGene
Hs.533432
Gene Chromosome Location
Chr.8: 56112942-56113982 on Build GRCh38
Transcripts
NM_005372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4702930 c.339C>T p.R113R Chr.8: 56113644-56113644 on Build GRCh38
6181249 c.553G>T p.V185F Chr.8: 56113430-56113430 on Build GRCh38
1635828 c.612C>A p.P204P Chr.8: 56113371-56113371 on Build GRCh38
6576935 c.629G>C p.S210T Chr.8: 56113354-56113354 on Build GRCh38
3728167 c.224A>G p.Y75C Chr.8: 56113759-56113759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761859165 Chr.8: 56113379-56113379 on Build GRCh38
rs751384600 Chr.8: 56113513-56113513 on Build GRCh38
rs369220995 Chr.8: 56113326-56113326 on Build GRCh38
rs757128773 Chr.8: 56113390-56113390 on Build GRCh38
rs749423112 Chr.8: 56113452-56113452 on Build GRCh38
rs943437335 Chr.8: 56113420-56113420 on Build GRCh38
rs757717531 Chr.8: 56113524-56113524 on Build GRCh38
rs748317252 Chr.8: 56113443-56113443 on Build GRCh38
rs758447455 Chr.8: 56113402-56113402 on Build GRCh38
rs763439301 Chr.8: 56113300-56113300 on Build GRCh38
rs529847429 Chr.8: 56113521-56113521 on Build GRCh38
rs763875775 Chr.8: 56113350-56113350 on Build GRCh38
rs750288981 Chr.8: 56113500-56113500 on Build GRCh38
rs759362843 Chr.8: 56113709-56113709 on Build GRCh38
rs747365374 Chr.8: 56113657-56113657 on Build GRCh38
rs774225819 Chr.8: 56113348-56113348 on Build GRCh38
rs779116609 Chr.8: 56113635-56113635 on Build GRCh38
rs185687051 Chr.8: 56113474-56113474 on Build GRCh38
rs766662365 Chr.8: 56113304-56113304 on Build GRCh38
rs142291866 Chr.8: 56113529-56113529 on Build GRCh38
rs752568092 Chr.8: 56113735-56113735 on Build GRCh38
rs756230428 Chr.8: 56113515-56113515 on Build GRCh38
rs777366450 Chr.8: 56113439-56113441 on Build GRCh38
rs776081262 Chr.8: 56113565-56113565 on Build GRCh38
rs200951438 Chr.8: 56113641-56113641 on Build GRCh38
rs1044569526 Chr.8: 56113648-56113648 on Build GRCh38
rs754914458 Chr.8: 56113622-56113622 on Build GRCh38
rs746035809 Chr.8: 56113640-56113640 on Build GRCh38
rs969068314 Chr.8: 56113733-56113733 on Build GRCh38
rs762844655 Chr.8: 56113597-56113597 on Build GRCh38
rs778799399 Chr.8: 56113394-56113394 on Build GRCh38
rs930795508 Chr.8: 56113387-56113387 on Build GRCh38
rs146390181 Chr.8: 56113575-56113575 on Build GRCh38
rs777179743 Chr.8: 56113673-56113673 on Build GRCh38
rs761515530 Chr.8: 56113496-56113496 on Build GRCh38
rs763458012 Chr.8: 56113480-56113480 on Build GRCh38
rs745827984 Chr.8: 56113395-56113395 on Build GRCh38
rs1048011369 Chr.8: 56113684-56113684 on Build GRCh38
rs769756297 Chr.8: 56113444-56113444 on Build GRCh38
rs928821421 Chr.8: 56113667-56113667 on Build GRCh38
rs753546945 Chr.8: 56113490-56113490 on Build GRCh38
rs914676231 Chr.8: 56113601-56113601 on Build GRCh38
rs766028889 Chr.8: 56113603-56113603 on Build GRCh38
rs986716211 Chr.8: 56113495-56113495 on Build GRCh38
rs572846115 Chr.8: 56113314-56113314 on Build GRCh38
rs990606886 Chr.8: 56113593-56113593 on Build GRCh38
rs372552034 Chr.8: 56113608-56113608 on Build GRCh38
rs376785210 Chr.8: 56113634-56113634 on Build GRCh38
rs753977054 Chr.8: 56113629-56113629 on Build GRCh38
rs889181910 Chr.8: 56113419-56113419 on Build GRCh38
rs772358722 Chr.8: 56113551-56113551 on Build GRCh38
rs561039574 Chr.8: 56113511-56113511 on Build GRCh38
rs780117512 Chr.8: 56113412-56113412 on Build GRCh38
rs781167645 Chr.8: 56113625-56113625 on Build GRCh38
rs368963717 Chr.8: 56113579-56113579 on Build GRCh38
rs35392772 Chr.8: 56113670-56113670 on Build GRCh38
rs577121918 Chr.8: 56113299-56113299 on Build GRCh38
rs774862139 Chr.8: 56113311-56113311 on Build GRCh38
rs750095075 Chr.8: 56113594-56113594 on Build GRCh38
rs749358950 Chr.8: 56113520-56113520 on Build GRCh38
rs760278351 Chr.8: 56113609-56113609 on Build GRCh38
rs773486880 Chr.8: 56113445-56113445 on Build GRCh38
rs758253167 Chr.8: 56113508-56113508 on Build GRCh38
rs764947261 Chr.8: 56113499-56113499 on Build GRCh38
rs552123421 Chr.8: 56113742-56113742 on Build GRCh38
rs767422321 Chr.8: 56113732-56113732 on Build GRCh38
rs956068265 Chr.8: 56113530-56113530 on Build GRCh38
rs962831190 Chr.8: 56113396-56113396 on Build GRCh38
rs373661675 Chr.8: 56113685-56113685 on Build GRCh38
rs370735077 Chr.8: 56113743-56113743 on Build GRCh38
rs761480996 Chr.8: 56113683-56113683 on Build GRCh38
rs376820512 Chr.8: 56113467-56113467 on Build GRCh38
rs546370933 Chr.8: 56113522-56113522 on Build GRCh38
rs919543361 Chr.8: 56113383-56113383 on Build GRCh38
rs757893643 Chr.8: 56113426-56113428 on Build GRCh38
rs372217648 Chr.8: 56113346-56113346 on Build GRCh38
rs761381034 Chr.8: 56113584-56113584 on Build GRCh38
rs752304134 Chr.8: 56113339-56113339 on Build GRCh38
rs751930901 Chr.8: 56113469-56113469 on Build GRCh38
rs767623793 Chr.8: 56113458-56113458 on Build GRCh38
rs34532635 Chr.8: 56113696-56113696 on Build GRCh38
rs776040946 Chr.8: 56113583-56113583 on Build GRCh38
rs765039256 Chr.8: 56113588-56113588 on Build GRCh38
rs777676899 Chr.8: 56113517-56113517 on Build GRCh38
rs753368455 Chr.8: 56113359-56113359 on Build GRCh38
rs564318673 Chr.8: 56113536-56113536 on Build GRCh38
rs972725148 Chr.8: 56113373-56113373 on Build GRCh38
rs774064952 Chr.8: 56113698-56113698 on Build GRCh38

View Full Product Details