Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60232158-60232666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA8
Entrez Gene ID
767
Gene Name
carbonic anhydrase 8
Gene Aliases
CA-RP, CA-VIII, CALS, CAMRQ3, CARP
UniGene
Hs.654388
Gene Chromosome Location
Chr.8: 60185420-60281423 on Build GRCh38
Transcripts
NM_001321837.1 NM_001321838.1 NM_001321839.1 NM_004056.5 NR_135821.1 XM_017013818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1132720 c.490G>A p.A164T Chr.8: 60232307-60232307 on Build GRCh38
1100755 c.494T>G p.I165S Chr.8: 60232303-60232303 on Build GRCh38
7207900 c.451G>T p.G151C Chr.8: 60232346-60232346 on Build GRCh38
5589971 c.479C>T p.P160L Chr.8: 60232318-60232318 on Build GRCh38
6013174 c.435G>A p.W145* Chr.8: 60232362-60232362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571931082 Chr.8: 60232562-60232562 on Build GRCh38
rs768293535 Chr.8: 60232308-60232308 on Build GRCh38
rs763154535 Chr.8: 60232246-60232246 on Build GRCh38
rs771677875 Chr.8: 60232282-60232282 on Build GRCh38
rs556165338 Chr.8: 60232318-60232318 on Build GRCh38
rs1045220497 Chr.8: 60232342-60232342 on Build GRCh38
rs749721295 Chr.8: 60232276-60232276 on Build GRCh38
rs752814536 Chr.8: 60232351-60232351 on Build GRCh38
rs780978826 Chr.8: 60232348-60232348 on Build GRCh38
rs537454715 Chr.8: 60232317-60232317 on Build GRCh38
rs767757358 Chr.8: 60232345-60232345 on Build GRCh38
rs770513940 Chr.8: 60232314-60232314 on Build GRCh38
rs954806449 Chr.8: 60232226-60232226 on Build GRCh38
rs140301131 Chr.8: 60232545-60232545 on Build GRCh38
rs538787010 Chr.8: 60232618-60232620 on Build GRCh38
rs751843637 Chr.8: 60232259-60232259 on Build GRCh38
rs974576578 Chr.8: 60232217-60232217 on Build GRCh38
rs11784742 Chr.8: 60232587-60232587 on Build GRCh38
rs757751935 Chr.8: 60232389-60232389 on Build GRCh38
rs749909756 Chr.8: 60232401-60232401 on Build GRCh38
rs766498562 Chr.8: 60232257-60232257 on Build GRCh38
rs963244956 Chr.8: 60232213-60232213 on Build GRCh38
rs773044449 Chr.8: 60232393-60232393 on Build GRCh38
rs769624801 Chr.8: 60232531-60232531 on Build GRCh38
rs79267946 Chr.8: 60232322-60232322 on Build GRCh38
rs969638329 Chr.8: 60232310-60232310 on Build GRCh38
rs113893454 Chr.8: 60232200-60232200 on Build GRCh38
rs778369328 Chr.8: 60232274-60232274 on Build GRCh38
rs746839542 Chr.8: 60232307-60232307 on Build GRCh38
rs954557275 Chr.8: 60232582-60232582 on Build GRCh38
rs151060395 Chr.8: 60232242-60232242 on Build GRCh38
rs538581020 Chr.8: 60232487-60232487 on Build GRCh38
rs775347412 Chr.8: 60232241-60232241 on Build GRCh38
rs776381649 Chr.8: 60232243-60232243 on Build GRCh38
rs956792758 Chr.8: 60232607-60232607 on Build GRCh38
rs1030917638 Chr.8: 60232596-60232596 on Build GRCh38
rs768508481 Chr.8: 60232427-60232427 on Build GRCh38
rs73683396 Chr.8: 60232231-60232231 on Build GRCh38
rs562027926 Chr.8: 60232260-60232260 on Build GRCh38
rs141808308 Chr.8: 60232368-60232368 on Build GRCh38
rs199747452 Chr.8: 60232261-60232261 on Build GRCh38
rs757851957 Chr.8: 60232420-60232420 on Build GRCh38
rs996911318 Chr.8: 60232542-60232542 on Build GRCh38
rs775194207 Chr.8: 60232289-60232289 on Build GRCh38
rs374622415 Chr.8: 60232278-60232278 on Build GRCh38
rs754359811 Chr.8: 60232384-60232384 on Build GRCh38
rs10102080 Chr.8: 60232237-60232237 on Build GRCh38
rs566313488 Chr.8: 60232219-60232219 on Build GRCh38
rs943899078 Chr.8: 60232212-60232212 on Build GRCh38
rs1056859593 Chr.8: 60232550-60232550 on Build GRCh38
rs745760343 Chr.8: 60232475-60232475 on Build GRCh38
rs148297638 Chr.8: 60232316-60232316 on Build GRCh38
rs114827865 Chr.8: 60232432-60232432 on Build GRCh38
rs965219361 Chr.8: 60232447-60232447 on Build GRCh38
rs761134546 Chr.8: 60232355-60232355 on Build GRCh38
rs1032341504 Chr.8: 60232397-60232397 on Build GRCh38
rs887140712 Chr.8: 60232446-60232446 on Build GRCh38
rs1028795533 Chr.8: 60232220-60232220 on Build GRCh38
rs368475479 Chr.8: 60232399-60232399 on Build GRCh38
rs899925398 Chr.8: 60232538-60232538 on Build GRCh38
rs542662241 Chr.8: 60232586-60232586 on Build GRCh38
rs756768339 Chr.8: 60232277-60232277 on Build GRCh38
rs779784677 Chr.8: 60232422-60232422 on Build GRCh38
rs746574538 Chr.8: 60232424-60232424 on Build GRCh38
rs975667090 Chr.8: 60232472-60232472 on Build GRCh38
rs758083149 Chr.8: 60232230-60232230 on Build GRCh38
rs989116300 Chr.8: 60232616-60232616 on Build GRCh38
rs576519781 Chr.8: 60232603-60232603 on Build GRCh38
rs1038340454 Chr.8: 60232500-60232500 on Build GRCh38
rs949545305 Chr.8: 60232598-60232598 on Build GRCh38
rs547214296 Chr.8: 60232315-60232315 on Build GRCh38
rs189339509 Chr.8: 60232181-60232181 on Build GRCh38

View Full Product Details