Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60591777-60592281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB2A
Entrez Gene ID
5862
Gene Name
RAB2A, member RAS oncogene family
Gene Aliases
LHX, RAB2
UniGene
Hs.369017
Gene Chromosome Location
Chr.8: 60516910-60623644 on Build GRCh38
Transcripts
NM_001242644.1 NM_002865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
343779 c.416G>A p.R139Q Chr.8: 60591911-60591911 on Build GRCh38
1457682 c.461C>T p.S154F Chr.8: 60591956-60591956 on Build GRCh38
1663793 c.377G>A p.R126K Chr.8: 60591872-60591872 on Build GRCh38
4843610 c.380G>A p.R127K Chr.8: 60591875-60591875 on Build GRCh38
1100759 c.392_394delAAG p.E133delE Chr.8: 60591887-60591889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759121999 Chr.8: 60591833-60591833 on Build GRCh38
rs757627796 Chr.8: 60592005-60592005 on Build GRCh38
rs373343553 Chr.8: 60592090-60592090 on Build GRCh38
rs1033755141 Chr.8: 60592044-60592044 on Build GRCh38
rs932312756 Chr.8: 60592130-60592130 on Build GRCh38
rs781427038 Chr.8: 60591811-60591811 on Build GRCh38
rs373117062 Chr.8: 60591974-60591974 on Build GRCh38
rs994127590 Chr.8: 60592009-60592009 on Build GRCh38
rs144969709 Chr.8: 60592160-60592161 on Build GRCh38
rs771550552 Chr.8: 60591826-60591826 on Build GRCh38
rs1016806103 Chr.8: 60591957-60591957 on Build GRCh38
rs375296362 Chr.8: 60591997-60591997 on Build GRCh38
rs200028335 Chr.8: 60591993-60591993 on Build GRCh38
rs932428801 Chr.8: 60591832-60591832 on Build GRCh38
rs1033638977 Chr.8: 60592259-60592259 on Build GRCh38
rs1049457057 Chr.8: 60591894-60591894 on Build GRCh38
rs567221003 Chr.8: 60592191-60592191 on Build GRCh38
rs530696032 Chr.8: 60592067-60592067 on Build GRCh38
rs765168370 Chr.8: 60591915-60591915 on Build GRCh38
rs775294429 Chr.8: 60591852-60591852 on Build GRCh38
rs370866047 Chr.8: 60591924-60591924 on Build GRCh38
rs767562111 Chr.8: 60591975-60591975 on Build GRCh38
rs1002199811 Chr.8: 60592240-60592240 on Build GRCh38
rs913698959 Chr.8: 60592088-60592088 on Build GRCh38
rs774941255 Chr.8: 60591830-60591830 on Build GRCh38
rs540804431 Chr.8: 60592217-60592217 on Build GRCh38
rs767732632 Chr.8: 60592035-60592035 on Build GRCh38
rs888206371 Chr.8: 60592159-60592159 on Build GRCh38
rs775400923 Chr.8: 60591996-60591996 on Build GRCh38
rs763831821 Chr.8: 60591880-60591880 on Build GRCh38
rs188810877 Chr.8: 60592205-60592205 on Build GRCh38
rs574446045 Chr.8: 60592229-60592229 on Build GRCh38
rs571157583 Chr.8: 60591900-60591900 on Build GRCh38
rs989328648 Chr.8: 60592065-60592065 on Build GRCh38
rs1037885960 Chr.8: 60592161-60592162 on Build GRCh38
rs73683524 Chr.8: 60592224-60592224 on Build GRCh38
rs568521272 Chr.8: 60592027-60592027 on Build GRCh38
rs778169475 Chr.8: 60591817-60591817 on Build GRCh38
rs770106859 Chr.8: 60591816-60591816 on Build GRCh38
rs569221654 Chr.8: 60592101-60592101 on Build GRCh38
rs146537724 Chr.8: 60592100-60592100 on Build GRCh38
rs750325171 Chr.8: 60591939-60591939 on Build GRCh38
rs141013917 Chr.8: 60591882-60591882 on Build GRCh38
rs779593461 Chr.8: 60592011-60592011 on Build GRCh38
rs377385072 Chr.8: 60591844-60591844 on Build GRCh38
rs1049506722 Chr.8: 60592137-60592137 on Build GRCh38
rs1035940738 Chr.8: 60592177-60592177 on Build GRCh38
rs752904410 Chr.8: 60591980-60591980 on Build GRCh38
rs866712222 Chr.8: 60592160-60592160 on Build GRCh38
rs535892012 Chr.8: 60592211-60592211 on Build GRCh38
rs199581780 Chr.8: 60591925-60591925 on Build GRCh38
rs769990701 Chr.8: 60592193-60592193 on Build GRCh38
rs779533840 Chr.8: 60592192-60592192 on Build GRCh38
rs970698576 Chr.8: 60592025-60592025 on Build GRCh38
rs897963652 Chr.8: 60592179-60592179 on Build GRCh38
rs558421884 Chr.8: 60592126-60592126 on Build GRCh38
rs373386761 Chr.8: 60591813-60591813 on Build GRCh38
rs756355395 Chr.8: 60591982-60591982 on Build GRCh38
rs761975340 Chr.8: 60591854-60591854 on Build GRCh38
rs750592323 Chr.8: 60592061-60592061 on Build GRCh38
rs892888458 Chr.8: 60592066-60592066 on Build GRCh38
rs115496123 Chr.8: 60592111-60592111 on Build GRCh38

View Full Product Details