Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60622414-60622937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB2A
Entrez Gene ID
5862
Gene Name
RAB2A, member RAS oncogene family
Gene Aliases
LHX, RAB2
UniGene
Hs.369017
Gene Chromosome Location
Chr.8: 60516910-60623644 on Build GRCh38
Transcripts
NM_001242644.1 NM_002865.2

SNP Detail

SNP ID Chromosome Location
rs967295395 Chr.8: 60622640-60622640 on Build GRCh38
rs907262777 Chr.8: 60622507-60622507 on Build GRCh38
rs538380311 Chr.8: 60622790-60622790 on Build GRCh38
rs79970848 Chr.8: 60622445-60622445 on Build GRCh38
rs946449948 Chr.8: 60622529-60622529 on Build GRCh38
rs987962204 Chr.8: 60622485-60622485 on Build GRCh38
rs531143485 Chr.8: 60622689-60622689 on Build GRCh38
rs965896176 Chr.8: 60622479-60622479 on Build GRCh38
rs889807638 Chr.8: 60622733-60622733 on Build GRCh38
rs941669427 Chr.8: 60622828-60622828 on Build GRCh38
rs552535426 Chr.8: 60622705-60622705 on Build GRCh38
rs1003384908 Chr.8: 60622528-60622528 on Build GRCh38
rs937674851 Chr.8: 60622621-60622621 on Build GRCh38
rs776727275 Chr.8: 60622484-60622484 on Build GRCh38
rs1037163637 Chr.8: 60622675-60622675 on Build GRCh38
rs759476655 Chr.8: 60622494-60622494 on Build GRCh38
rs75974140 Chr.8: 60622906-60622906 on Build GRCh38
rs979615744 Chr.8: 60622857-60622857 on Build GRCh38
rs958699870 Chr.8: 60622555-60622555 on Build GRCh38
rs923114435 Chr.8: 60622694-60622694 on Build GRCh38
rs548493450 Chr.8: 60622652-60622652 on Build GRCh38
rs954452048 Chr.8: 60622708-60622708 on Build GRCh38
rs75837686 Chr.8: 60622720-60622720 on Build GRCh38
rs913690146 Chr.8: 60622492-60622492 on Build GRCh38
rs986288322 Chr.8: 60622789-60622789 on Build GRCh38
rs1018907614 Chr.8: 60622741-60622741 on Build GRCh38
rs1008215663 Chr.8: 60622739-60622739 on Build GRCh38
rs996011014 Chr.8: 60622714-60622714 on Build GRCh38
rs118165570 Chr.8: 60622902-60622902 on Build GRCh38
rs138129268 Chr.8: 60622635-60622635 on Build GRCh38
rs988015864 Chr.8: 60622820-60622820 on Build GRCh38
rs649634 Chr.8: 60622566-60622566 on Build GRCh38
rs527452829 Chr.8: 60622598-60622598 on Build GRCh38
rs149557425 Chr.8: 60622854-60622854 on Build GRCh38
rs192628540 Chr.8: 60622662-60622662 on Build GRCh38
rs2930041 Chr.8: 60622808-60622808 on Build GRCh38
rs9437 Chr.8: 60622858-60622858 on Build GRCh38
rs752485713 Chr.8: 60622641-60622641 on Build GRCh38
rs1043448088 Chr.8: 60622557-60622557 on Build GRCh38
rs994326478 Chr.8: 60622437-60622437 on Build GRCh38
rs750639422 Chr.8: 60622852-60622852 on Build GRCh38

View Full Product Details