Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73065802-73066305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBSPON
Entrez Gene ID
157869
Gene Name
somatomedin B and thrombospondin type 1 domain containing
Gene Aliases
C8orf84, RPESP
UniGene
Hs.439040
Gene Chromosome Location
Chr.8: 73063077-73093272 on Build GRCh38
Transcripts
NM_153225.3 XM_017013145.1

SNP Detail

SNP ID Chromosome Location
rs529907509 Chr.8: 73066016-73066016 on Build GRCh38
rs1047428 Chr.8: 73066005-73066005 on Build GRCh38
rs79617270 Chr.8: 73065909-73065909 on Build GRCh38
rs895462400 Chr.8: 73066094-73066094 on Build GRCh38
rs1023830141 Chr.8: 73065859-73065860 on Build GRCh38
rs905360963 Chr.8: 73066140-73066140 on Build GRCh38
rs534187982 Chr.8: 73066144-73066144 on Build GRCh38
rs145422398 Chr.8: 73066027-73066027 on Build GRCh38
rs181851027 Chr.8: 73065958-73065958 on Build GRCh38
rs1010338116 Chr.8: 73065937-73065937 on Build GRCh38
rs995778422 Chr.8: 73065911-73065911 on Build GRCh38
rs965456103 Chr.8: 73066014-73066014 on Build GRCh38
rs532108868 Chr.8: 73065893-73065893 on Build GRCh38
rs1043128094 Chr.8: 73065939-73065939 on Build GRCh38
rs1004449138 Chr.8: 73066245-73066245 on Build GRCh38
rs369280492 Chr.8: 73066050-73066050 on Build GRCh38
rs951657702 Chr.8: 73065964-73065964 on Build GRCh38
rs924558191 Chr.8: 73066082-73066082 on Build GRCh38
rs748221854 Chr.8: 73066091-73066091 on Build GRCh38
rs1012654198 Chr.8: 73065848-73065849 on Build GRCh38
rs912549657 Chr.8: 73066004-73066004 on Build GRCh38
rs975445425 Chr.8: 73066017-73066017 on Build GRCh38
rs781282081 Chr.8: 73066089-73066089 on Build GRCh38
rs566687619 Chr.8: 73066102-73066102 on Build GRCh38
rs983220274 Chr.8: 73065992-73065992 on Build GRCh38
rs1027144373 Chr.8: 73066161-73066161 on Build GRCh38
rs1057113827 Chr.8: 73065899-73065899 on Build GRCh38
rs1042893914 Chr.8: 73066055-73066055 on Build GRCh38
rs7841807 Chr.8: 73066244-73066244 on Build GRCh38
rs771322227 Chr.8: 73065916-73065921 on Build GRCh38
rs947169965 Chr.8: 73066031-73066031 on Build GRCh38
rs927565479 Chr.8: 73065845-73065845 on Build GRCh38
rs1019591566 Chr.8: 73066283-73066283 on Build GRCh38
rs552336920 Chr.8: 73066149-73066149 on Build GRCh38
rs878926313 Chr.8: 73065965-73065968 on Build GRCh38
rs1044136199 Chr.8: 73066120-73066120 on Build GRCh38
rs1027608524 Chr.8: 73065941-73065941 on Build GRCh38
rs997617987 Chr.8: 73066143-73066143 on Build GRCh38
rs934583418 Chr.8: 73066083-73066083 on Build GRCh38

View Full Product Details