Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24466700-24467247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM7
Entrez Gene ID
8756
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
ADAM 7, ADAM-7, EAPI, GP-83, GP83
UniGene
Hs.116147
Gene Chromosome Location
Chr.8: 24440996-24509565 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5037462 c.534A>G p.K178K Chr.8: 24466943-24466943 on Build GRCh38
5612970 c.530G>T p.R177I Chr.8: 24466939-24466939 on Build GRCh38
4977740 c.408C>T p.N136N Chr.8: 24466817-24466817 on Build GRCh38
7354214 c.500A>C p.N167T Chr.8: 24466909-24466909 on Build GRCh38
142117 c.539T>C p.V180A Chr.8: 24466948-24466948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756537927 Chr.8: 24467062-24467062 on Build GRCh38
rs868821595 Chr.8: 24466945-24466945 on Build GRCh38
rs763965256 Chr.8: 24466897-24466897 on Build GRCh38
rs748550554 Chr.8: 24466953-24466953 on Build GRCh38
rs757940668 Chr.8: 24467079-24467079 on Build GRCh38
rs760465080 Chr.8: 24466894-24466894 on Build GRCh38
rs973716636 Chr.8: 24467142-24467142 on Build GRCh38
rs367766886 Chr.8: 24467218-24467218 on Build GRCh38
rs757498090 Chr.8: 24466809-24466809 on Build GRCh38
rs773959713 Chr.8: 24466877-24466877 on Build GRCh38
rs781475833 Chr.8: 24466853-24466853 on Build GRCh38
rs544931596 Chr.8: 24467169-24467170 on Build GRCh38
rs761113889 Chr.8: 24466954-24466954 on Build GRCh38
rs759628671 Chr.8: 24467027-24467027 on Build GRCh38
rs748959286 Chr.8: 24466863-24466863 on Build GRCh38
rs541450223 Chr.8: 24467041-24467041 on Build GRCh38
rs941088975 Chr.8: 24467222-24467222 on Build GRCh38
rs985912973 Chr.8: 24466763-24466763 on Build GRCh38
rs1013941195 Chr.8: 24467088-24467088 on Build GRCh38
rs750105683 Chr.8: 24467077-24467077 on Build GRCh38
rs183779862 Chr.8: 24467011-24467011 on Build GRCh38
rs949790205 Chr.8: 24467191-24467191 on Build GRCh38
rs574706805 Chr.8: 24467037-24467037 on Build GRCh38
rs17052018 Chr.8: 24467145-24467145 on Build GRCh38
rs768678285 Chr.8: 24466981-24466981 on Build GRCh38
rs996736034 Chr.8: 24467006-24467006 on Build GRCh38
rs762159774 Chr.8: 24466917-24466917 on Build GRCh38
rs755387849 Chr.8: 24466946-24466946 on Build GRCh38
rs139013320 Chr.8: 24466956-24466956 on Build GRCh38
rs778867805 Chr.8: 24466900-24466900 on Build GRCh38
rs200526194 Chr.8: 24466869-24466869 on Build GRCh38
rs761753009 Chr.8: 24466990-24466990 on Build GRCh38
rs376519230 Chr.8: 24466756-24466756 on Build GRCh38
rs559607040 Chr.8: 24466990-24467019 on Build GRCh38
rs770915822 Chr.8: 24466971-24466971 on Build GRCh38
rs752117930 Chr.8: 24466944-24466944 on Build GRCh38
rs771615867 Chr.8: 24466961-24466961 on Build GRCh38
rs918175439 Chr.8: 24467180-24467180 on Build GRCh38
rs911395416 Chr.8: 24466800-24466800 on Build GRCh38
rs527289130 Chr.8: 24467075-24467075 on Build GRCh38
rs531887202 Chr.8: 24467216-24467216 on Build GRCh38
rs201342977 Chr.8: 24466834-24466834 on Build GRCh38
rs773800358 Chr.8: 24466765-24466765 on Build GRCh38
rs781566017 Chr.8: 24466951-24466951 on Build GRCh38
rs751550309 Chr.8: 24467016-24467016 on Build GRCh38
rs751157751 Chr.8: 24466896-24466896 on Build GRCh38
rs773569621 Chr.8: 24467004-24467004 on Build GRCh38
rs905646423 Chr.8: 24467215-24467215 on Build GRCh38
rs763792277 Chr.8: 24466799-24466799 on Build GRCh38
rs113368922 Chr.8: 24467185-24467185 on Build GRCh38
rs770439108 Chr.8: 24466761-24466761 on Build GRCh38
rs1031446910 Chr.8: 24467071-24467071 on Build GRCh38
rs569204775 Chr.8: 24467179-24467179 on Build GRCh38
rs148882775 Chr.8: 24467111-24467111 on Build GRCh38
rs755190811 Chr.8: 24466821-24466823 on Build GRCh38
rs775266459 Chr.8: 24466788-24466788 on Build GRCh38
rs745709897 Chr.8: 24467029-24467029 on Build GRCh38
rs1005373029 Chr.8: 24466994-24466994 on Build GRCh38
rs745503970 Chr.8: 24466960-24466960 on Build GRCh38
rs552219254 Chr.8: 24467103-24467103 on Build GRCh38
rs750655080 Chr.8: 24466918-24466918 on Build GRCh38
rs776682016 Chr.8: 24466989-24466989 on Build GRCh38
rs755370535 Chr.8: 24466849-24466849 on Build GRCh38
rs746544036 Chr.8: 24467082-24467082 on Build GRCh38
rs1031415700 Chr.8: 24466808-24466808 on Build GRCh38
rs758568601 Chr.8: 24466927-24466927 on Build GRCh38
rs754367650 Chr.8: 24467132-24467132 on Build GRCh38
rs549789518 Chr.8: 24466980-24466980 on Build GRCh38
rs376371469 Chr.8: 24466993-24466993 on Build GRCh38
rs560051955 Chr.8: 24467074-24467074 on Build GRCh38
rs768263765 Chr.8: 24467083-24467083 on Build GRCh38
rs779649409 Chr.8: 24467081-24467081 on Build GRCh38
rs749494721 Chr.8: 24466758-24466758 on Build GRCh38
rs774807746 Chr.8: 24466965-24466965 on Build GRCh38
rs116973115 Chr.8: 24467128-24467128 on Build GRCh38
rs143068519 Chr.8: 24466817-24466817 on Build GRCh38
rs562817965 Chr.8: 24467195-24467195 on Build GRCh38
rs766298658 Chr.8: 24466930-24466930 on Build GRCh38
rs142174925 Chr.8: 24466959-24466959 on Build GRCh38
rs201646264 Chr.8: 24466891-24466891 on Build GRCh38
rs137855268 Chr.8: 24466806-24466806 on Build GRCh38
rs745430698 Chr.8: 24466886-24466886 on Build GRCh38
rs536229556 Chr.8: 24466772-24466772 on Build GRCh38
rs935545570 Chr.8: 24466835-24466835 on Build GRCh38
rs772981326 Chr.8: 24466752-24466752 on Build GRCh38
rs985474154 Chr.8: 24467084-24467084 on Build GRCh38
rs529899254 Chr.8: 24467213-24467213 on Build GRCh38
rs370524448 Chr.8: 24466876-24466876 on Build GRCh38
rs771469786 Chr.8: 24466887-24466887 on Build GRCh38
rs781340208 Chr.8: 24467092-24467092 on Build GRCh38
rs531493022 Chr.8: 24466829-24466829 on Build GRCh38
rs776521318 Chr.8: 24466898-24466898 on Build GRCh38
rs1052802267 Chr.8: 24466843-24466843 on Build GRCh38
rs748170542 Chr.8: 24466968-24466970 on Build GRCh38
rs534259238 Chr.8: 24466818-24466818 on Build GRCh38

View Full Product Details