Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24481955-24482473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM7
Entrez Gene ID
8756
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
ADAM 7, ADAM-7, EAPI, GP-83, GP83
UniGene
Hs.116147
Gene Chromosome Location
Chr.8: 24440996-24509565 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
142118 c.727C>T p.H243Y Chr.8: 24482163-24482163 on Build GRCh38
6593716 c.844C>T p.R282W Chr.8: 24482280-24482280 on Build GRCh38
6352522 c.730G>A p.V244M Chr.8: 24482166-24482166 on Build GRCh38
1098334 c.766G>T p.D256Y Chr.8: 24482202-24482202 on Build GRCh38
5596216 c.834C>T p.I278I Chr.8: 24482270-24482270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750423597 Chr.8: 24482226-24482226 on Build GRCh38
rs779521824 Chr.8: 24482262-24482262 on Build GRCh38
rs746265017 Chr.8: 24482276-24482276 on Build GRCh38
rs943752273 Chr.8: 24482039-24482039 on Build GRCh38
rs149396012 Chr.8: 24482189-24482189 on Build GRCh38
rs781236804 Chr.8: 24482170-24482170 on Build GRCh38
rs1053658077 Chr.8: 24482120-24482120 on Build GRCh38
rs768395546 Chr.8: 24482210-24482210 on Build GRCh38
rs772265231 Chr.8: 24482278-24482278 on Build GRCh38
rs945629795 Chr.8: 24482250-24482250 on Build GRCh38
rs199786542 Chr.8: 24482227-24482227 on Build GRCh38
rs142885831 Chr.8: 24481990-24481990 on Build GRCh38
rs201308689 Chr.8: 24482240-24482240 on Build GRCh38
rs774059964 Chr.8: 24482363-24482363 on Build GRCh38
rs760323611 Chr.8: 24482195-24482195 on Build GRCh38
rs761398118 Chr.8: 24482213-24482213 on Build GRCh38
rs751797225 Chr.8: 24482161-24482161 on Build GRCh38
rs4872240 Chr.8: 24482081-24482081 on Build GRCh38
rs771434508 Chr.8: 24482199-24482199 on Build GRCh38
rs772682709 Chr.8: 24482288-24482288 on Build GRCh38
rs976493816 Chr.8: 24482347-24482347 on Build GRCh38
rs760277054 Chr.8: 24482204-24482204 on Build GRCh38
rs753023284 Chr.8: 24482336-24482336 on Build GRCh38
rs11348969 Chr.8: 24482384-24482384 on Build GRCh38
rs895153995 Chr.8: 24482359-24482359 on Build GRCh38
rs778488740 Chr.8: 24482174-24482174 on Build GRCh38
rs763535548 Chr.8: 24482111-24482111 on Build GRCh38
rs774808165 Chr.8: 24482200-24482200 on Build GRCh38
rs13255694 Chr.8: 24482166-24482166 on Build GRCh38
rs774101592 Chr.8: 24482309-24482309 on Build GRCh38
rs367914998 Chr.8: 24482106-24482106 on Build GRCh38
rs147421261 Chr.8: 24482340-24482340 on Build GRCh38
rs779761107 Chr.8: 24482152-24482152 on Build GRCh38
rs548148422 Chr.8: 24482421-24482421 on Build GRCh38
rs79127838 Chr.8: 24482115-24482115 on Build GRCh38
rs754001688 Chr.8: 24482345-24482345 on Build GRCh38
rs1005515759 Chr.8: 24482367-24482368 on Build GRCh38
rs765209429 Chr.8: 24482129-24482129 on Build GRCh38
rs373522515 Chr.8: 24482237-24482237 on Build GRCh38
rs921135743 Chr.8: 24482078-24482078 on Build GRCh38
rs557551645 Chr.8: 24482245-24482245 on Build GRCh38
rs895346900 Chr.8: 24482234-24482234 on Build GRCh38
rs953892401 Chr.8: 24482388-24482388 on Build GRCh38
rs758258853 Chr.8: 24482149-24482149 on Build GRCh38
rs147649440 Chr.8: 24482171-24482171 on Build GRCh38
rs544890798 Chr.8: 24482368-24482368 on Build GRCh38
rs140597991 Chr.8: 24482182-24482182 on Build GRCh38
rs398086432 Chr.8: 24482390-24482390 on Build GRCh38
rs965011407 Chr.8: 24482325-24482325 on Build GRCh38
rs976464093 Chr.8: 24482048-24482048 on Build GRCh38
rs766206304 Chr.8: 24482229-24482229 on Build GRCh38
rs908290037 Chr.8: 24482017-24482017 on Build GRCh38
rs547306323 Chr.8: 24482220-24482220 on Build GRCh38
rs762414081 Chr.8: 24482294-24482294 on Build GRCh38
rs569773461 Chr.8: 24482279-24482279 on Build GRCh38
rs777987045 Chr.8: 24482244-24482244 on Build GRCh38
rs200376967 Chr.8: 24482172-24482172 on Build GRCh38
rs565468431 Chr.8: 24481991-24481991 on Build GRCh38
rs973676210 Chr.8: 24481987-24481987 on Build GRCh38
rs775285246 Chr.8: 24482099-24482099 on Build GRCh38
rs945250814 Chr.8: 24482409-24482409 on Build GRCh38
rs754683685 Chr.8: 24482236-24482236 on Build GRCh38
rs149722616 Chr.8: 24482280-24482280 on Build GRCh38
rs1042296661 Chr.8: 24482414-24482414 on Build GRCh38
rs963208101 Chr.8: 24482046-24482046 on Build GRCh38
rs756330227 Chr.8: 24482241-24482241 on Build GRCh38
rs761198575 Chr.8: 24482128-24482128 on Build GRCh38
rs180986443 Chr.8: 24482062-24482062 on Build GRCh38
rs375488680 Chr.8: 24482384-24482384 on Build GRCh38
rs757807464 Chr.8: 24482255-24482255 on Build GRCh38
rs545945948 Chr.8: 24482069-24482069 on Build GRCh38
rs530048532 Chr.8: 24482395-24482395 on Build GRCh38
rs910236169 Chr.8: 24482202-24482202 on Build GRCh38
rs893683580 Chr.8: 24482142-24482142 on Build GRCh38
rs989693878 Chr.8: 24482391-24482391 on Build GRCh38
rs756383329 Chr.8: 24482337-24482337 on Build GRCh38
rs749745197 Chr.8: 24482176-24482176 on Build GRCh38
rs747949424 Chr.8: 24482251-24482251 on Build GRCh38
rs751404831 Chr.8: 24482233-24482233 on Build GRCh38
rs141296535 Chr.8: 24481978-24481978 on Build GRCh38
rs769347524 Chr.8: 24482283-24482283 on Build GRCh38
rs750349883 Chr.8: 24482143-24482143 on Build GRCh38
rs575093318 Chr.8: 24482394-24482394 on Build GRCh38
rs766329095 Chr.8: 24482301-24482301 on Build GRCh38
rs370887425 Chr.8: 24482351-24482351 on Build GRCh38
rs529112526 Chr.8: 24482219-24482219 on Build GRCh38
rs906684540 Chr.8: 24482293-24482293 on Build GRCh38
rs987250499 Chr.8: 24482445-24482445 on Build GRCh38
rs542541711 Chr.8: 24482001-24482001 on Build GRCh38
rs776126909 Chr.8: 24482211-24482211 on Build GRCh38
rs560479929 Chr.8: 24482003-24482003 on Build GRCh38
rs145549411 Chr.8: 24482311-24482311 on Build GRCh38
rs575256369 Chr.8: 24482000-24482000 on Build GRCh38
rs1026170451 Chr.8: 24482417-24482417 on Build GRCh38

View Full Product Details