Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27460797-27461289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

SNP Detail

SNP ID Chromosome Location
rs925910890 Chr.8: 27460889-27460889 on Build GRCh38
rs940210331 Chr.8: 27461247-27461247 on Build GRCh38
rs2292974 Chr.8: 27460874-27460874 on Build GRCh38
rs2292976 Chr.8: 27461027-27461027 on Build GRCh38
rs909328944 Chr.8: 27461010-27461010 on Build GRCh38
rs181384212 Chr.8: 27460866-27460866 on Build GRCh38
rs752939421 Chr.8: 27461186-27461186 on Build GRCh38
rs991955767 Chr.8: 27460939-27460939 on Build GRCh38
rs76428534 Chr.8: 27461245-27461245 on Build GRCh38
rs542172546 Chr.8: 27461243-27461243 on Build GRCh38
rs2292975 Chr.8: 27460908-27460908 on Build GRCh38
rs919404477 Chr.8: 27460944-27460944 on Build GRCh38
rs1038996400 Chr.8: 27461254-27461254 on Build GRCh38
rs778798757 Chr.8: 27460831-27460831 on Build GRCh38
rs892850363 Chr.8: 27461065-27461065 on Build GRCh38
rs79796894 Chr.8: 27461006-27461006 on Build GRCh38
rs1008730716 Chr.8: 27461076-27461076 on Build GRCh38
rs943412642 Chr.8: 27461251-27461251 on Build GRCh38
rs570546296 Chr.8: 27461248-27461248 on Build GRCh38
rs1038593040 Chr.8: 27461012-27461012 on Build GRCh38
rs543706264 Chr.8: 27460977-27460977 on Build GRCh38
rs971107554 Chr.8: 27461106-27461106 on Build GRCh38
rs1012677814 Chr.8: 27461075-27461075 on Build GRCh38
rs977410907 Chr.8: 27460886-27460886 on Build GRCh38
rs2292977 Chr.8: 27461118-27461118 on Build GRCh38
rs768247154 Chr.8: 27460830-27460830 on Build GRCh38
rs898802412 Chr.8: 27461020-27461020 on Build GRCh38
rs940243341 Chr.8: 27461011-27461011 on Build GRCh38
rs548984913 Chr.8: 27461083-27461083 on Build GRCh38
rs574004327 Chr.8: 27461150-27461150 on Build GRCh38
rs1045916715 Chr.8: 27460980-27460980 on Build GRCh38
rs888892837 Chr.8: 27461057-27461057 on Build GRCh38
rs998681323 Chr.8: 27461045-27461045 on Build GRCh38
rs75002794 Chr.8: 27460993-27460993 on Build GRCh38
rs950852359 Chr.8: 27461220-27461220 on Build GRCh38
rs1050585345 Chr.8: 27461035-27461035 on Build GRCh38
rs77335612 Chr.8: 27461233-27461233 on Build GRCh38
rs971488359 Chr.8: 27461221-27461221 on Build GRCh38
rs1032752447 Chr.8: 27460862-27460862 on Build GRCh38
rs760687186 Chr.8: 27460961-27460961 on Build GRCh38
rs903011280 Chr.8: 27461041-27461041 on Build GRCh38
rs552243531 Chr.8: 27461249-27461249 on Build GRCh38
rs920217846 Chr.8: 27461257-27461257 on Build GRCh38
rs957591139 Chr.8: 27461162-27461162 on Build GRCh38
rs991531471 Chr.8: 27461182-27461182 on Build GRCh38
rs987330936 Chr.8: 27460852-27460852 on Build GRCh38
rs1032943133 Chr.8: 27461148-27461148 on Build GRCh38
rs773081362 Chr.8: 27460941-27460941 on Build GRCh38
rs998750870 Chr.8: 27461145-27461145 on Build GRCh38
rs528556599 Chr.8: 27461164-27461164 on Build GRCh38
rs572058986 Chr.8: 27460858-27460858 on Build GRCh38
rs1032721372 Chr.8: 27461048-27461048 on Build GRCh38
rs916029184 Chr.8: 27460940-27460940 on Build GRCh38
rs967209575 Chr.8: 27461136-27461136 on Build GRCh38

View Full Product Details