Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27467122-27467590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6276841 c.340-9C>A p.? Chr.8: 27467347-27467347 on Build GRCh38
3925095 c.395C>T p.S132F Chr.8: 27467283-27467283 on Build GRCh38
7180383 c.449+1G>A p.? Chr.8: 27467228-27467228 on Build GRCh38
3374885 c.380T>C p.F127S Chr.8: 27467298-27467298 on Build GRCh38
6312104 c.444C>A p.Y148* Chr.8: 27467234-27467234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017665537 Chr.8: 27467562-27467562 on Build GRCh38
rs748810147 Chr.8: 27467198-27467198 on Build GRCh38
rs758217117 Chr.8: 27467229-27467231 on Build GRCh38
rs751816159 Chr.8: 27467197-27467197 on Build GRCh38
rs746274421 Chr.8: 27467317-27467317 on Build GRCh38
rs759799198 Chr.8: 27467370-27467370 on Build GRCh38
rs773956983 Chr.8: 27467276-27467276 on Build GRCh38
rs995697228 Chr.8: 27467530-27467530 on Build GRCh38
rs374195332 Chr.8: 27467308-27467308 on Build GRCh38
rs750588205 Chr.8: 27467226-27467226 on Build GRCh38
rs374903443 Chr.8: 27467240-27467240 on Build GRCh38
rs752422797 Chr.8: 27467316-27467316 on Build GRCh38
rs780846003 Chr.8: 27467242-27467242 on Build GRCh38
rs769789191 Chr.8: 27467187-27467187 on Build GRCh38
rs778279138 Chr.8: 27467324-27467324 on Build GRCh38
rs191458051 Chr.8: 27467538-27467538 on Build GRCh38
rs773105816 Chr.8: 27467359-27467359 on Build GRCh38
rs767749199 Chr.8: 27467208-27467208 on Build GRCh38
rs541282922 Chr.8: 27467331-27467331 on Build GRCh38
rs948717378 Chr.8: 27467439-27467439 on Build GRCh38
rs748005557 Chr.8: 27467180-27467180 on Build GRCh38
rs557339825 Chr.8: 27467455-27467455 on Build GRCh38
rs368584036 Chr.8: 27467220-27467220 on Build GRCh38
rs1030301231 Chr.8: 27467145-27467145 on Build GRCh38
rs1044754212 Chr.8: 27467443-27467443 on Build GRCh38
rs775960558 Chr.8: 27467213-27467213 on Build GRCh38
rs559567251 Chr.8: 27467176-27467176 on Build GRCh38
rs548983616 Chr.8: 27467383-27467383 on Build GRCh38
rs373046238 Chr.8: 27467353-27467353 on Build GRCh38
rs890456672 Chr.8: 27467515-27467515 on Build GRCh38
rs200169352 Chr.8: 27467206-27467206 on Build GRCh38
rs758688566 Chr.8: 27467235-27467235 on Build GRCh38
rs745399034 Chr.8: 27467325-27467325 on Build GRCh38
rs145898029 Chr.8: 27467445-27467445 on Build GRCh38
rs777391476 Chr.8: 27467275-27467275 on Build GRCh38
rs761127621 Chr.8: 27467288-27467288 on Build GRCh38
rs556886679 Chr.8: 27467552-27467552 on Build GRCh38
rs924412195 Chr.8: 27467379-27467379 on Build GRCh38
rs749906968 Chr.8: 27467531-27467531 on Build GRCh38
rs779790900 Chr.8: 27467330-27467330 on Build GRCh38
rs990009490 Chr.8: 27467405-27467405 on Build GRCh38
rs891398 Chr.8: 27467305-27467305 on Build GRCh38
rs777304017 Chr.8: 27467298-27467298 on Build GRCh38
rs571461388 Chr.8: 27467441-27467441 on Build GRCh38
rs757256139 Chr.8: 27467323-27467323 on Build GRCh38
rs753696515 Chr.8: 27467377-27467377 on Build GRCh38
rs368477570 Chr.8: 27467354-27467354 on Build GRCh38
rs891676966 Chr.8: 27467544-27467544 on Build GRCh38
rs934556384 Chr.8: 27467150-27467150 on Build GRCh38
rs934525432 Chr.8: 27467403-27467403 on Build GRCh38
rs928273961 Chr.8: 27467436-27467436 on Build GRCh38
rs138682847 Chr.8: 27467295-27467295 on Build GRCh38
rs557478158 Chr.8: 27467435-27467435 on Build GRCh38
rs55698463 Chr.8: 27467407-27467407 on Build GRCh38
rs780763744 Chr.8: 27467367-27467367 on Build GRCh38
rs772332837 Chr.8: 27467175-27467175 on Build GRCh38
rs1054314849 Chr.8: 27467160-27467160 on Build GRCh38
rs1037809461 Chr.8: 27467475-27467475 on Build GRCh38
rs897313454 Chr.8: 27467518-27467518 on Build GRCh38
rs908082104 Chr.8: 27467442-27467442 on Build GRCh38
rs766634989 Chr.8: 27467315-27467315 on Build GRCh38
rs534405187 Chr.8: 27467409-27467409 on Build GRCh38
rs777177252 Chr.8: 27467332-27467332 on Build GRCh38
rs990354931 Chr.8: 27467378-27467378 on Build GRCh38
rs765451065 Chr.8: 27467223-27467223 on Build GRCh38
rs975875474 Chr.8: 27467356-27467356 on Build GRCh38
rs569942227 Chr.8: 27467319-27467319 on Build GRCh38
rs751801366 Chr.8: 27467369-27467369 on Build GRCh38
rs796052297 Chr.8: 27467246-27467246 on Build GRCh38
rs371067379 Chr.8: 27467248-27467248 on Build GRCh38
rs748755253 Chr.8: 27467261-27467261 on Build GRCh38
rs574300545 Chr.8: 27467503-27467503 on Build GRCh38
rs772694686 Chr.8: 27467196-27467196 on Build GRCh38
rs573914101 Chr.8: 27467148-27467148 on Build GRCh38
rs777692776 Chr.8: 27467249-27467249 on Build GRCh38
rs748032811 Chr.8: 27467247-27467247 on Build GRCh38
rs150112824 Chr.8: 27467277-27467277 on Build GRCh38
rs1037130126 Chr.8: 27467509-27467509 on Build GRCh38
rs763842246 Chr.8: 27467373-27467373 on Build GRCh38
rs765201434 Chr.8: 27467306-27467306 on Build GRCh38
rs553318792 Chr.8: 27467561-27467561 on Build GRCh38
rs770502863 Chr.8: 27467202-27467202 on Build GRCh38
rs758535216 Chr.8: 27467163-27467163 on Build GRCh38
rs775662796 Chr.8: 27467284-27467284 on Build GRCh38
rs758534127 Chr.8: 27467313-27467313 on Build GRCh38
rs770447351 Chr.8: 27467262-27467262 on Build GRCh38
rs190952749 Chr.8: 27467174-27467174 on Build GRCh38
rs2565061 Chr.8: 27467327-27467327 on Build GRCh38
rs763096433 Chr.8: 27467365-27467365 on Build GRCh38
rs989910987 Chr.8: 27467201-27467202 on Build GRCh38
rs1008135646 Chr.8: 27467329-27467329 on Build GRCh38
rs202012980 Chr.8: 27467217-27467217 on Build GRCh38
rs969773867 Chr.8: 27467408-27467408 on Build GRCh38
rs771821921 Chr.8: 27467278-27467278 on Build GRCh38
rs979874380 Chr.8: 27467425-27467425 on Build GRCh38
rs1048479171 Chr.8: 27467275-27467275 on Build GRCh38

View Full Product Details