Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144317702-144318192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGAT1
Entrez Gene ID
8694
Gene Name
diacylglycerol O-acyltransferase 1
Gene Aliases
ARAT, ARGP1, DGAT, DIAR7
UniGene
Hs.521954
Gene Chromosome Location
Chr.8: 144314583-144326919 on Build GRCh38
Transcripts
NM_012079.5 XM_011517356.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6440682 c.847C>T p.L283F Chr.8: 144317922-144317922 on Build GRCh38
1097203 c.821G>A p.R274Q Chr.8: 144317948-144317948 on Build GRCh38
4445143 c.811C>A p.P271T Chr.8: 144317958-144317958 on Build GRCh38
6841490 c.749G>A p.R250H Chr.8: 144318097-144318097 on Build GRCh38
5549329 c.714G>A p.P238P Chr.8: 144318132-144318132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893874730 Chr.8: 144318065-144318065 on Build GRCh38
rs782136428 Chr.8: 144317732-144317732 on Build GRCh38
rs782431268 Chr.8: 144317865-144317865 on Build GRCh38
rs782563878 Chr.8: 144317897-144317897 on Build GRCh38
rs1053882558 Chr.8: 144318064-144318064 on Build GRCh38
rs782535361 Chr.8: 144318032-144318032 on Build GRCh38
rs367844653 Chr.8: 144318115-144318115 on Build GRCh38
rs781907411 Chr.8: 144317737-144317737 on Build GRCh38
rs782101705 Chr.8: 144318078-144318078 on Build GRCh38
rs371388715 Chr.8: 144318069-144318069 on Build GRCh38
rs781982207 Chr.8: 144318080-144318080 on Build GRCh38
rs782758517 Chr.8: 144318030-144318030 on Build GRCh38
rs782370319 Chr.8: 144317729-144317729 on Build GRCh38
rs782608010 Chr.8: 144317868-144317868 on Build GRCh38
rs782077320 Chr.8: 144317831-144317832 on Build GRCh38
rs140478833 Chr.8: 144318152-144318152 on Build GRCh38
rs782501227 Chr.8: 144318006-144318006 on Build GRCh38
rs782452538 Chr.8: 144318166-144318166 on Build GRCh38
rs935852307 Chr.8: 144318140-144318140 on Build GRCh38
rs368923478 Chr.8: 144317974-144317974 on Build GRCh38
rs782395052 Chr.8: 144318063-144318063 on Build GRCh38
rs961402262 Chr.8: 144318108-144318108 on Build GRCh38
rs782614403 Chr.8: 144317811-144317811 on Build GRCh38
rs782569114 Chr.8: 144318109-144318109 on Build GRCh38
rs781798310 Chr.8: 144318165-144318165 on Build GRCh38
rs201160986 Chr.8: 144317846-144317846 on Build GRCh38
rs373467033 Chr.8: 144318133-144318133 on Build GRCh38
rs376345760 Chr.8: 144317728-144317728 on Build GRCh38
rs781870073 Chr.8: 144318031-144318031 on Build GRCh38
rs782740287 Chr.8: 144318149-144318149 on Build GRCh38
rs782343457 Chr.8: 144317901-144317901 on Build GRCh38
rs782804473 Chr.8: 144317957-144317957 on Build GRCh38
rs782702737 Chr.8: 144317948-144317948 on Build GRCh38
rs782082318 Chr.8: 144317830-144317830 on Build GRCh38
rs151256660 Chr.8: 144318126-144318126 on Build GRCh38
rs376031466 Chr.8: 144318075-144318075 on Build GRCh38
rs782238113 Chr.8: 144318121-144318121 on Build GRCh38
rs782275693 Chr.8: 144317877-144317877 on Build GRCh38
rs782148617 Chr.8: 144318155-144318155 on Build GRCh38
rs781945965 Chr.8: 144317727-144317727 on Build GRCh38
rs782820690 Chr.8: 144318159-144318159 on Build GRCh38
rs782511616 Chr.8: 144317873-144317873 on Build GRCh38
rs1044305222 Chr.8: 144318043-144318043 on Build GRCh38
rs782161523 Chr.8: 144318130-144318130 on Build GRCh38
rs375938839 Chr.8: 144317832-144317832 on Build GRCh38
rs782564453 Chr.8: 144317894-144317894 on Build GRCh38
rs376658941 Chr.8: 144318042-144318042 on Build GRCh38
rs782641257 Chr.8: 144317760-144317760 on Build GRCh38
rs782247930 Chr.8: 144317928-144317928 on Build GRCh38
rs781969331 Chr.8: 144317927-144317927 on Build GRCh38
rs781895334 Chr.8: 144318102-144318102 on Build GRCh38
rs782645151 Chr.8: 144317721-144317721 on Build GRCh38
rs113582880 Chr.8: 144317761-144317761 on Build GRCh38
rs191371257 Chr.8: 144318067-144318067 on Build GRCh38
rs782152356 Chr.8: 144317872-144317873 on Build GRCh38
rs149913927 Chr.8: 144317980-144317980 on Build GRCh38
rs527813714 Chr.8: 144318085-144318085 on Build GRCh38
rs782679330 Chr.8: 144317801-144317801 on Build GRCh38
rs939644432 Chr.8: 144317905-144317905 on Build GRCh38
rs782317402 Chr.8: 144317826-144317826 on Build GRCh38
rs782246517 Chr.8: 144318057-144318057 on Build GRCh38
rs373473438 Chr.8: 144317995-144317995 on Build GRCh38
rs146322937 Chr.8: 144318119-144318119 on Build GRCh38
rs782415001 Chr.8: 144317726-144317726 on Build GRCh38
rs781875216 Chr.8: 144318154-144318154 on Build GRCh38
rs782553235 Chr.8: 144317955-144317955 on Build GRCh38
rs782795347 Chr.8: 144317733-144317733 on Build GRCh38
rs781903129 Chr.8: 144317849-144317849 on Build GRCh38
rs782391605 Chr.8: 144318125-144318125 on Build GRCh38
rs781871790 Chr.8: 144318171-144318172 on Build GRCh38
rs141487799 Chr.8: 144318114-144318114 on Build GRCh38
rs200768512 Chr.8: 144318068-144318068 on Build GRCh38
rs781925298 Chr.8: 144318029-144318030 on Build GRCh38
rs372168844 Chr.8: 144317871-144317871 on Build GRCh38
rs782043842 Chr.8: 144318095-144318095 on Build GRCh38
rs781929904 Chr.8: 144318135-144318135 on Build GRCh38
rs782802515 Chr.8: 144318003-144318003 on Build GRCh38
rs781791106 Chr.8: 144317960-144317960 on Build GRCh38
rs782054104 Chr.8: 144317739-144317739 on Build GRCh38
rs200287520 Chr.8: 144318087-144318087 on Build GRCh38
rs781990889 Chr.8: 144317731-144317731 on Build GRCh38
rs781882807 Chr.8: 144317951-144317951 on Build GRCh38
rs782085129 Chr.8: 144318073-144318073 on Build GRCh38
rs782046312 Chr.8: 144318005-144318005 on Build GRCh38
rs782250375 Chr.8: 144317812-144317812 on Build GRCh38
rs782276475 Chr.8: 144317723-144317723 on Build GRCh38
rs782665180 Chr.8: 144317876-144317876 on Build GRCh38
rs543812582 Chr.8: 144318004-144318004 on Build GRCh38
rs144983092 Chr.8: 144317997-144317997 on Build GRCh38
rs782306228 Chr.8: 144317825-144317825 on Build GRCh38
rs373363244 Chr.8: 144317964-144317964 on Build GRCh38
rs782152635 Chr.8: 144317944-144317944 on Build GRCh38
rs146196839 Chr.8: 144317972-144317972 on Build GRCh38
rs55962377 Chr.8: 144318103-144318103 on Build GRCh38
rs139792712 Chr.8: 144317963-144317963 on Build GRCh38
rs781852964 Chr.8: 144317841-144317841 on Build GRCh38
rs781914459 Chr.8: 144317988-144317988 on Build GRCh38
rs782383355 Chr.8: 144317930-144317930 on Build GRCh38
rs781822558 Chr.8: 144318015-144318015 on Build GRCh38
rs188787685 Chr.8: 144317744-144317744 on Build GRCh38
rs781961359 Chr.8: 144317824-144317825 on Build GRCh38
rs782141799 Chr.8: 144317998-144317998 on Build GRCh38
rs1036959933 Chr.8: 144317909-144317909 on Build GRCh38
rs148665132 Chr.8: 144318093-144318093 on Build GRCh38
rs782202977 Chr.8: 144317899-144317899 on Build GRCh38
rs141523495 Chr.8: 144317967-144317967 on Build GRCh38
rs782495198 Chr.8: 144318021-144318021 on Build GRCh38
rs370182734 Chr.8: 144318148-144318148 on Build GRCh38
rs901153033 Chr.8: 144318077-144318077 on Build GRCh38
rs376764170 Chr.8: 144317943-144317943 on Build GRCh38
rs139081729 Chr.8: 144317977-144317977 on Build GRCh38
rs56408073 Chr.8: 144317749-144317749 on Build GRCh38
rs782078569 Chr.8: 144318142-144318142 on Build GRCh38
rs863225093 Chr.8: 144317794-144317794 on Build GRCh38
rs1044230336 Chr.8: 144317942-144317942 on Build GRCh38
rs782658376 Chr.8: 144317893-144317893 on Build GRCh38
rs782127538 Chr.8: 144317842-144317842 on Build GRCh38
rs782349298 Chr.8: 144317810-144317810 on Build GRCh38
rs146155230 Chr.8: 144317950-144317950 on Build GRCh38
rs782528548 Chr.8: 144317780-144317780 on Build GRCh38
rs371214641 Chr.8: 144318052-144318052 on Build GRCh38
rs782187728 Chr.8: 144317990-144317990 on Build GRCh38
rs782187957 Chr.8: 144318097-144318097 on Build GRCh38
rs782176186 Chr.8: 144317805-144317805 on Build GRCh38
rs782478665 Chr.8: 144317748-144317748 on Build GRCh38
rs570222469 Chr.8: 144317971-144317971 on Build GRCh38
rs782707652 Chr.8: 144318096-144318096 on Build GRCh38
rs782703963 Chr.8: 144317741-144317741 on Build GRCh38
rs782676187 Chr.8: 144318036-144318036 on Build GRCh38

View Full Product Details