Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6532765-6533273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
UniGene
Hs.584238
Gene Chromosome Location
Chr.9: 6532464-6645692 on Build GRCh38
Transcripts
NM_000170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1490060 c.3032_3033insCC p.F1012fs*>10 Chr.9: 6533047-6533048 on Build GRCh38
6671424 c.3054G>A p.A1018A Chr.9: 6533026-6533026 on Build GRCh38
4687092 c.3003C>T p.T1001T Chr.9: 6533077-6533077 on Build GRCh38
1176675 c.2937G>T p.E979D Chr.9: 6533143-6533143 on Build GRCh38
7288793 c.2996T>A p.V999D Chr.9: 6533084-6533084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942841834 Chr.9: 6532848-6532848 on Build GRCh38
rs142004524 Chr.9: 6533125-6533125 on Build GRCh38
rs772574530 Chr.9: 6533142-6533142 on Build GRCh38
rs533019569 Chr.9: 6533022-6533022 on Build GRCh38
rs112299567 Chr.9: 6533061-6533061 on Build GRCh38
rs369837674 Chr.9: 6533126-6533126 on Build GRCh38
rs553188892 Chr.9: 6532816-6532816 on Build GRCh38
rs543333246 Chr.9: 6532921-6532921 on Build GRCh38
rs374892231 Chr.9: 6533210-6533210 on Build GRCh38
rs777045138 Chr.9: 6533156-6533156 on Build GRCh38
rs1053035680 Chr.9: 6532891-6532891 on Build GRCh38
rs189753047 Chr.9: 6532979-6532979 on Build GRCh38
rs749409925 Chr.9: 6533026-6533026 on Build GRCh38
rs1019872837 Chr.9: 6533028-6533028 on Build GRCh38
rs200053325 Chr.9: 6533018-6533018 on Build GRCh38
rs762513013 Chr.9: 6533184-6533184 on Build GRCh38
rs867198626 Chr.9: 6532951-6532951 on Build GRCh38
rs949553434 Chr.9: 6533160-6533160 on Build GRCh38
rs753397431 Chr.9: 6533006-6533006 on Build GRCh38
rs932008577 Chr.9: 6532849-6532849 on Build GRCh38
rs764113678 Chr.9: 6533195-6533195 on Build GRCh38
rs148273307 Chr.9: 6532902-6532902 on Build GRCh38
rs999601385 Chr.9: 6532859-6532859 on Build GRCh38
rs140028660 Chr.9: 6533124-6533124 on Build GRCh38
rs774386131 Chr.9: 6533036-6533036 on Build GRCh38
rs758421388 Chr.9: 6533109-6533109 on Build GRCh38
rs912371579 Chr.9: 6532808-6532808 on Build GRCh38
rs369026023 Chr.9: 6533211-6533211 on Build GRCh38
rs758402823 Chr.9: 6533013-6533013 on Build GRCh38
rs940369337 Chr.9: 6532798-6532798 on Build GRCh38
rs951314581 Chr.9: 6532790-6532790 on Build GRCh38
rs759229432 Chr.9: 6533086-6533086 on Build GRCh38
rs1011300122 Chr.9: 6533232-6533232 on Build GRCh38
rs528875416 Chr.9: 6532938-6532938 on Build GRCh38
rs555776146 Chr.9: 6533078-6533078 on Build GRCh38
rs765132272 Chr.9: 6532980-6532980 on Build GRCh38
rs573934284 Chr.9: 6532801-6532801 on Build GRCh38
rs762287499 Chr.9: 6532966-6532966 on Build GRCh38
rs541456045 Chr.9: 6532804-6532804 on Build GRCh38
rs751846463 Chr.9: 6533073-6533073 on Build GRCh38
rs752239336 Chr.9: 6533105-6533105 on Build GRCh38
rs760781247 Chr.9: 6533185-6533185 on Build GRCh38
rs764054067 Chr.9: 6533110-6533110 on Build GRCh38
rs201262788 Chr.9: 6533205-6533205 on Build GRCh38
rs761780200 Chr.9: 6533204-6533204 on Build GRCh38
rs776192576 Chr.9: 6533081-6533081 on Build GRCh38
rs757398632 Chr.9: 6533089-6533089 on Build GRCh38
rs1044876419 Chr.9: 6532925-6532925 on Build GRCh38
rs760437541 Chr.9: 6532995-6532995 on Build GRCh38
rs904923660 Chr.9: 6532953-6532953 on Build GRCh38
rs775143393 Chr.9: 6533178-6533178 on Build GRCh38
rs976174465 Chr.9: 6532827-6532827 on Build GRCh38
rs749512886 Chr.9: 6533117-6533117 on Build GRCh38
rs751400703 Chr.9: 6533112-6533112 on Build GRCh38
rs577665662 Chr.9: 6532829-6532829 on Build GRCh38
rs777768041 Chr.9: 6533014-6533014 on Build GRCh38
rs1034808665 Chr.9: 6533032-6533032 on Build GRCh38
rs1049111024 Chr.9: 6532828-6532828 on Build GRCh38
rs773562990 Chr.9: 6532991-6532991 on Build GRCh38
rs765894083 Chr.9: 6533000-6533000 on Build GRCh38
rs893779931 Chr.9: 6532958-6532958 on Build GRCh38
rs550514750 Chr.9: 6533144-6533144 on Build GRCh38
rs935574554 Chr.9: 6532861-6532861 on Build GRCh38
rs576854663 Chr.9: 6533015-6533015 on Build GRCh38
rs372606760 Chr.9: 6533209-6533209 on Build GRCh38
rs747426447 Chr.9: 6533147-6533147 on Build GRCh38
rs570239401 Chr.9: 6533053-6533053 on Build GRCh38
rs150149798 Chr.9: 6533027-6533027 on Build GRCh38
rs1043696226 Chr.9: 6532850-6532850 on Build GRCh38
rs2228098 Chr.9: 6533010-6533010 on Build GRCh38
rs751501264 Chr.9: 6533198-6533198 on Build GRCh38
rs773642256 Chr.9: 6533070-6533070 on Build GRCh38
rs138786708 Chr.9: 6533168-6533168 on Build GRCh38
rs764814250 Chr.9: 6533012-6533012 on Build GRCh38
rs886741775 Chr.9: 6533238-6533238 on Build GRCh38
rs150624881 Chr.9: 6533154-6533154 on Build GRCh38
rs200397751 Chr.9: 6533172-6533172 on Build GRCh38
rs772557873 Chr.9: 6533069-6533069 on Build GRCh38
rs192587467 Chr.9: 6533065-6533065 on Build GRCh38
rs146045718 Chr.9: 6533116-6533116 on Build GRCh38
rs1003297138 Chr.9: 6533021-6533021 on Build GRCh38
rs12350734 Chr.9: 6532855-6532855 on Build GRCh38
rs951774955 Chr.9: 6533113-6533113 on Build GRCh38
rs1044996669 Chr.9: 6533233-6533233 on Build GRCh38
rs1052505781 Chr.9: 6533155-6533155 on Build GRCh38
rs925268812 Chr.9: 6533120-6533120 on Build GRCh38
rs562050625 Chr.9: 6532936-6532936 on Build GRCh38
rs975199715 Chr.9: 6533077-6533077 on Build GRCh38
rs149336868 Chr.9: 6533114-6533114 on Build GRCh38
rs1013753804 Chr.9: 6532924-6532924 on Build GRCh38
rs138640017 Chr.9: 6533092-6533092 on Build GRCh38
rs769512301 Chr.9: 6533174-6533174 on Build GRCh38
rs557062703 Chr.9: 6533176-6533176 on Build GRCh38
rs1057515605 Chr.9: 6533133-6533133 on Build GRCh38
rs545105557 Chr.9: 6532835-6532835 on Build GRCh38
rs995199042 Chr.9: 6532963-6532963 on Build GRCh38
rs971738825 Chr.9: 6532809-6532809 on Build GRCh38
rs184498089 Chr.9: 6532788-6532788 on Build GRCh38
rs760263601 Chr.9: 6532899-6532899 on Build GRCh38

View Full Product Details